Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32972728-32972965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD2
Entrez Gene ID
6046
Gene Name
bromodomain containing 2
Gene Aliases
D6S113E, FSH, FSRG1, NAT, O27.1.1, RING3, RNF3
UniGene
Hs.75243
Gene Chromosome Location
Chr.6: 32968660-32981505 on Build GRCh38
Transcripts
NM_001113182.2 NM_001199455.1 NM_001291986.1 NM_005104.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1077922 c.29+1G>A p.? Chr.6: 32972928-32972928 on Build GRCh38
5561321 c.25A>G p.N9D Chr.6: 32972923-32972923 on Build GRCh38
6403582 c.12C>A p.N4K Chr.6: 32972910-32972910 on Build GRCh38
1621496 c.14T>A p.V5E Chr.6: 32972912-32972912 on Build GRCh38
6845397 c.29A>C p.K10T Chr.6: 32972927-32972927 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757288990 Chr.6: 32972857-32972857 on Build GRCh38
rs558971120 Chr.6: 32972804-32972804 on Build GRCh38
rs142520217 Chr.6: 32972918-32972918 on Build GRCh38
rs2071571 Chr.6: 32972844-32972844 on Build GRCh38
rs919945373 Chr.6: 32972799-32972799 on Build GRCh38
rs768407759 Chr.6: 32972930-32972930 on Build GRCh38
rs767346915 Chr.6: 32972859-32972859 on Build GRCh38
rs576387027 Chr.6: 32972796-32972796 on Build GRCh38
rs973834011 Chr.6: 32972800-32972800 on Build GRCh38
rs367803227 Chr.6: 32972890-32972890 on Build GRCh38
rs776972257 Chr.6: 32972877-32972877 on Build GRCh38
rs200590068 Chr.6: 32972812-32972812 on Build GRCh38
rs777834793 Chr.6: 32972915-32972915 on Build GRCh38
rs775207207 Chr.6: 32972923-32972923 on Build GRCh38
rs752766238 Chr.6: 32972913-32972913 on Build GRCh38
rs959266138 Chr.6: 32972814-32972814 on Build GRCh38
rs750308944 Chr.6: 32972902-32972902 on Build GRCh38
rs900023500 Chr.6: 32972757-32972757 on Build GRCh38
rs917820303 Chr.6: 32972828-32972828 on Build GRCh38
rs942771943 Chr.6: 32972794-32972794 on Build GRCh38
rs766257762 Chr.6: 32972889-32972890 on Build GRCh38
rs1013918914 Chr.6: 32972830-32972830 on Build GRCh38
rs569785807 Chr.6: 32972935-32972935 on Build GRCh38
rs199876176 Chr.6: 32972931-32972931 on Build GRCh38
rs769381730 Chr.6: 32972884-32972884 on Build GRCh38
rs758763027 Chr.6: 32972866-32972866 on Build GRCh38
rs992416863 Chr.6: 32972822-32972822 on Build GRCh38
rs763892067 Chr.6: 32972854-32972854 on Build GRCh38
rs912712229 Chr.6: 32972780-32972780 on Build GRCh38
rs11555089 Chr.6: 32972827-32972827 on Build GRCh38
rs778267283 Chr.6: 32972868-32972868 on Build GRCh38
rs373976120 Chr.6: 32972889-32972889 on Build GRCh38
rs991968769 Chr.6: 32972936-32972936 on Build GRCh38
rs147219068 Chr.6: 32972910-32972910 on Build GRCh38
rs554732133 Chr.6: 32972747-32972747 on Build GRCh38
rs763653224 Chr.6: 32972892-32972892 on Build GRCh38
rs962429747 Chr.6: 32972798-32972799 on Build GRCh38
rs370485049 Chr.6: 32972862-32972862 on Build GRCh38
rs926441150 Chr.6: 32972809-32972809 on Build GRCh38
rs6916791 Chr.6: 32972840-32972840 on Build GRCh38
rs201319601 Chr.6: 32972921-32972921 on Build GRCh38
rs78280497 Chr.6: 32972777-32972777 on Build GRCh38
rs748307554 Chr.6: 32972863-32972863 on Build GRCh38
rs1054623729 Chr.6: 32972752-32972752 on Build GRCh38
rs776044199 Chr.6: 32972781-32972781 on Build GRCh38
rs201642612 Chr.6: 32972895-32972895 on Build GRCh38
rs769119110 Chr.6: 32972849-32972849 on Build GRCh38
rs11555088 Chr.6: 32972775-32972775 on Build GRCh38
rs574317080 Chr.6: 32972806-32972806 on Build GRCh38
rs773135501 Chr.6: 32972939-32972939 on Build GRCh38
rs761492965 Chr.6: 32972896-32972896 on Build GRCh38
rs552660151 Chr.6: 32972845-32972845 on Build GRCh38
rs913511389 Chr.6: 32972765-32972765 on Build GRCh38
rs140566548 Chr.6: 32972917-32972917 on Build GRCh38
rs775295477 Chr.6: 32972887-32972887 on Build GRCh38
rs745403599 Chr.6: 32972882-32972882 on Build GRCh38
rs377684930 Chr.6: 32972920-32972920 on Build GRCh38
rs540613957 Chr.6: 32972797-32972797 on Build GRCh38
rs760592838 Chr.6: 32972880-32972881 on Build GRCh38
rs767534205 Chr.6: 32972898-32972898 on Build GRCh38
rs957770050 Chr.6: 32972867-32972867 on Build GRCh38
rs771703104 Chr.6: 32972937-32972937 on Build GRCh38
rs554586362 Chr.6: 32972922-32972922 on Build GRCh38
rs748797118 Chr.6: 32972925-32972925 on Build GRCh38
rs931275772 Chr.6: 32972807-32972807 on Build GRCh38
rs368022125 Chr.6: 32972855-32972855 on Build GRCh38
rs201899070 Chr.6: 32972813-32972813 on Build GRCh38
rs771515658 Chr.6: 32972874-32972874 on Build GRCh38
rs995206152 Chr.6: 32972919-32972921 on Build GRCh38
rs955831966 Chr.6: 32972782-32972782 on Build GRCh38
rs564641147 Chr.6: 32972784-32972784 on Build GRCh38
rs747322559 Chr.6: 32972872-32972872 on Build GRCh38
rs570969275 Chr.6: 32972861-32972861 on Build GRCh38
rs758704519 Chr.6: 32972914-32972914 on Build GRCh38
rs765229853 Chr.6: 32972912-32972912 on Build GRCh38

View Full Product Details