Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34623638-34623911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID3C
Entrez Gene ID
138715
Gene Name
AT-rich interaction domain 3C
Gene Aliases
-
UniGene
Hs.534549
Gene Chromosome Location
Chr.9: 34621360-34633112 on Build GRCh38
Transcripts
NM_001017363.1 XM_017014283.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4645681 c.646G>A p.A216T Chr.9: 34623644-34623644 on Build GRCh38
7221089 c.576-7C>T p.? Chr.9: 34623721-34623721 on Build GRCh38
6536759 c.607G>C p.E203Q Chr.9: 34623683-34623683 on Build GRCh38
3907038 c.568C>T p.R190C Chr.9: 34623871-34623871 on Build GRCh38
5989272 c.556G>T p.A186S Chr.9: 34623883-34623883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974256657 Chr.9: 34623725-34623725 on Build GRCh38
rs758739197 Chr.9: 34623871-34623871 on Build GRCh38
rs925748238 Chr.9: 34623805-34623805 on Build GRCh38
rs754225762 Chr.9: 34623696-34623696 on Build GRCh38
rs764865701 Chr.9: 34623662-34623662 on Build GRCh38
rs751024693 Chr.9: 34623689-34623689 on Build GRCh38
rs754386668 Chr.9: 34623851-34623851 on Build GRCh38
rs778108570 Chr.9: 34623876-34623876 on Build GRCh38
rs866952933 Chr.9: 34623677-34623677 on Build GRCh38
rs766858093 Chr.9: 34623827-34623827 on Build GRCh38
rs1055660969 Chr.9: 34623819-34623819 on Build GRCh38
rs745850068 Chr.9: 34623893-34623893 on Build GRCh38
rs745706625 Chr.9: 34623747-34623747 on Build GRCh38
rs937144327 Chr.9: 34623806-34623806 on Build GRCh38
rs527399007 Chr.9: 34623779-34623779 on Build GRCh38
rs756664788 Chr.9: 34623690-34623690 on Build GRCh38
rs775193878 Chr.9: 34623667-34623667 on Build GRCh38
rs771031503 Chr.9: 34623728-34623728 on Build GRCh38
rs761310010 Chr.9: 34623825-34623825 on Build GRCh38
rs774023997 Chr.9: 34623734-34623734 on Build GRCh38
rs376209558 Chr.9: 34623889-34623889 on Build GRCh38
rs755360459 Chr.9: 34623699-34623699 on Build GRCh38
rs751753753 Chr.9: 34623883-34623883 on Build GRCh38
rs544991892 Chr.9: 34623695-34623695 on Build GRCh38
rs532640422 Chr.9: 34623674-34623674 on Build GRCh38
rs149879108 Chr.9: 34623692-34623692 on Build GRCh38
rs10814129 Chr.9: 34623721-34623721 on Build GRCh38
rs778018688 Chr.9: 34623719-34623719 on Build GRCh38
rs762617594 Chr.9: 34623813-34623813 on Build GRCh38
rs758649179 Chr.9: 34623710-34623710 on Build GRCh38
rs757057977 Chr.9: 34623694-34623694 on Build GRCh38
rs759342451 Chr.9: 34623658-34623658 on Build GRCh38
rs550487304 Chr.9: 34623788-34623788 on Build GRCh38
rs968085346 Chr.9: 34623783-34623783 on Build GRCh38
rs775108204 Chr.9: 34623758-34623758 on Build GRCh38
rs748382089 Chr.9: 34623700-34623700 on Build GRCh38
rs753059376 Chr.9: 34623869-34623869 on Build GRCh38
rs777370297 Chr.9: 34623690-34623695 on Build GRCh38
rs763625725 Chr.9: 34623686-34623686 on Build GRCh38
rs759890577 Chr.9: 34623863-34623863 on Build GRCh38
rs892077792 Chr.9: 34623800-34623800 on Build GRCh38
rs751374358 Chr.9: 34623689-34623690 on Build GRCh38
rs1031037371 Chr.9: 34623693-34623693 on Build GRCh38
rs765660656 Chr.9: 34623867-34623867 on Build GRCh38
rs757346189 Chr.9: 34623886-34623886 on Build GRCh38
rs768276005 Chr.9: 34623820-34623820 on Build GRCh38
rs776624949 Chr.9: 34623730-34623730 on Build GRCh38
rs569247402 Chr.9: 34623744-34623744 on Build GRCh38
rs769636179 Chr.9: 34623757-34623757 on Build GRCh38

View Full Product Details