Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 107610157-107610406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD11
Entrez Gene ID
121551
Gene Name
BTB domain containing 11
Gene Aliases
ABTB2B
UniGene
Hs.271272
Gene Chromosome Location
Chr.12: 107317168-107659642 on Build GRCh38
Transcripts
NM_001017523.1 NM_001018072.1 XM_005268645.3 XM_011537909.2 XM_011537910.2 XM_017018807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1185016 c.1682C>T p.A561V Chr.12: 107610228-107610228 on Build GRCh38
6613305 c.353G>A p.R118Q Chr.12: 107610288-107610288 on Build GRCh38
5965119 c.1769C>T p.S590F Chr.12: 107610315-107610315 on Build GRCh38
5775716 c.352C>T p.R118* Chr.12: 107610287-107610287 on Build GRCh38
6579041 c.336G>A p.R112R Chr.12: 107610271-107610271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770851833 Chr.12: 107610270-107610270 on Build GRCh38
rs551383603 Chr.12: 107610196-107610196 on Build GRCh38
rs927584618 Chr.12: 107610369-107610369 on Build GRCh38
rs145154300 Chr.12: 107610275-107610275 on Build GRCh38
rs779619469 Chr.12: 107610208-107610208 on Build GRCh38
rs766719761 Chr.12: 107610345-107610345 on Build GRCh38
rs144974823 Chr.12: 107610326-107610326 on Build GRCh38
rs754687077 Chr.12: 107610229-107610229 on Build GRCh38
rs780850772 Chr.12: 107610365-107610365 on Build GRCh38
rs972267016 Chr.12: 107610315-107610315 on Build GRCh38
rs751966736 Chr.12: 107610349-107610349 on Build GRCh38
rs750705568 Chr.12: 107610197-107610197 on Build GRCh38
rs947774241 Chr.12: 107610346-107610346 on Build GRCh38
rs149087396 Chr.12: 107610328-107610328 on Build GRCh38
rs780656543 Chr.12: 107610232-107610232 on Build GRCh38
rs777195666 Chr.12: 107610193-107610193 on Build GRCh38
rs921242352 Chr.12: 107610367-107610367 on Build GRCh38
rs758807119 Chr.12: 107610206-107610206 on Build GRCh38
rs777505296 Chr.12: 107610381-107610381 on Build GRCh38
rs370299158 Chr.12: 107610385-107610385 on Build GRCh38
rs916117649 Chr.12: 107610337-107610337 on Build GRCh38
rs773222745 Chr.12: 107610239-107610239 on Build GRCh38
rs373253511 Chr.12: 107610344-107610344 on Build GRCh38
rs953321307 Chr.12: 107610379-107610379 on Build GRCh38
rs1018888722 Chr.12: 107610269-107610269 on Build GRCh38
rs376237566 Chr.12: 107610366-107610366 on Build GRCh38
rs200134340 Chr.12: 107610238-107610238 on Build GRCh38
rs142214499 Chr.12: 107610329-107610329 on Build GRCh38
rs61747550 Chr.12: 107610260-107610260 on Build GRCh38
rs750948386 Chr.12: 107610341-107610341 on Build GRCh38
rs764264260 Chr.12: 107610191-107610191 on Build GRCh38
rs760959109 Chr.12: 107610314-107610314 on Build GRCh38
rs754591296 Chr.12: 107610363-107610363 on Build GRCh38
rs765608294 Chr.12: 107610331-107610331 on Build GRCh38
rs200596550 Chr.12: 107610209-107610209 on Build GRCh38

View Full Product Details