Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49689987-49690216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADM5
Entrez Gene ID
199800
Gene Name
adrenomedullin 5 (putative)
Gene Aliases
AM5, C19orf76
UniGene
Hs.311193
Gene Chromosome Location
Chr.19: 49688685-49690990 on Build GRCh38
Transcripts
NM_001101340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5390319 c.95C>T p.P32L Chr.19: 49690126-49690126 on Build GRCh38
439994 c.77G>A p.R26Q Chr.19: 49690108-49690108 on Build GRCh38
3362955 c.149C>A p.A50E Chr.19: 49690180-49690180 on Build GRCh38
5957358 c.167T>C p.I56T Chr.19: 49690198-49690198 on Build GRCh38
4667202 c.112G>A p.V38I Chr.19: 49690143-49690143 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs984130220 Chr.19: 49690119-49690119 on Build GRCh38
rs865921197 Chr.19: 49690158-49690158 on Build GRCh38
rs45613034 Chr.19: 49690180-49690180 on Build GRCh38
rs76092430 Chr.19: 49690107-49690107 on Build GRCh38
rs79588684 Chr.19: 49690110-49690110 on Build GRCh38
rs752846646 Chr.19: 49690056-49690056 on Build GRCh38
rs776597429 Chr.19: 49690178-49690178 on Build GRCh38
rs2288919 Chr.19: 49690174-49690174 on Build GRCh38
rs929276365 Chr.19: 49690112-49690112 on Build GRCh38
rs76964367 Chr.19: 49690109-49690109 on Build GRCh38
rs968995732 Chr.19: 49690015-49690015 on Build GRCh38
rs746551412 Chr.19: 49690168-49690168 on Build GRCh38
rs563358995 Chr.19: 49690067-49690067 on Build GRCh38
rs927816680 Chr.19: 49690031-49690031 on Build GRCh38
rs545215438 Chr.19: 49690051-49690051 on Build GRCh38
rs751151826 Chr.19: 49690137-49690137 on Build GRCh38
rs546582373 Chr.19: 49690195-49690195 on Build GRCh38
rs757623949 Chr.19: 49690103-49690103 on Build GRCh38
rs993340452 Chr.19: 49690094-49690094 on Build GRCh38
rs369177737 Chr.19: 49690184-49690184 on Build GRCh38
rs767812493 Chr.19: 49690193-49690193 on Build GRCh38
rs756989881 Chr.19: 49690144-49690144 on Build GRCh38
rs769542749 Chr.19: 49690181-49690181 on Build GRCh38
rs969317754 Chr.19: 49690010-49690010 on Build GRCh38
rs764738826 Chr.19: 49690093-49690093 on Build GRCh38
rs754375777 Chr.19: 49690100-49690100 on Build GRCh38
rs756223730 Chr.19: 49690065-49690065 on Build GRCh38
rs779335298 Chr.19: 49690118-49690118 on Build GRCh38
rs779520761 Chr.19: 49690141-49690141 on Build GRCh38
rs373026253 Chr.19: 49690127-49690127 on Build GRCh38
rs549073119 Chr.19: 49690136-49690136 on Build GRCh38
rs985303776 Chr.19: 49690032-49690032 on Build GRCh38
rs868071671 Chr.19: 49690196-49690196 on Build GRCh38
rs762608036 Chr.19: 49690190-49690190 on Build GRCh38
rs866935616 Chr.19: 49690128-49690128 on Build GRCh38

View Full Product Details