Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158292207-158292436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD1B
Entrez Gene ID
910
Gene Name
CD1b molecule
Gene Aliases
CD1, CD1A, R1
UniGene
Hs.1310
Gene Chromosome Location
Chr.1: 158286841-158332038 on Build GRCh38
Transcripts
XM_017002785.1 NM_001765.2 XM_005245579.4

Gene Symbol
CD1C
Entrez Gene ID
911
Gene Name
CD1c molecule
Gene Aliases
BDCA1, CD1, CD1A, R7
UniGene
Hs.132448
Gene Chromosome Location
Chr.1: 158289773-158294774 on Build GRCh38
Transcripts
XM_017002785.1 NM_001765.2 XM_005245579.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5823912 c.508T>A p.Y170N Chr.1: 158292263-158292263 on Build GRCh38
1688838 c.511G>A p.E171K Chr.1: 158292266-158292266 on Build GRCh38
3418174 c.455C>A p.S152Y Chr.1: 158292210-158292210 on Build GRCh38
3360418 c.516C>T p.G172G Chr.1: 158292271-158292271 on Build GRCh38
87744 c.594G>A p.M198I Chr.1: 158292349-158292349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762541123 Chr.1: 158292302-158292302 on Build GRCh38
rs756972442 Chr.1: 158292347-158292347 on Build GRCh38
rs775092697 Chr.1: 158292402-158292402 on Build GRCh38
rs912254103 Chr.1: 158292297-158292297 on Build GRCh38
rs905216602 Chr.1: 158292270-158292270 on Build GRCh38
rs748673410 Chr.1: 158292357-158292357 on Build GRCh38
rs142044323 Chr.1: 158292271-158292271 on Build GRCh38
rs760726167 Chr.1: 158292407-158292407 on Build GRCh38
rs767151325 Chr.1: 158292399-158292399 on Build GRCh38
rs568774285 Chr.1: 158292334-158292334 on Build GRCh38
rs377534018 Chr.1: 158292313-158292313 on Build GRCh38
rs751303260 Chr.1: 158292242-158292243 on Build GRCh38
rs768705274 Chr.1: 158292281-158292281 on Build GRCh38
rs548847032 Chr.1: 158292328-158292328 on Build GRCh38
rs781184826 Chr.1: 158292353-158292353 on Build GRCh38
rs535534188 Chr.1: 158292310-158292310 on Build GRCh38
rs564937461 Chr.1: 158292291-158292291 on Build GRCh38
rs771185947 Chr.1: 158292387-158292387 on Build GRCh38
rs368552932 Chr.1: 158292244-158292244 on Build GRCh38
rs763819494 Chr.1: 158292329-158292329 on Build GRCh38
rs755119731 Chr.1: 158292352-158292352 on Build GRCh38
rs777559090 Chr.1: 158292233-158292233 on Build GRCh38
rs1029113591 Chr.1: 158292269-158292269 on Build GRCh38
rs551035923 Chr.1: 158292381-158292381 on Build GRCh38
rs965671764 Chr.1: 158292322-158292322 on Build GRCh38
rs139538152 Chr.1: 158292314-158292314 on Build GRCh38
rs1045058762 Chr.1: 158292239-158292239 on Build GRCh38
rs1053652004 Chr.1: 158292415-158292415 on Build GRCh38
rs747223540 Chr.1: 158292351-158292351 on Build GRCh38
rs773574801 Chr.1: 158292296-158292296 on Build GRCh38
rs764939458 Chr.1: 158292414-158292414 on Build GRCh38
rs770293197 Chr.1: 158292362-158292362 on Build GRCh38
rs773557839 Chr.1: 158292379-158292379 on Build GRCh38
rs759928678 Chr.1: 158292315-158292315 on Build GRCh38
rs766092724 Chr.1: 158292305-158292305 on Build GRCh38
rs146158051 Chr.1: 158292272-158292272 on Build GRCh38
rs763083931 Chr.1: 158292312-158292312 on Build GRCh38
rs550882869 Chr.1: 158292323-158292323 on Build GRCh38
rs768687130 Chr.1: 158292408-158292408 on Build GRCh38
rs753700863 Chr.1: 158292413-158292413 on Build GRCh38
rs3138102 Chr.1: 158292410-158292410 on Build GRCh38
rs774213272 Chr.1: 158292393-158292393 on Build GRCh38
rs762306011 Chr.1: 158292286-158292286 on Build GRCh38
rs372819385 Chr.1: 158292383-158292383 on Build GRCh38
rs1027985102 Chr.1: 158292380-158292380 on Build GRCh38
rs754643486 Chr.1: 158292309-158292310 on Build GRCh38
rs746365992 Chr.1: 158292246-158292246 on Build GRCh38
rs777182596 Chr.1: 158292284-158292284 on Build GRCh38
rs759196612 Chr.1: 158292396-158292396 on Build GRCh38

View Full Product Details