Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97122796-97123065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD36
Entrez Gene ID
375248
Gene Name
ankyrin repeat domain 36
Gene Aliases
UNQ2430
UniGene
Hs.541894
Gene Chromosome Location
Chr.2: 97113186-97264521 on Build GRCh38
Transcripts
NM_001164315.1 XM_006712514.3 XM_011511130.1 XM_011511131.1 XM_017004009.1 XM_017004010.1 XM_017004011.1 XM_017004012.1 XM_017004013.1 XM_017004014.1 XM_017004015.1 XM_017004016.1 XM_017004017.1 XM_017004018.1 XM_017004019.1 XM_017004020.1 XM_017004021.1 XM_017004022.1 XM_017004023.1 XM_017004024.1 XM_017004025.1 XM_017004026.1 XM_017004027.1 XM_017004028.1 XM_017004029.1 XM_017004030.1 XM_017004031.1 XM_017004032.1 XM_017004033.1 XM_017004034.1 XM_017004035.1 XM_017004036.1 XM_017004037.1 XM_017004038.1 XM_017004039.1 XM_017004040.1 XM_017004041.1 XM_017004042.1 XM_017004043.1 XM_017004044.1 XM_017004045.1 XM_017004046.1 XM_017004047.1 XM_017004048.1 XM_017004049.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1024033 c.577A>G p.I193V Chr.2: 97122977-97122977 on Build GRCh38
3840314 c.565A>C p.N189H Chr.2: 97122965-97122965 on Build GRCh38
6261141 c.593G>T p.R198I Chr.2: 97122993-97122993 on Build GRCh38
6501780 c.559A>G p.K187E Chr.2: 97122959-97122959 on Build GRCh38
6261142 c.593G>T p.R198I Chr.2: 97122993-97122993 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544103818 Chr.2: 97122831-97122831 on Build GRCh38
rs756903791 Chr.2: 97122846-97122846 on Build GRCh38
rs771176336 Chr.2: 97123019-97123019 on Build GRCh38
rs559966737 Chr.2: 97122995-97122995 on Build GRCh38
rs574084137 Chr.2: 97122962-97122962 on Build GRCh38
rs779423591 Chr.2: 97122923-97122923 on Build GRCh38
rs368158747 Chr.2: 97123020-97123020 on Build GRCh38
rs917475314 Chr.2: 97123034-97123034 on Build GRCh38
rs781059768 Chr.2: 97122854-97122854 on Build GRCh38
rs138036717 Chr.2: 97122990-97122990 on Build GRCh38
rs1005229885 Chr.2: 97122921-97122921 on Build GRCh38
rs755337089 Chr.2: 97122916-97122916 on Build GRCh38
rs758239797 Chr.2: 97122868-97122868 on Build GRCh38
rs185041929 Chr.2: 97123037-97123037 on Build GRCh38
rs775287671 Chr.2: 97123038-97123038 on Build GRCh38
rs754240834 Chr.2: 97122908-97122908 on Build GRCh38
rs192445907 Chr.2: 97122920-97122920 on Build GRCh38

View Full Product Details