Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15258779-15259022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB9
Entrez Gene ID
140462
Gene Name
ankyrin repeat and SOCS box containing 9
Gene Aliases
-
UniGene
Hs.19404
Gene Chromosome Location
Chr.X: 15243987-15270476 on Build GRCh38
Transcripts
NM_001031739.2 NM_001168530.1 NM_001168531.1 NM_024087.2 XM_005274446.1 XM_011545458.1 XM_011545460.1 XM_017029283.1 XM_017029284.1 XM_017029285.1 XM_017029286.1

SNP Detail

SNP ID Chromosome Location
rs763038775 Chr.X: 15258891-15258891 on Build GRCh38
rs761949853 Chr.X: 15258845-15258845 on Build GRCh38
rs752213261 Chr.X: 15258951-15258951 on Build GRCh38
rs751887101 Chr.X: 15258917-15258917 on Build GRCh38
rs765167058 Chr.X: 15258850-15258850 on Build GRCh38
rs779869281 Chr.X: 15258980-15258980 on Build GRCh38
rs973098246 Chr.X: 15258972-15258972 on Build GRCh38
rs760083246 Chr.X: 15258939-15258939 on Build GRCh38
rs755653116 Chr.X: 15258952-15258952 on Build GRCh38
rs773501274 Chr.X: 15258870-15258870 on Build GRCh38
rs781111448 Chr.X: 15258948-15258948 on Build GRCh38
rs756450144 Chr.X: 15258817-15258817 on Build GRCh38
rs766764678 Chr.X: 15258902-15258902 on Build GRCh38
rs1031229149 Chr.X: 15258976-15258976 on Build GRCh38
rs1012655320 Chr.X: 15258815-15258815 on Build GRCh38
rs1001049363 Chr.X: 15258954-15258954 on Build GRCh38
rs757453227 Chr.X: 15258821-15258821 on Build GRCh38

View Full Product Details