Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47171317-47171580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFGAP2
Entrez Gene ID
84364
Gene Name
ADP ribosylation factor GTPase activating protein 2
Gene Aliases
IRZ, NBLA10535, ZFP289, ZNF289
UniGene
Hs.436204
Gene Chromosome Location
Chr.11: 47164298-47177503 on Build GRCh38
Transcripts
NM_001242832.1 NM_032389.4 XM_005253166.1 XM_005253167.1 XM_005253168.2 XM_006718346.1 XM_011520405.1 XM_017018413.1 XM_017018414.1 XM_017018415.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
466846 c.854G>T p.R285L Chr.11: 47171513-47171513 on Build GRCh38
5341533 c.810-5C>G p.? Chr.11: 47171562-47171562 on Build GRCh38
1725029 c.917_918insTT p.L306fs*22 Chr.11: 47171449-47171450 on Build GRCh38
1981239 c.938G>A p.R313Q Chr.11: 47171429-47171429 on Build GRCh38
141338 c.853C>A p.R285S Chr.11: 47171514-47171514 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766680279 Chr.11: 47171437-47171437 on Build GRCh38
rs770562056 Chr.11: 47171405-47171405 on Build GRCh38
rs376079930 Chr.11: 47171471-47171471 on Build GRCh38
rs1015570008 Chr.11: 47171346-47171346 on Build GRCh38
rs760467199 Chr.11: 47171418-47171418 on Build GRCh38
rs776670169 Chr.11: 47171554-47171554 on Build GRCh38
rs755280223 Chr.11: 47171460-47171460 on Build GRCh38
rs143629499 Chr.11: 47171543-47171543 on Build GRCh38
rs570987895 Chr.11: 47171359-47171359 on Build GRCh38
rs748561500 Chr.11: 47171390-47171390 on Build GRCh38
rs914650135 Chr.11: 47171468-47171468 on Build GRCh38
rs778906631 Chr.11: 47171463-47171468 on Build GRCh38
rs765077743 Chr.11: 47171557-47171557 on Build GRCh38
rs770336255 Chr.11: 47171391-47171391 on Build GRCh38
rs141420803 Chr.11: 47171510-47171510 on Build GRCh38
rs751880003 Chr.11: 47171447-47171447 on Build GRCh38
rs774030572 Chr.11: 47171406-47171406 on Build GRCh38
rs558754699 Chr.11: 47171370-47171370 on Build GRCh38
rs913530036 Chr.11: 47171484-47171484 on Build GRCh38
rs778408239 Chr.11: 47171509-47171509 on Build GRCh38
rs146780643 Chr.11: 47171546-47171546 on Build GRCh38
rs771332378 Chr.11: 47171514-47171514 on Build GRCh38
rs575420278 Chr.11: 47171410-47171410 on Build GRCh38
rs374182918 Chr.11: 47171403-47171403 on Build GRCh38
rs775099593 Chr.11: 47171416-47171416 on Build GRCh38
rs960033314 Chr.11: 47171347-47171347 on Build GRCh38
rs557674772 Chr.11: 47171548-47171548 on Build GRCh38
rs761681129 Chr.11: 47171556-47171556 on Build GRCh38
rs781064553 Chr.11: 47171470-47171470 on Build GRCh38
rs980229811 Chr.11: 47171494-47171494 on Build GRCh38
rs559792907 Chr.11: 47171553-47171553 on Build GRCh38
rs371380560 Chr.11: 47171429-47171429 on Build GRCh38
rs774717068 Chr.11: 47171515-47171515 on Build GRCh38
rs1052299204 Chr.11: 47171397-47171397 on Build GRCh38
rs932936190 Chr.11: 47171538-47171538 on Build GRCh38
rs752788617 Chr.11: 47171494-47171496 on Build GRCh38
rs374493764 Chr.11: 47171487-47171487 on Build GRCh38
rs748272296 Chr.11: 47171472-47171472 on Build GRCh38
rs538823239 Chr.11: 47171369-47171369 on Build GRCh38
rs995675661 Chr.11: 47171368-47171368 on Build GRCh38
rs554859128 Chr.11: 47171420-47171420 on Build GRCh38
rs145672265 Chr.11: 47171411-47171411 on Build GRCh38
rs745484725 Chr.11: 47171528-47171528 on Build GRCh38
rs992884194 Chr.11: 47171354-47171354 on Build GRCh38
rs757185986 Chr.11: 47171432-47171432 on Build GRCh38
rs755214880 Chr.11: 47171446-47171446 on Build GRCh38
rs756564398 Chr.11: 47171474-47171474 on Build GRCh38

View Full Product Details