Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44147824-44148083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf53
Entrez Gene ID
78995
Gene Name
chromosome 17 open reading frame 53
Gene Aliases
-
UniGene
Hs.437059
Gene Chromosome Location
Chr.17: 44141902-44162477 on Build GRCh38
Transcripts
NM_001171251.2 NM_001321310.1 NM_001321311.1 NM_024032.4 XM_011525187.1 XM_011525188.1 XM_011525189.1 XM_011525190.1 XM_011525191.2 XM_011525192.1 XM_011525193.1 XM_011525194.1 XM_011525195.1 XM_017025031.1 XM_017025032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3970091 c.257C>T p.T86M Chr.17: 44148060-44148060 on Build GRCh38
6081232 c.137C>G p.S46C Chr.17: 44147940-44147940 on Build GRCh38
6315915 c.168G>T p.Q56H Chr.17: 44147971-44147971 on Build GRCh38
6146977 c.143G>T p.R48M Chr.17: 44147946-44147946 on Build GRCh38
4753694 c.275G>A p.R92H Chr.17: 44148078-44148078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924806498 Chr.17: 44147851-44147851 on Build GRCh38
rs879143201 Chr.17: 44147970-44147970 on Build GRCh38
rs751626122 Chr.17: 44147996-44147996 on Build GRCh38
rs758027614 Chr.17: 44148044-44148044 on Build GRCh38
rs765782184 Chr.17: 44147857-44147857 on Build GRCh38
rs373094846 Chr.17: 44147925-44147925 on Build GRCh38
rs773745843 Chr.17: 44147956-44147956 on Build GRCh38
rs149678700 Chr.17: 44147924-44147924 on Build GRCh38
rs768145806 Chr.17: 44147950-44147950 on Build GRCh38
rs748801257 Chr.17: 44147934-44147934 on Build GRCh38
rs1013576538 Chr.17: 44147977-44147977 on Build GRCh38
rs200005578 Chr.17: 44147960-44147960 on Build GRCh38
rs761956200 Chr.17: 44147942-44147942 on Build GRCh38
rs759952461 Chr.17: 44147974-44147974 on Build GRCh38
rs139563255 Chr.17: 44147976-44147976 on Build GRCh38
rs745929297 Chr.17: 44147892-44147892 on Build GRCh38
rs965809569 Chr.17: 44147853-44147853 on Build GRCh38
rs376559222 Chr.17: 44147973-44147973 on Build GRCh38
rs191707252 Chr.17: 44147891-44147891 on Build GRCh38
rs757466740 Chr.17: 44147997-44147997 on Build GRCh38
rs139451289 Chr.17: 44147904-44147904 on Build GRCh38
rs147787171 Chr.17: 44147933-44147933 on Build GRCh38
rs749177606 Chr.17: 44148038-44148038 on Build GRCh38
rs771744510 Chr.17: 44148064-44148064 on Build GRCh38
rs201117972 Chr.17: 44147981-44147981 on Build GRCh38
rs771664188 Chr.17: 44147964-44147964 on Build GRCh38
rs755448659 Chr.17: 44147854-44147854 on Build GRCh38
rs35879996 Chr.17: 44147882-44147882 on Build GRCh38
rs779998519 Chr.17: 44148031-44148031 on Build GRCh38
rs563980398 Chr.17: 44148060-44148060 on Build GRCh38
rs750227681 Chr.17: 44148017-44148017 on Build GRCh38
rs781316792 Chr.17: 44148013-44148013 on Build GRCh38
rs779960035 Chr.17: 44147852-44147852 on Build GRCh38
rs760328625 Chr.17: 44148066-44148066 on Build GRCh38
rs764187345 Chr.17: 44147988-44147988 on Build GRCh38
rs375631727 Chr.17: 44147872-44147872 on Build GRCh38
rs757445731 Chr.17: 44147914-44147914 on Build GRCh38
rs373854314 Chr.17: 44148022-44148022 on Build GRCh38
rs747661873 Chr.17: 44148061-44148061 on Build GRCh38
rs768499928 Chr.17: 44148048-44148048 on Build GRCh38
rs763388362 Chr.17: 44147987-44147987 on Build GRCh38
rs772916845 Chr.17: 44148065-44148065 on Build GRCh38
rs977227418 Chr.17: 44148062-44148062 on Build GRCh38

View Full Product Details