Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50880391-50880652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD12B
Entrez Gene ID
145447
Gene Name
abhydrolase domain containing 12B
Gene Aliases
BEM46L3, C14orf29, c14_5314
UniGene
Hs.271896
Gene Chromosome Location
Chr.14: 50872090-50904970 on Build GRCh38
Transcripts
NM_001206673.1 NM_181533.3 NM_181814.1 XM_006720052.1 XM_011536472.1 XM_011536474.2 XM_017021031.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7203365 c.137C>A p.A46D Chr.14: 50880484-50880484 on Build GRCh38
4658496 c.176G>A p.R59H Chr.14: 50880523-50880523 on Build GRCh38
6590822 c.408T>C p.R136R Chr.14: 50880524-50880524 on Build GRCh38
4658498 c.183G>A p.G61G Chr.14: 50880530-50880530 on Build GRCh38
4658495 c.407G>A p.R136H Chr.14: 50880523-50880523 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774341892 Chr.14: 50880563-50880568 on Build GRCh38
rs770378738 Chr.14: 50880436-50880436 on Build GRCh38
rs764564347 Chr.14: 50880550-50880550 on Build GRCh38
rs777420122 Chr.14: 50880417-50880417 on Build GRCh38
rs762019272 Chr.14: 50880457-50880457 on Build GRCh38
rs756654748 Chr.14: 50880548-50880548 on Build GRCh38
rs779667300 Chr.14: 50880602-50880602 on Build GRCh38
rs747981965 Chr.14: 50880614-50880614 on Build GRCh38
rs368116773 Chr.14: 50880574-50880574 on Build GRCh38
rs758948210 Chr.14: 50880446-50880446 on Build GRCh38
rs571246952 Chr.14: 50880632-50880632 on Build GRCh38
rs749622217 Chr.14: 50880486-50880486 on Build GRCh38
rs1025891747 Chr.14: 50880482-50880482 on Build GRCh38
rs371975948 Chr.14: 50880506-50880506 on Build GRCh38
rs750691867 Chr.14: 50880469-50880469 on Build GRCh38
rs760550383 Chr.14: 50880462-50880462 on Build GRCh38
rs752492456 Chr.14: 50880608-50880609 on Build GRCh38
rs531534584 Chr.14: 50880473-50880473 on Build GRCh38
rs775581616 Chr.14: 50880505-50880505 on Build GRCh38
rs774373488 Chr.14: 50880534-50880534 on Build GRCh38
rs769805122 Chr.14: 50880544-50880544 on Build GRCh38
rs770081676 Chr.14: 50880416-50880416 on Build GRCh38
rs753148607 Chr.14: 50880475-50880475 on Build GRCh38
rs894264737 Chr.14: 50880626-50880626 on Build GRCh38
rs768760134 Chr.14: 50880611-50880611 on Build GRCh38
rs770669064 Chr.14: 50880491-50880491 on Build GRCh38
rs377112021 Chr.14: 50880444-50880444 on Build GRCh38
rs781725974 Chr.14: 50880474-50880474 on Build GRCh38
rs560987049 Chr.14: 50880586-50880586 on Build GRCh38
rs746483075 Chr.14: 50880610-50880610 on Build GRCh38
rs759679516 Chr.14: 50880538-50880538 on Build GRCh38
rs143764492 Chr.14: 50880523-50880523 on Build GRCh38
rs772103938 Chr.14: 50880450-50880450 on Build GRCh38
rs778415828 Chr.14: 50880495-50880495 on Build GRCh38
rs186226043 Chr.14: 50880623-50880623 on Build GRCh38
rs767240814 Chr.14: 50880593-50880593 on Build GRCh38
rs139997584 Chr.14: 50880514-50880514 on Build GRCh38
rs368157046 Chr.14: 50880477-50880477 on Build GRCh38
rs891205724 Chr.14: 50880428-50880428 on Build GRCh38
rs538304820 Chr.14: 50880592-50880593 on Build GRCh38
rs745449820 Chr.14: 50880497-50880497 on Build GRCh38
rs778072071 Chr.14: 50880479-50880479 on Build GRCh38
rs768600574 Chr.14: 50880509-50880509 on Build GRCh38
rs924962222 Chr.14: 50880624-50880624 on Build GRCh38
rs748891865 Chr.14: 50880420-50880420 on Build GRCh38
rs150681086 Chr.14: 50880498-50880498 on Build GRCh38
rs776694397 Chr.14: 50880613-50880613 on Build GRCh38
rs757947582 Chr.14: 50880593-50880593 on Build GRCh38
rs745452955 Chr.14: 50880592-50880592 on Build GRCh38
rs776470941 Chr.14: 50880453-50880453 on Build GRCh38
rs977338550 Chr.14: 50880537-50880537 on Build GRCh38
rs201529226 Chr.14: 50880443-50880443 on Build GRCh38
rs774659987 Chr.14: 50880616-50880616 on Build GRCh38
rs200952659 Chr.14: 50880472-50880472 on Build GRCh38
rs374247021 Chr.14: 50880522-50880522 on Build GRCh38
rs947805856 Chr.14: 50880607-50880607 on Build GRCh38
rs1029739303 Chr.14: 50880421-50880421 on Build GRCh38
rs769420133 Chr.14: 50880615-50880615 on Build GRCh38
rs141865189 Chr.14: 50880471-50880471 on Build GRCh38
rs753234503 Chr.14: 50880544-50880544 on Build GRCh38
rs747543592 Chr.14: 50880590-50880590 on Build GRCh38

View Full Product Details