Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136993204-136993451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf142
Entrez Gene ID
286257
Gene Name
chromosome 9 open reading frame 142
Gene Aliases
PAXX, XLS
UniGene
Hs.409582
Gene Chromosome Location
Chr.9: 136992418-136993976 on Build GRCh38
Transcripts
NM_183241.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6292841 c.427G>T p.E143* Chr.9: 136993348-136993348 on Build GRCh38
3703521 c.471G>T p.G157G Chr.9: 136993392-136993392 on Build GRCh38
7213547 c.421+9G>T p.? Chr.9: 136993252-136993252 on Build GRCh38
5467647 c.490+9T>C p.? Chr.9: 136993420-136993420 on Build GRCh38
6616837 c.421+9delG p.? Chr.9: 136993252-136993252 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752173010 Chr.9: 136993428-136993428 on Build GRCh38
rs533507781 Chr.9: 136993304-136993304 on Build GRCh38
rs774501223 Chr.9: 136993368-136993368 on Build GRCh38
rs778710020 Chr.9: 136993232-136993232 on Build GRCh38
rs953705577 Chr.9: 136993285-136993285 on Build GRCh38
rs939610584 Chr.9: 136993257-136993257 on Build GRCh38
rs745789397 Chr.9: 136993431-136993431 on Build GRCh38
rs202216228 Chr.9: 136993239-136993239 on Build GRCh38
rs758047696 Chr.9: 136993418-136993418 on Build GRCh38
rs778477084 Chr.9: 136993314-136993314 on Build GRCh38
rs762140528 Chr.9: 136993276-136993276 on Build GRCh38
rs775866385 Chr.9: 136993267-136993267 on Build GRCh38
rs145852011 Chr.9: 136993352-136993352 on Build GRCh38
rs146967007 Chr.9: 136993424-136993424 on Build GRCh38
rs371701132 Chr.9: 136993279-136993279 on Build GRCh38
rs934386051 Chr.9: 136993415-136993415 on Build GRCh38
rs753304135 Chr.9: 136993306-136993306 on Build GRCh38
rs373617347 Chr.9: 136993341-136993341 on Build GRCh38
rs201336592 Chr.9: 136993270-136993270 on Build GRCh38
rs764801089 Chr.9: 136993405-136993405 on Build GRCh38
rs754538246 Chr.9: 136993404-136993404 on Build GRCh38
rs766803814 Chr.9: 136993387-136993387 on Build GRCh38
rs377073523 Chr.9: 136993394-136993394 on Build GRCh38
rs937304445 Chr.9: 136993426-136993426 on Build GRCh38
rs141325653 Chr.9: 136993351-136993351 on Build GRCh38
rs775689767 Chr.9: 136993245-136993245 on Build GRCh38
rs749316845 Chr.9: 136993367-136993367 on Build GRCh38
rs199908002 Chr.9: 136993224-136993224 on Build GRCh38
rs566004185 Chr.9: 136993381-136993381 on Build GRCh38
rs528735705 Chr.9: 136993419-136993419 on Build GRCh38
rs746969084 Chr.9: 136993348-136993348 on Build GRCh38
rs570101362 Chr.9: 136993425-136993425 on Build GRCh38
rs747829305 Chr.9: 136993235-136993235 on Build GRCh38
rs572498199 Chr.9: 136993248-136993248 on Build GRCh38
rs765916751 Chr.9: 136993267-136993267 on Build GRCh38
rs752334084 Chr.9: 136993416-136993416 on Build GRCh38
rs777594027 Chr.9: 136993338-136993338 on Build GRCh38
rs369629141 Chr.9: 136993307-136993307 on Build GRCh38
rs542127750 Chr.9: 136993393-136993393 on Build GRCh38
rs750930412 Chr.9: 136993297-136993297 on Build GRCh38
rs761201559 Chr.9: 136993382-136993382 on Build GRCh38
rs184303541 Chr.9: 136993317-136993317 on Build GRCh38
rs773083243 Chr.9: 136993237-136993237 on Build GRCh38
rs978669923 Chr.9: 136993353-136993353 on Build GRCh38
rs549427066 Chr.9: 136993380-136993380 on Build GRCh38
rs370028945 Chr.9: 136993423-136993423 on Build GRCh38
rs753247052 Chr.9: 136993345-136993347 on Build GRCh38
rs762060610 Chr.9: 136993371-136993371 on Build GRCh38
rs770035022 Chr.9: 136993241-136993241 on Build GRCh38
rs764374885 Chr.9: 136993251-136993251 on Build GRCh38
rs200249254 Chr.9: 136993299-136993299 on Build GRCh38
rs758196163 Chr.9: 136993319-136993319 on Build GRCh38

View Full Product Details