Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26372864-26373129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN3A2
Entrez Gene ID
11118
Gene Name
butyrophilin subfamily 3 member A2
Gene Aliases
BT3.2, BTF4, BTN3.2, CD277
UniGene
Hs.376046
Gene Chromosome Location
Chr.6: 26365159-26451697 on Build GRCh38
Transcripts
NM_001197246.2 NM_001197247.2 NM_001197248.2 NM_001197249.2 NM_007047.4 XM_005248827.3 XM_005248831.3 XM_005248832.3 XM_006714979.3 XM_006714980.3 XM_006714981.3 XM_006714982.3 XM_011514267.2 XM_011514268.2 XM_011514269.2 XM_011514270.2 XM_011514271.2 XM_017010211.1 XM_017010212.1 XM_017010213.1 XM_017010214.1 XM_017010215.1 XM_017010216.1 XM_017010217.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5577289 c.761G>A p.G254E Chr.6: 26372942-26372942 on Build GRCh38
5577288 c.761G>A p.G254E Chr.6: 26372942-26372942 on Build GRCh38
3410851 c.793G>A p.G265R Chr.6: 26372974-26372974 on Build GRCh38
1696994 c.896A>T p.E299V Chr.6: 26373077-26373077 on Build GRCh38
301780 c.788T>C p.L263P Chr.6: 26372969-26372969 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568843545 Chr.6: 26372929-26372929 on Build GRCh38
rs772400538 Chr.6: 26373016-26373016 on Build GRCh38
rs780468088 Chr.6: 26372914-26372914 on Build GRCh38
rs774078806 Chr.6: 26373089-26373090 on Build GRCh38
rs754314730 Chr.6: 26372940-26372940 on Build GRCh38
rs568607568 Chr.6: 26372987-26372987 on Build GRCh38
rs554201845 Chr.6: 26373082-26373082 on Build GRCh38
rs927970646 Chr.6: 26373004-26373004 on Build GRCh38
rs760733555 Chr.6: 26372931-26372931 on Build GRCh38
rs762532058 Chr.6: 26372956-26372957 on Build GRCh38
rs751198622 Chr.6: 26372956-26372956 on Build GRCh38
rs371708538 Chr.6: 26372888-26372888 on Build GRCh38
rs780580226 Chr.6: 26372970-26372970 on Build GRCh38
rs144393024 Chr.6: 26372973-26372973 on Build GRCh38
rs982086366 Chr.6: 26372980-26372980 on Build GRCh38
rs959875487 Chr.6: 26372960-26372960 on Build GRCh38
rs767585614 Chr.6: 26372882-26372882 on Build GRCh38
rs767997124 Chr.6: 26373014-26373018 on Build GRCh38
rs759993496 Chr.6: 26373077-26373077 on Build GRCh38
rs529887353 Chr.6: 26372892-26372892 on Build GRCh38
rs111392726 Chr.6: 26373080-26373080 on Build GRCh38
rs750068318 Chr.6: 26373101-26373101 on Build GRCh38
rs779284406 Chr.6: 26372950-26372950 on Build GRCh38
rs372417646 Chr.6: 26372915-26372915 on Build GRCh38
rs749190080 Chr.6: 26372978-26372978 on Build GRCh38
rs774129855 Chr.6: 26372991-26372991 on Build GRCh38
rs146618645 Chr.6: 26373079-26373079 on Build GRCh38
rs368782404 Chr.6: 26372913-26372913 on Build GRCh38
rs142279595 Chr.6: 26372900-26372900 on Build GRCh38
rs903114249 Chr.6: 26372887-26372887 on Build GRCh38
rs531941877 Chr.6: 26373074-26373074 on Build GRCh38
rs755382991 Chr.6: 26372942-26372942 on Build GRCh38
rs560756333 Chr.6: 26372881-26372881 on Build GRCh38
rs12174631 Chr.6: 26372922-26372922 on Build GRCh38
rs532046699 Chr.6: 26372905-26372905 on Build GRCh38
rs745794122 Chr.6: 26372906-26372906 on Build GRCh38
rs12174639 Chr.6: 26372893-26372893 on Build GRCh38
rs766426965 Chr.6: 26373032-26373032 on Build GRCh38
rs767499921 Chr.6: 26372923-26372923 on Build GRCh38
rs566739896 Chr.6: 26372899-26372899 on Build GRCh38
rs777332474 Chr.6: 26372898-26372898 on Build GRCh38
rs766940767 Chr.6: 26372936-26372936 on Build GRCh38
rs745427991 Chr.6: 26372971-26372971 on Build GRCh38
rs369533018 Chr.6: 26373106-26373106 on Build GRCh38
rs534380775 Chr.6: 26373020-26373020 on Build GRCh38
rs200135502 Chr.6: 26373098-26373098 on Build GRCh38
rs780178954 Chr.6: 26372974-26372974 on Build GRCh38
rs947204049 Chr.6: 26373083-26373083 on Build GRCh38
rs750926111 Chr.6: 26372884-26372884 on Build GRCh38
rs760786726 Chr.6: 26373027-26373027 on Build GRCh38
rs139423944 Chr.6: 26372962-26372962 on Build GRCh38
rs767266552 Chr.6: 26373090-26373090 on Build GRCh38
rs754362000 Chr.6: 26373060-26373060 on Build GRCh38
rs761954790 Chr.6: 26372993-26372993 on Build GRCh38

View Full Product Details