Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 48233965-48234210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA13
Entrez Gene ID
154664
Gene Name
ATP binding cassette subfamily A member 13
Gene Aliases
-
UniGene
Hs.226568
Gene Chromosome Location
Chr.7: 48171424-48647497 on Build GRCh38
Transcripts
NM_152701.4 XM_011515130.2 XM_011515131.2 XM_011515132.2 XM_011515133.2 XM_011515134.2 XM_011515136.2 XM_011515137.2 XM_011515138.2 XM_011515139.2 XM_011515141.2 XM_011515142.2 XM_011515143.2 XM_011515144.2 XM_011515145.2 XM_011515146.2 XM_011515147.2 XM_011515148.2 XM_011515149.2 XM_017011767.1 XM_017011768.1 XM_017011769.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5345371 c.726G>T p.L242L Chr.7: 48234145-48234145 on Build GRCh38
3715748 c.856C>T p.H286Y Chr.7: 48234110-48234110 on Build GRCh38
3639267 c.669C>T p.S223S Chr.7: 48234088-48234088 on Build GRCh38
3639269 c.729G>A p.K243K Chr.7: 48234148-48234148 on Build GRCh38
6347988 c.658G>T p.D220Y Chr.7: 48234077-48234077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771747708 Chr.7: 48234174-48234174 on Build GRCh38
rs376125013 Chr.7: 48234026-48234026 on Build GRCh38
rs746090084 Chr.7: 48233999-48233999 on Build GRCh38
rs114914138 Chr.7: 48234175-48234175 on Build GRCh38
rs757158140 Chr.7: 48234084-48234084 on Build GRCh38
rs762336102 Chr.7: 48234041-48234041 on Build GRCh38
rs367866741 Chr.7: 48234088-48234088 on Build GRCh38
rs1008876353 Chr.7: 48234186-48234186 on Build GRCh38
rs372241108 Chr.7: 48234179-48234179 on Build GRCh38
rs780391805 Chr.7: 48234109-48234109 on Build GRCh38
rs367697881 Chr.7: 48234184-48234184 on Build GRCh38
rs780624607 Chr.7: 48234020-48234020 on Build GRCh38
rs370659927 Chr.7: 48234151-48234151 on Build GRCh38
rs370501374 Chr.7: 48234052-48234052 on Build GRCh38
rs770362374 Chr.7: 48234043-48234043 on Build GRCh38
rs760427001 Chr.7: 48234066-48234066 on Build GRCh38
rs566275204 Chr.7: 48234057-48234057 on Build GRCh38
rs202012795 Chr.7: 48234180-48234180 on Build GRCh38
rs938327760 Chr.7: 48234016-48234016 on Build GRCh38
rs745357121 Chr.7: 48234164-48234164 on Build GRCh38
rs375547406 Chr.7: 48234159-48234159 on Build GRCh38
rs779001315 Chr.7: 48233998-48233998 on Build GRCh38
rs1043197281 Chr.7: 48234093-48234093 on Build GRCh38
rs777039751 Chr.7: 48234039-48234039 on Build GRCh38
rs983540467 Chr.7: 48234172-48234172 on Build GRCh38
rs750166574 Chr.7: 48234055-48234057 on Build GRCh38
rs755813741 Chr.7: 48234070-48234087 on Build GRCh38
rs772352143 Chr.7: 48234004-48234004 on Build GRCh38
rs373967596 Chr.7: 48234025-48234025 on Build GRCh38
rs755419260 Chr.7: 48234124-48234124 on Build GRCh38
rs758745044 Chr.7: 48234097-48234097 on Build GRCh38
rs759248586 Chr.7: 48234053-48234053 on Build GRCh38
rs770332276 Chr.7: 48234158-48234158 on Build GRCh38
rs369906254 Chr.7: 48234067-48234067 on Build GRCh38
rs996749414 Chr.7: 48234160-48234160 on Build GRCh38
rs372215692 Chr.7: 48234115-48234115 on Build GRCh38
rs753741777 Chr.7: 48234075-48234075 on Build GRCh38
rs781567190 Chr.7: 48234150-48234150 on Build GRCh38
rs552464039 Chr.7: 48234056-48234056 on Build GRCh38

View Full Product Details