Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7089356-7089629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1R
Entrez Gene ID
715
Gene Name
complement C1r subcomponent
Gene Aliases
-
UniGene
Hs.524224
Gene Chromosome Location
Chr.12: 7080209-7092447 on Build GRCh38
Transcripts
NM_001733.4 XM_017019915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7358355 c.595G>A p.D199N Chr.12: 7089605-7089605 on Build GRCh38
3872461 c.664C>T p.L222L Chr.12: 7089439-7089439 on Build GRCh38
294194 c.689delC p.P230fs*100 Chr.12: 7089414-7089414 on Build GRCh38
3872462 c.577G>A p.G193S Chr.12: 7089623-7089623 on Build GRCh38
6880407 c.585G>A p.E195E Chr.12: 7089615-7089615 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111937645 Chr.12: 7089514-7089514 on Build GRCh38
rs367700816 Chr.12: 7089415-7089415 on Build GRCh38
rs766984810 Chr.12: 7089573-7089574 on Build GRCh38
rs756745476 Chr.12: 7089545-7089545 on Build GRCh38
rs769139608 Chr.12: 7089549-7089549 on Build GRCh38
rs747484081 Chr.12: 7089548-7089548 on Build GRCh38
rs762993517 Chr.12: 7089513-7089513 on Build GRCh38
rs369628991 Chr.12: 7089464-7089464 on Build GRCh38
rs746129350 Chr.12: 7089428-7089428 on Build GRCh38
rs747824483 Chr.12: 7089434-7089434 on Build GRCh38
rs751494513 Chr.12: 7089419-7089419 on Build GRCh38
rs370601700 Chr.12: 7089406-7089406 on Build GRCh38
rs772664085 Chr.12: 7089395-7089395 on Build GRCh38
rs748580260 Chr.12: 7089571-7089571 on Build GRCh38
rs199899342 Chr.12: 7089405-7089405 on Build GRCh38
rs752287162 Chr.12: 7089533-7089533 on Build GRCh38
rs959113863 Chr.12: 7089431-7089431 on Build GRCh38
rs770450354 Chr.12: 7089470-7089470 on Build GRCh38
rs777447841 Chr.12: 7089535-7089535 on Build GRCh38
rs112616697 Chr.12: 7089491-7089491 on Build GRCh38
rs1009481107 Chr.12: 7089410-7089410 on Build GRCh38
rs189155429 Chr.12: 7089414-7089414 on Build GRCh38
rs777271496 Chr.12: 7089562-7089562 on Build GRCh38
rs760052651 Chr.12: 7089397-7089397 on Build GRCh38
rs765619313 Chr.12: 7089504-7089504 on Build GRCh38
rs200735593 Chr.12: 7089450-7089450 on Build GRCh38
rs772395563 Chr.12: 7089401-7089401 on Build GRCh38
rs926334802 Chr.12: 7089429-7089429 on Build GRCh38
rs773610671 Chr.12: 7089578-7089578 on Build GRCh38
rs1126605 Chr.12: 7089608-7089608 on Build GRCh38
rs761199969 Chr.12: 7089471-7089471 on Build GRCh38
rs768487195 Chr.12: 7089378-7089378 on Build GRCh38
rs267603660 Chr.12: 7089605-7089605 on Build GRCh38
rs762077783 Chr.12: 7089407-7089407 on Build GRCh38
rs763194636 Chr.12: 7089584-7089584 on Build GRCh38
rs768921131 Chr.12: 7089475-7089475 on Build GRCh38
rs763446444 Chr.12: 7089554-7089554 on Build GRCh38
rs998548080 Chr.12: 7089570-7089570 on Build GRCh38
rs747767420 Chr.12: 7089383-7089383 on Build GRCh38
rs375378142 Chr.12: 7089517-7089517 on Build GRCh38
rs371821985 Chr.12: 7089416-7089416 on Build GRCh38
rs368895945 Chr.12: 7089546-7089546 on Build GRCh38
rs767238087 Chr.12: 7089575-7089575 on Build GRCh38
rs751484583 Chr.12: 7089516-7089516 on Build GRCh38
rs901283712 Chr.12: 7089496-7089496 on Build GRCh38
rs752425153 Chr.12: 7089522-7089522 on Build GRCh38
rs1000247449 Chr.12: 7089582-7089582 on Build GRCh38
rs4519167 Chr.12: 7089601-7089601 on Build GRCh38
rs376068614 Chr.12: 7089426-7089426 on Build GRCh38
rs750747014 Chr.12: 7089510-7089510 on Build GRCh38
rs535296088 Chr.12: 7089379-7089379 on Build GRCh38
rs768951004 Chr.12: 7089536-7089536 on Build GRCh38
rs773714106 Chr.12: 7089413-7089413 on Build GRCh38
rs572440545 Chr.12: 7089508-7089508 on Build GRCh38
rs368607035 Chr.12: 7089427-7089427 on Build GRCh38
rs374193306 Chr.12: 7089374-7089374 on Build GRCh38
rs769328977 Chr.12: 7089387-7089387 on Build GRCh38
rs777077198 Chr.12: 7089476-7089476 on Build GRCh38
rs73046140 Chr.12: 7089518-7089518 on Build GRCh38
rs113182443 Chr.12: 7089443-7089443 on Build GRCh38
rs762130885 Chr.12: 7089478-7089478 on Build GRCh38

View Full Product Details