Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81192181-81192426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP131
Entrez Gene ID
22994
Gene Name
centrosomal protein 131
Gene Aliases
AZ1, AZI1, ZA1
UniGene
Hs.514578
Gene Chromosome Location
Chr.17: 81189593-81222989 on Build GRCh38
Transcripts
NM_001009811.3 NM_001319228.1 NM_001319229.1 NM_014984.3 XM_011524547.2 XM_011524548.2 XM_011524550.2 XM_011524551.2 XM_011524552.2 XM_011524553.2 XM_011524555.2 XM_017024369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3523519 c.2470G>A p.E824K Chr.17: 81192362-81192362 on Build GRCh38
3756007 c.2496C>T p.A832A Chr.17: 81192336-81192336 on Build GRCh38
3523518 c.2578G>A p.E860K Chr.17: 81192362-81192362 on Build GRCh38
1387226 c.2609G>A p.R870H Chr.17: 81192331-81192331 on Build GRCh38
1387227 c.2501G>A p.R834H Chr.17: 81192331-81192331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778589604 Chr.17: 81192280-81192280 on Build GRCh38
rs780691586 Chr.17: 81192392-81192392 on Build GRCh38
rs764333781 Chr.17: 81192350-81192350 on Build GRCh38
rs987521682 Chr.17: 81192334-81192334 on Build GRCh38
rs774958669 Chr.17: 81192397-81192397 on Build GRCh38
rs111662238 Chr.17: 81192394-81192394 on Build GRCh38
rs750298089 Chr.17: 81192365-81192365 on Build GRCh38
rs758337262 Chr.17: 81192394-81192395 on Build GRCh38
rs775990178 Chr.17: 81192404-81192404 on Build GRCh38
rs867691963 Chr.17: 81192373-81192373 on Build GRCh38
rs746731644 Chr.17: 81192389-81192389 on Build GRCh38
rs533349678 Chr.17: 81192207-81192207 on Build GRCh38
rs113590232 Chr.17: 81192336-81192336 on Build GRCh38
rs549532630 Chr.17: 81192272-81192272 on Build GRCh38
rs1003169954 Chr.17: 81192246-81192246 on Build GRCh38
rs2279916 Chr.17: 81192401-81192401 on Build GRCh38
rs576170952 Chr.17: 81192363-81192363 on Build GRCh38
rs1052956655 Chr.17: 81192212-81192212 on Build GRCh38
rs142714736 Chr.17: 81192379-81192379 on Build GRCh38
rs776115714 Chr.17: 81192304-81192304 on Build GRCh38
rs568037087 Chr.17: 81192287-81192287 on Build GRCh38
rs891591849 Chr.17: 81192213-81192213 on Build GRCh38
rs870187 Chr.17: 81192303-81192303 on Build GRCh38
rs894929412 Chr.17: 81192257-81192257 on Build GRCh38
rs201824599 Chr.17: 81192366-81192366 on Build GRCh38
rs751329243 Chr.17: 81192402-81192402 on Build GRCh38
rs754556585 Chr.17: 81192391-81192391 on Build GRCh38
rs779796959 Chr.17: 81192295-81192295 on Build GRCh38
rs1042299404 Chr.17: 81192208-81192208 on Build GRCh38
rs370319596 Chr.17: 81192399-81192399 on Build GRCh38
rs2279917 Chr.17: 81192357-81192357 on Build GRCh38
rs757154064 Chr.17: 81192271-81192271 on Build GRCh38
rs750520168 Chr.17: 81192353-81192355 on Build GRCh38
rs1041619184 Chr.17: 81192247-81192273 on Build GRCh38
rs754758468 Chr.17: 81192406-81192407 on Build GRCh38
rs779978850 Chr.17: 81192398-81192401 on Build GRCh38
rs116806556 Chr.17: 81192324-81192324 on Build GRCh38
rs906208751 Chr.17: 81192232-81192232 on Build GRCh38
rs771409290 Chr.17: 81192331-81192331 on Build GRCh38
rs570087010 Chr.17: 81192222-81192222 on Build GRCh38
rs373546590 Chr.17: 81192220-81192220 on Build GRCh38
rs771371939 Chr.17: 81192396-81192396 on Build GRCh38
rs759904172 Chr.17: 81192335-81192335 on Build GRCh38
rs368110005 Chr.17: 81192402-81192402 on Build GRCh38
rs760963477 Chr.17: 81192346-81192346 on Build GRCh38
rs565585486 Chr.17: 81192332-81192332 on Build GRCh38
rs865776730 Chr.17: 81192344-81192344 on Build GRCh38
rs531414588 Chr.17: 81192224-81192224 on Build GRCh38
rs745631405 Chr.17: 81192286-81192286 on Build GRCh38
rs753881831 Chr.17: 81192270-81192270 on Build GRCh38
rs747644761 Chr.17: 81192395-81192395 on Build GRCh38

View Full Product Details