Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33256722-33256983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG1
Entrez Gene ID
573
Gene Name
BCL2 associated athanogene 1
Gene Aliases
BAG-1, HAP, RAP46
UniGene
Hs.377484
Gene Chromosome Location
Chr.9: 33252471-33264761 on Build GRCh38
Transcripts
NM_001172415.1 NM_004323.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
753616 c.868G>A p.E290K Chr.9: 33256818-33256818 on Build GRCh38
2772477 c.789C>T p.P263P Chr.9: 33256897-33256897 on Build GRCh38
3395791 c.523G>A p.E175K Chr.9: 33256818-33256818 on Build GRCh38
5932363 c.778-5C>T p.? Chr.9: 33256913-33256913 on Build GRCh38
5932364 c.433-5C>T p.? Chr.9: 33256913-33256913 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747570862 Chr.9: 33256800-33256800 on Build GRCh38
rs750072265 Chr.9: 33256757-33256757 on Build GRCh38
rs768587971 Chr.9: 33256952-33256952 on Build GRCh38
rs746206567 Chr.9: 33256926-33256926 on Build GRCh38
rs896820477 Chr.9: 33256940-33256941 on Build GRCh38
rs899933890 Chr.9: 33256822-33256822 on Build GRCh38
rs770085672 Chr.9: 33256953-33256953 on Build GRCh38
rs751413256 Chr.9: 33256868-33256868 on Build GRCh38
rs941271347 Chr.9: 33256852-33256852 on Build GRCh38
rs755881556 Chr.9: 33256771-33256771 on Build GRCh38
rs867402358 Chr.9: 33256923-33256923 on Build GRCh38
rs148464069 Chr.9: 33256827-33256827 on Build GRCh38
rs778082997 Chr.9: 33256789-33256789 on Build GRCh38
rs769913302 Chr.9: 33256803-33256806 on Build GRCh38
rs985326716 Chr.9: 33256792-33256792 on Build GRCh38
rs778512738 Chr.9: 33256911-33256911 on Build GRCh38
rs753542393 Chr.9: 33256901-33256901 on Build GRCh38
rs377359750 Chr.9: 33256841-33256841 on Build GRCh38
rs143706180 Chr.9: 33256856-33256856 on Build GRCh38
rs770211672 Chr.9: 33256930-33256930 on Build GRCh38
rs750303225 Chr.9: 33256874-33256874 on Build GRCh38
rs192414877 Chr.9: 33256893-33256893 on Build GRCh38
rs574806930 Chr.9: 33256830-33256830 on Build GRCh38
rs775787205 Chr.9: 33256842-33256842 on Build GRCh38
rs1009569924 Chr.9: 33256883-33256883 on Build GRCh38
rs775529887 Chr.9: 33256939-33256939 on Build GRCh38
rs374125409 Chr.9: 33256913-33256913 on Build GRCh38
rs370851662 Chr.9: 33256782-33256782 on Build GRCh38
rs781520774 Chr.9: 33256918-33256918 on Build GRCh38
rs777018751 Chr.9: 33256811-33256811 on Build GRCh38
rs767399114 Chr.9: 33256873-33256873 on Build GRCh38
rs764260677 Chr.9: 33256862-33256862 on Build GRCh38
rs761881834 Chr.9: 33256870-33256870 on Build GRCh38
rs754395663 Chr.9: 33256905-33256905 on Build GRCh38
rs765879496 Chr.9: 33256896-33256896 on Build GRCh38
rs748814717 Chr.9: 33256781-33256781 on Build GRCh38
rs933787860 Chr.9: 33256786-33256786 on Build GRCh38
rs749557183 Chr.9: 33256942-33256942 on Build GRCh38
rs771240750 Chr.9: 33256804-33256804 on Build GRCh38
rs747479504 Chr.9: 33256912-33256912 on Build GRCh38

View Full Product Details