Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 13001372-13001645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP192
Entrez Gene ID
55125
Gene Name
centrosomal protein 192
Gene Aliases
PPP1R62
UniGene
Hs.100914
Gene Chromosome Location
Chr.18: 12991307-13125053 on Build GRCh38
Transcripts
NM_032142.3 XM_005258107.3 XM_005258109.3 XM_006722326.3 XM_006722327.3 XM_006722330.3 XM_011525673.2 XM_017025803.1 XM_017025804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
148402 c.221G>C p.G74A Chr.18: 13001513-13001513 on Build GRCh38
7360877 c.199G>T p.G67W Chr.18: 13001491-13001491 on Build GRCh38
3756072 c.232G>A p.G78R Chr.18: 13001524-13001524 on Build GRCh38
7360876 c.199G>A p.G67R Chr.18: 13001491-13001491 on Build GRCh38
3422057 c.181G>T p.A61S Chr.18: 13001473-13001473 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs569650034 Chr.18: 13001425-13001425 on Build GRCh38
rs1032424546 Chr.18: 13001603-13001603 on Build GRCh38
rs142767003 Chr.18: 13001411-13001411 on Build GRCh38
rs756777298 Chr.18: 13001397-13001397 on Build GRCh38
rs1017519939 Chr.18: 13001533-13001533 on Build GRCh38
rs565948559 Chr.18: 13001518-13001518 on Build GRCh38
rs771920309 Chr.18: 13001451-13001451 on Build GRCh38
rs993196635 Chr.18: 13001495-13001495 on Build GRCh38
rs114856311 Chr.18: 13001491-13001491 on Build GRCh38
rs948405633 Chr.18: 13001443-13001443 on Build GRCh38
rs966546259 Chr.18: 13001556-13001556 on Build GRCh38
rs904105744 Chr.18: 13001514-13001514 on Build GRCh38
rs561852357 Chr.18: 13001440-13001440 on Build GRCh38
rs1047758815 Chr.18: 13001506-13001506 on Build GRCh38
rs11080618 Chr.18: 13001524-13001524 on Build GRCh38
rs984734003 Chr.18: 13001440-13001441 on Build GRCh38
rs116318687 Chr.18: 13001593-13001593 on Build GRCh38
rs374489828 Chr.18: 13001578-13001578 on Build GRCh38
rs759262244 Chr.18: 13001512-13001512 on Build GRCh38
rs201553106 Chr.18: 13001489-13001489 on Build GRCh38
rs866763825 Chr.18: 13001523-13001523 on Build GRCh38
rs1044075599 Chr.18: 13001445-13001445 on Build GRCh38
rs966122531 Chr.18: 13001393-13001393 on Build GRCh38
rs191073576 Chr.18: 13001463-13001463 on Build GRCh38
rs764553056 Chr.18: 13001534-13001534 on Build GRCh38
rs530564973 Chr.18: 13001459-13001459 on Build GRCh38
rs1051633470 Chr.18: 13001405-13001405 on Build GRCh38
rs4499304 Chr.18: 13001513-13001513 on Build GRCh38
rs200338892 Chr.18: 13001454-13001454 on Build GRCh38
rs759388099 Chr.18: 13001511-13001511 on Build GRCh38

View Full Product Details