Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109488938-109489195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7L2
Entrez Gene ID
127002
Gene Name
ataxin 7 like 2
Gene Aliases
-
UniGene
Hs.118248
Gene Chromosome Location
Chr.1: 109483904-109492804 on Build GRCh38
Transcripts
NM_153340.4 XM_005270445.3 XM_005270446.2 XM_011540638.2 XM_011540639.2 XM_011540641.2 XM_011540644.2 XM_011540645.1 XM_011540646.2 XM_017000267.1 XM_017000268.1 XM_017000269.1 XM_017000270.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5498737 c.1030C>T p.L344L Chr.1: 109489093-109489093 on Build GRCh38
4403139 c.921C>T p.L307L Chr.1: 109488984-109488984 on Build GRCh38
3470880 c.947C>T p.S316F Chr.1: 109489010-109489010 on Build GRCh38
3862120 c.932C>T p.S311F Chr.1: 109488995-109488995 on Build GRCh38
1245839 c.1001G>A p.R334H Chr.1: 109489064-109489064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150011982 Chr.1: 109488988-109488988 on Build GRCh38
rs145282300 Chr.1: 109489031-109489031 on Build GRCh38
rs377046901 Chr.1: 109489026-109489026 on Build GRCh38
rs1036105032 Chr.1: 109489118-109489118 on Build GRCh38
rs1002627147 Chr.1: 109489154-109489154 on Build GRCh38
rs773103259 Chr.1: 109488981-109488981 on Build GRCh38
rs140809312 Chr.1: 109489086-109489086 on Build GRCh38
rs374989764 Chr.1: 109489146-109489146 on Build GRCh38
rs749735932 Chr.1: 109489041-109489041 on Build GRCh38
rs748271434 Chr.1: 109489068-109489068 on Build GRCh38
rs746711045 Chr.1: 109489032-109489032 on Build GRCh38
rs563050311 Chr.1: 109489072-109489072 on Build GRCh38
rs887254452 Chr.1: 109489098-109489098 on Build GRCh38
rs369719228 Chr.1: 109489161-109489161 on Build GRCh38
rs560744155 Chr.1: 109489157-109489157 on Build GRCh38
rs148728891 Chr.1: 109488989-109488989 on Build GRCh38
rs760517710 Chr.1: 109488984-109488984 on Build GRCh38
rs780349000 Chr.1: 109489038-109489038 on Build GRCh38
rs958409300 Chr.1: 109489172-109489172 on Build GRCh38
rs766137202 Chr.1: 109488985-109488985 on Build GRCh38
rs766195458 Chr.1: 109489088-109489088 on Build GRCh38
rs751205779 Chr.1: 109489129-109489129 on Build GRCh38
rs569693572 Chr.1: 109489121-109489121 on Build GRCh38
rs757878260 Chr.1: 109489119-109489119 on Build GRCh38
rs774299743 Chr.1: 109488967-109488967 on Build GRCh38
rs141453920 Chr.1: 109489010-109489010 on Build GRCh38
rs759464970 Chr.1: 109489104-109489104 on Build GRCh38
rs776038541 Chr.1: 109488959-109488959 on Build GRCh38
rs368111387 Chr.1: 109489105-109489105 on Build GRCh38
rs374236558 Chr.1: 109489064-109489064 on Build GRCh38
rs752668234 Chr.1: 109489117-109489117 on Build GRCh38
rs763650004 Chr.1: 109489123-109489123 on Build GRCh38
rs370697701 Chr.1: 109488961-109488961 on Build GRCh38
rs756863896 Chr.1: 109489132-109489132 on Build GRCh38
rs372003543 Chr.1: 109489145-109489145 on Build GRCh38
rs527763050 Chr.1: 109489162-109489162 on Build GRCh38
rs768775684 Chr.1: 109488964-109488964 on Build GRCh38
rs539122228 Chr.1: 109489012-109489012 on Build GRCh38
rs913427036 Chr.1: 109489044-109489044 on Build GRCh38
rs779532123 Chr.1: 109489136-109489136 on Build GRCh38
rs372039916 Chr.1: 109489033-109489033 on Build GRCh38
rs865958554 Chr.1: 109489005-109489005 on Build GRCh38
rs944145382 Chr.1: 109488982-109488982 on Build GRCh38
rs986487148 Chr.1: 109489092-109489092 on Build GRCh38
rs749636222 Chr.1: 109489133-109489133 on Build GRCh38
rs767873540 Chr.1: 109488968-109488968 on Build GRCh38
rs755471477 Chr.1: 109489135-109489135 on Build GRCh38
rs764634685 Chr.1: 109489113-109489114 on Build GRCh38
rs764742217 Chr.1: 109488992-109488992 on Build GRCh38
rs773547460 Chr.1: 109489084-109489084 on Build GRCh38
rs546430852 Chr.1: 109489058-109489058 on Build GRCh38

View Full Product Details