Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 51025164-51025405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COBL
Entrez Gene ID
23242
Gene Name
cordon-bleu WH2 repeat protein
Gene Aliases
-
UniGene
Hs.99141
Gene Chromosome Location
Chr.7: 51016212-51316818 on Build GRCh38
Transcripts
NM_001287436.1 NM_015198.3 XM_005271750.1 XM_005271751.1 XM_005271756.1 XM_011515234.1 XM_011515235.1 XM_011515236.1 XM_011515237.1 XM_011515238.1 XM_011515239.1 XM_011515240.1 XM_011515241.1 XM_017011898.1 XM_017011899.1 XM_017011900.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7143465 c.3678C>G p.F1226L Chr.7: 51025199-51025199 on Build GRCh38
4628651 c.3760T>C p.S1254P Chr.7: 51025363-51025363 on Build GRCh38
3881668 c.3703G>A p.A1235T Chr.7: 51025174-51025174 on Build GRCh38
5624668 c.3700A>C p.T1234P Chr.7: 51025177-51025177 on Build GRCh38
6294119 c.3881C>A p.P1294Q Chr.7: 51025242-51025242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778585556 Chr.7: 51025371-51025371 on Build GRCh38
rs149902151 Chr.7: 51025340-51025340 on Build GRCh38
rs1055327304 Chr.7: 51025334-51025334 on Build GRCh38
rs746110032 Chr.7: 51025314-51025314 on Build GRCh38
rs140576062 Chr.7: 51025308-51025308 on Build GRCh38
rs778921084 Chr.7: 51025337-51025337 on Build GRCh38
rs761308896 Chr.7: 51025228-51025228 on Build GRCh38
rs541523816 Chr.7: 51025285-51025285 on Build GRCh38
rs749408434 Chr.7: 51025211-51025211 on Build GRCh38
rs895138378 Chr.7: 51025335-51025335 on Build GRCh38
rs766033325 Chr.7: 51025255-51025255 on Build GRCh38
rs775925510 Chr.7: 51025245-51025245 on Build GRCh38
rs2240087 Chr.7: 51025316-51025316 on Build GRCh38
rs202117145 Chr.7: 51025329-51025329 on Build GRCh38
rs762209143 Chr.7: 51025333-51025333 on Build GRCh38
rs764298231 Chr.7: 51025247-51025247 on Build GRCh38
rs767073730 Chr.7: 51025347-51025347 on Build GRCh38
rs779641267 Chr.7: 51025183-51025183 on Build GRCh38
rs143763360 Chr.7: 51025193-51025193 on Build GRCh38
rs199986848 Chr.7: 51025216-51025216 on Build GRCh38
rs201787564 Chr.7: 51025279-51025279 on Build GRCh38
rs769194381 Chr.7: 51025328-51025328 on Build GRCh38
rs199610932 Chr.7: 51025268-51025268 on Build GRCh38
rs748130662 Chr.7: 51025194-51025194 on Build GRCh38
rs528601904 Chr.7: 51025372-51025372 on Build GRCh38
rs754498434 Chr.7: 51025267-51025267 on Build GRCh38
rs966432121 Chr.7: 51025191-51025191 on Build GRCh38
rs751212887 Chr.7: 51025264-51025264 on Build GRCh38
rs761181233 Chr.7: 51025246-51025246 on Build GRCh38
rs941400845 Chr.7: 51025300-51025300 on Build GRCh38
rs546157122 Chr.7: 51025353-51025353 on Build GRCh38
rs764057362 Chr.7: 51025360-51025360 on Build GRCh38
rs1129262 Chr.7: 51025352-51025352 on Build GRCh38
rs909889035 Chr.7: 51025292-51025292 on Build GRCh38
rs754586195 Chr.7: 51025190-51025190 on Build GRCh38
rs184520448 Chr.7: 51025369-51025369 on Build GRCh38
rs753748024 Chr.7: 51025363-51025363 on Build GRCh38
rs1009719023 Chr.7: 51025244-51025244 on Build GRCh38
rs368735072 Chr.7: 51025322-51025322 on Build GRCh38
rs1010875572 Chr.7: 51025336-51025336 on Build GRCh38
rs774100799 Chr.7: 51025339-51025339 on Build GRCh38
rs745914504 Chr.7: 51025231-51025231 on Build GRCh38
rs200941312 Chr.7: 51025248-51025248 on Build GRCh38
rs555546127 Chr.7: 51025307-51025307 on Build GRCh38
rs139118175 Chr.7: 51025327-51025327 on Build GRCh38
rs147224749 Chr.7: 51025295-51025295 on Build GRCh38
rs767939422 Chr.7: 51025250-51025250 on Build GRCh38
rs201212787 Chr.7: 51025212-51025212 on Build GRCh38
rs557642612 Chr.7: 51025196-51025196 on Build GRCh38
rs201622555 Chr.7: 51025208-51025208 on Build GRCh38
rs751067480 Chr.7: 51025184-51025184 on Build GRCh38
rs772166686 Chr.7: 51025234-51025234 on Build GRCh38
rs991065802 Chr.7: 51025291-51025291 on Build GRCh38

View Full Product Details