Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 7799530-7799757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM261
Entrez Gene ID
90871
Gene Name
transmembrane protein 261
Gene Aliases
C9orf123
UniGene
Hs.7517
Gene Chromosome Location
Chr.9: 7796490-7799806 on Build GRCh38
Transcripts
NM_001318058.1 NM_001318059.1 NM_033428.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6184174 c.144G>T p.W48C Chr.9: 7799591-7799591 on Build GRCh38
6246569 c.46C>A p.P16T Chr.9: 7799689-7799689 on Build GRCh38
1462978 c.181G>A p.A61T Chr.9: 7799554-7799554 on Build GRCh38
5951606 c.116C>T p.A39V Chr.9: 7799619-7799619 on Build GRCh38
6040708 c.70G>T p.A24S Chr.9: 7799665-7799665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771098822 Chr.9: 7799717-7799717 on Build GRCh38
rs200925039 Chr.9: 7799739-7799739 on Build GRCh38
rs754727220 Chr.9: 7799610-7799610 on Build GRCh38
rs760810148 Chr.9: 7799714-7799714 on Build GRCh38
rs761827796 Chr.9: 7799581-7799581 on Build GRCh38
rs757857229 Chr.9: 7799621-7799621 on Build GRCh38
rs770841963 Chr.9: 7799637-7799637 on Build GRCh38
rs756076416 Chr.9: 7799740-7799740 on Build GRCh38
rs1127430 Chr.9: 7799653-7799653 on Build GRCh38
rs372232070 Chr.9: 7799573-7799573 on Build GRCh38
rs764274268 Chr.9: 7799578-7799578 on Build GRCh38
rs14059 Chr.9: 7799606-7799606 on Build GRCh38
rs766971555 Chr.9: 7799665-7799665 on Build GRCh38
rs955200716 Chr.9: 7799642-7799642 on Build GRCh38
rs200927073 Chr.9: 7799673-7799673 on Build GRCh38
rs574961082 Chr.9: 7799736-7799736 on Build GRCh38
rs757632905 Chr.9: 7799646-7799654 on Build GRCh38
rs535287379 Chr.9: 7799613-7799613 on Build GRCh38
rs781664708 Chr.9: 7799648-7799649 on Build GRCh38
rs750910088 Chr.9: 7799586-7799586 on Build GRCh38
rs200162312 Chr.9: 7799685-7799685 on Build GRCh38
rs142833833 Chr.9: 7799700-7799700 on Build GRCh38
rs762557809 Chr.9: 7799732-7799732 on Build GRCh38
rs773341638 Chr.9: 7799659-7799659 on Build GRCh38
rs772052546 Chr.9: 7799656-7799656 on Build GRCh38
rs967981335 Chr.9: 7799654-7799654 on Build GRCh38
rs745755025 Chr.9: 7799696-7799696 on Build GRCh38
rs1005234595 Chr.9: 7799600-7799600 on Build GRCh38
rs747015103 Chr.9: 7799630-7799630 on Build GRCh38
rs139533992 Chr.9: 7799680-7799680 on Build GRCh38
rs982969960 Chr.9: 7799710-7799710 on Build GRCh38
rs376872204 Chr.9: 7799738-7799738 on Build GRCh38
rs776405685 Chr.9: 7799563-7799563 on Build GRCh38
rs765590439 Chr.9: 7799675-7799675 on Build GRCh38
rs748387426 Chr.9: 7799704-7799704 on Build GRCh38
rs754295822 Chr.9: 7799672-7799672 on Build GRCh38
rs756626454 Chr.9: 7799595-7799595 on Build GRCh38
rs769487435 Chr.9: 7799698-7799698 on Build GRCh38
rs146989557 Chr.9: 7799723-7799723 on Build GRCh38
rs778399806 Chr.9: 7799611-7799611 on Build GRCh38
rs779833544 Chr.9: 7799702-7799702 on Build GRCh38
rs753088927 Chr.9: 7799727-7799727 on Build GRCh38
rs950230683 Chr.9: 7799689-7799689 on Build GRCh38
rs557764771 Chr.9: 7799663-7799663 on Build GRCh38
rs758061935 Chr.9: 7799678-7799678 on Build GRCh38
rs140096578 Chr.9: 7799734-7799734 on Build GRCh38
rs149537395 Chr.9: 7799641-7799641 on Build GRCh38
rs765584714 Chr.9: 7799724-7799724 on Build GRCh38
rs745628156 Chr.9: 7799640-7799640 on Build GRCh38
rs773479686 Chr.9: 7799712-7799712 on Build GRCh38
rs376861063 Chr.9: 7799677-7799677 on Build GRCh38
rs368028558 Chr.9: 7799679-7799679 on Build GRCh38
rs781137305 Chr.9: 7799694-7799694 on Build GRCh38
rs553896593 Chr.9: 7799624-7799624 on Build GRCh38
rs774527150 Chr.9: 7799579-7799579 on Build GRCh38
rs757226215 Chr.9: 7799682-7799682 on Build GRCh38
rs777302221 Chr.9: 7799720-7799720 on Build GRCh38
rs772325105 Chr.9: 7799711-7799711 on Build GRCh38
rs766920814 Chr.9: 7799598-7799598 on Build GRCh38

View Full Product Details