Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48298029-48298278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC114
Entrez Gene ID
93233
Gene Name
coiled-coil domain containing 114
Gene Aliases
CILD20
UniGene
Hs.112645
Gene Chromosome Location
Chr.19: 48296452-48322793 on Build GRCh38
Transcripts
NM_144577.3 XM_005259413.3 XM_005259414.3 XM_005259415.3 XM_005259416.4 XM_011527515.2 XM_011527516.2 XM_011527517.1 XM_017027483.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1612514 c.672+9C>T p.? Chr.19: 48298168-48298168 on Build GRCh38
3536473 c.731T>G p.L244R Chr.19: 48298039-48298039 on Build GRCh38
712098 c.1353T>C p.L451L Chr.19: 48298038-48298038 on Build GRCh38
3107165 c.1355C>T p.P452L Chr.19: 48298036-48298036 on Build GRCh38
1304854 c.1263C>A p.L421L Chr.19: 48298207-48298207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141720286 Chr.19: 48298080-48298080 on Build GRCh38
rs200795319 Chr.19: 48298168-48298168 on Build GRCh38
rs745518243 Chr.19: 48298187-48298188 on Build GRCh38
rs559114844 Chr.19: 48298138-48298138 on Build GRCh38
rs772097281 Chr.19: 48298128-48298128 on Build GRCh38
rs143475766 Chr.19: 48298072-48298072 on Build GRCh38
rs746635107 Chr.19: 48298109-48298109 on Build GRCh38
rs199600077 Chr.19: 48298079-48298079 on Build GRCh38
rs201233397 Chr.19: 48298144-48298144 on Build GRCh38
rs377546100 Chr.19: 48298119-48298119 on Build GRCh38
rs8111297 Chr.19: 48298157-48298157 on Build GRCh38
rs768975947 Chr.19: 48298213-48298213 on Build GRCh38
rs1014304171 Chr.19: 48298182-48298182 on Build GRCh38
rs773122864 Chr.19: 48298123-48298123 on Build GRCh38
rs138500585 Chr.19: 48298257-48298257 on Build GRCh38
rs575579848 Chr.19: 48298243-48298243 on Build GRCh38
rs775678608 Chr.19: 48298205-48298205 on Build GRCh38
rs776541413 Chr.19: 48298118-48298118 on Build GRCh38
rs765137896 Chr.19: 48298051-48298051 on Build GRCh38
rs1019912297 Chr.19: 48298192-48298192 on Build GRCh38
rs769435813 Chr.19: 48298229-48298230 on Build GRCh38
rs371519705 Chr.19: 48298167-48298167 on Build GRCh38
rs759627494 Chr.19: 48298125-48298125 on Build GRCh38
rs368172509 Chr.19: 48298184-48298184 on Build GRCh38
rs150259992 Chr.19: 48298077-48298077 on Build GRCh38
rs933511254 Chr.19: 48298156-48298156 on Build GRCh38
rs777717616 Chr.19: 48298177-48298177 on Build GRCh38
rs751149003 Chr.19: 48298245-48298245 on Build GRCh38
rs759247817 Chr.19: 48298190-48298190 on Build GRCh38
rs147082322 Chr.19: 48298256-48298256 on Build GRCh38
rs8111947 Chr.19: 48298169-48298169 on Build GRCh38
rs199822195 Chr.19: 48298221-48298221 on Build GRCh38
rs372851757 Chr.19: 48298053-48298053 on Build GRCh38
rs868247822 Chr.19: 48298076-48298076 on Build GRCh38
rs779554530 Chr.19: 48298083-48298083 on Build GRCh38
rs765660029 Chr.19: 48298220-48298220 on Build GRCh38
rs776657117 Chr.19: 48298214-48298214 on Build GRCh38
rs201704290 Chr.19: 48298120-48298120 on Build GRCh38
rs750890147 Chr.19: 48298145-48298145 on Build GRCh38
rs757921685 Chr.19: 48298099-48298099 on Build GRCh38
rs34816095 Chr.19: 48298160-48298161 on Build GRCh38
rs770833619 Chr.19: 48298201-48298201 on Build GRCh38
rs769522909 Chr.19: 48298170-48298170 on Build GRCh38
rs370797734 Chr.19: 48298108-48298108 on Build GRCh38
rs374202479 Chr.19: 48298115-48298115 on Build GRCh38
rs752848327 Chr.19: 48298068-48298068 on Build GRCh38
rs371473952 Chr.19: 48298137-48298137 on Build GRCh38
rs375854089 Chr.19: 48298054-48298054 on Build GRCh38
rs760663326 Chr.19: 48298211-48298211 on Build GRCh38
rs768293391 Chr.19: 48298230-48298230 on Build GRCh38
rs374508237 Chr.19: 48298130-48298130 on Build GRCh38
rs751016041 Chr.19: 48298164-48298164 on Build GRCh38
rs764190012 Chr.19: 48298141-48298141 on Build GRCh38
rs745426106 Chr.19: 48298095-48298095 on Build GRCh38
rs754292609 Chr.19: 48298143-48298143 on Build GRCh38

View Full Product Details