Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75949471-75949744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOX1
Entrez Gene ID
51
Gene Name
acyl-CoA oxidase 1
Gene Aliases
ACOX, PALMCOX, SCOX
UniGene
Hs.464137
Gene Chromosome Location
Chr.17: 75941507-75979434 on Build GRCh38
Transcripts
NM_001185039.1 NM_004035.6 NM_007292.5 XM_011524868.2 XM_011524869.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6035698 c.1478+9T>A p.? Chr.17: 75949709-75949709 on Build GRCh38
4658895 c.1502_1503insA p.N501fs*22 Chr.17: 75949576-75949577 on Build GRCh38
1727204 c.1570G>T p.V524F Chr.17: 75949509-75949509 on Build GRCh38
4630342 c.1493C>T p.A498V Chr.17: 75949586-75949586 on Build GRCh38
4069735 c.1469G>A p.R490H Chr.17: 75949727-75949727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769829197 Chr.17: 75949652-75949652 on Build GRCh38
rs779728173 Chr.17: 75949621-75949621 on Build GRCh38
rs190584775 Chr.17: 75949704-75949704 on Build GRCh38
rs761772318 Chr.17: 75949546-75949546 on Build GRCh38
rs150349294 Chr.17: 75949514-75949514 on Build GRCh38
rs199908120 Chr.17: 75949705-75949705 on Build GRCh38
rs371908627 Chr.17: 75949642-75949642 on Build GRCh38
rs572144497 Chr.17: 75949573-75949573 on Build GRCh38
rs758806297 Chr.17: 75949689-75949689 on Build GRCh38
rs77139740 Chr.17: 75949559-75949559 on Build GRCh38
rs748052592 Chr.17: 75949650-75949650 on Build GRCh38
rs773019791 Chr.17: 75949684-75949684 on Build GRCh38
rs138050619 Chr.17: 75949547-75949547 on Build GRCh38
rs964973324 Chr.17: 75949562-75949562 on Build GRCh38
rs1042213728 Chr.17: 75949501-75949501 on Build GRCh38
rs562585860 Chr.17: 75949690-75949690 on Build GRCh38
rs749761908 Chr.17: 75949505-75949505 on Build GRCh38
rs201115257 Chr.17: 75949640-75949640 on Build GRCh38
rs368066448 Chr.17: 75949604-75949604 on Build GRCh38
rs768128743 Chr.17: 75949529-75949529 on Build GRCh38
rs754006646 Chr.17: 75949708-75949708 on Build GRCh38
rs372131625 Chr.17: 75949607-75949607 on Build GRCh38
rs778466063 Chr.17: 75949497-75949497 on Build GRCh38
rs976060091 Chr.17: 75949598-75949598 on Build GRCh38
rs756311982 Chr.17: 75949581-75949581 on Build GRCh38
rs775885844 Chr.17: 75949698-75949698 on Build GRCh38
rs776575018 Chr.17: 75949538-75949538 on Build GRCh38
rs542364930 Chr.17: 75949689-75949689 on Build GRCh38
rs771228961 Chr.17: 75949506-75949506 on Build GRCh38
rs540115931 Chr.17: 75949584-75949584 on Build GRCh38
rs750177407 Chr.17: 75949554-75949554 on Build GRCh38
rs760216161 Chr.17: 75949525-75949525 on Build GRCh38
rs8064955 Chr.17: 75949659-75949659 on Build GRCh38
rs758656332 Chr.17: 75949615-75949615 on Build GRCh38
rs768188018 Chr.17: 75949637-75949637 on Build GRCh38
rs758791373 Chr.17: 75949523-75949523 on Build GRCh38
rs755821796 Chr.17: 75949665-75949665 on Build GRCh38
rs746306059 Chr.17: 75949614-75949614 on Build GRCh38
rs1041070293 Chr.17: 75949649-75949649 on Build GRCh38
rs752938493 Chr.17: 75949576-75949576 on Build GRCh38
rs775115439 Chr.17: 75949518-75949518 on Build GRCh38
rs9901420 Chr.17: 75949664-75949664 on Build GRCh38
rs779270053 Chr.17: 75949512-75949512 on Build GRCh38
rs779271033 Chr.17: 75949608-75949608 on Build GRCh38
rs112638345 Chr.17: 75949638-75949638 on Build GRCh38
rs374558539 Chr.17: 75949626-75949626 on Build GRCh38
rs745520282 Chr.17: 75949513-75949513 on Build GRCh38
rs8069565 Chr.17: 75949605-75949605 on Build GRCh38

View Full Product Details