Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 1801471-1801742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA2D4
Entrez Gene ID
93589
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 4
Gene Aliases
RCD4
UniGene
Hs.13768
Gene Chromosome Location
Chr.12: 1791957-1922606 on Build GRCh38
Transcripts
NM_172364.4 XM_011521041.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4958690 c.2764C>T p.Q922* Chr.12: 1801602-1801602 on Build GRCh38
7210194 c.2723C>T p.T908M Chr.12: 1801643-1801643 on Build GRCh38
5560149 c.2771T>G p.L924R Chr.12: 1801595-1801595 on Build GRCh38
2004023 c.2724G>A p.T908T Chr.12: 1801642-1801642 on Build GRCh38
2004019 c.2774G>T p.S925I Chr.12: 1801592-1801592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557477003 Chr.12: 1801700-1801700 on Build GRCh38
rs62621429 Chr.12: 1801620-1801620 on Build GRCh38
rs757410335 Chr.12: 1801527-1801527 on Build GRCh38
rs962058834 Chr.12: 1801663-1801663 on Build GRCh38
rs935694433 Chr.12: 1801508-1801508 on Build GRCh38
rs369468455 Chr.12: 1801627-1801627 on Build GRCh38
rs762559441 Chr.12: 1801642-1801642 on Build GRCh38
rs888444063 Chr.12: 1801617-1801617 on Build GRCh38
rs182211477 Chr.12: 1801706-1801706 on Build GRCh38
rs762460256 Chr.12: 1801688-1801688 on Build GRCh38
rs776204790 Chr.12: 1801663-1801664 on Build GRCh38
rs138140709 Chr.12: 1801716-1801716 on Build GRCh38
rs1054244037 Chr.12: 1801518-1801518 on Build GRCh38
rs575121085 Chr.12: 1801654-1801654 on Build GRCh38
rs186355821 Chr.12: 1801528-1801528 on Build GRCh38
rs189581999 Chr.12: 1801689-1801689 on Build GRCh38
rs372209257 Chr.12: 1801536-1801536 on Build GRCh38
rs553401040 Chr.12: 1801567-1801567 on Build GRCh38
rs768577254 Chr.12: 1801587-1801587 on Build GRCh38
rs771321239 Chr.12: 1801501-1801501 on Build GRCh38
rs750654875 Chr.12: 1801550-1801550 on Build GRCh38
rs749464580 Chr.12: 1801604-1801604 on Build GRCh38
rs550683986 Chr.12: 1801502-1801502 on Build GRCh38
rs886049125 Chr.12: 1801583-1801583 on Build GRCh38
rs372098009 Chr.12: 1801630-1801630 on Build GRCh38
rs967603664 Chr.12: 1801701-1801701 on Build GRCh38
rs557429511 Chr.12: 1801554-1801554 on Build GRCh38
rs199562767 Chr.12: 1801644-1801644 on Build GRCh38
rs779959638 Chr.12: 1801576-1801576 on Build GRCh38
rs571129509 Chr.12: 1801586-1801586 on Build GRCh38
rs553620421 Chr.12: 1801643-1801643 on Build GRCh38
rs575639373 Chr.12: 1801704-1801704 on Build GRCh38
rs761469853 Chr.12: 1801683-1801684 on Build GRCh38
rs568835066 Chr.12: 1801525-1801525 on Build GRCh38
rs994873933 Chr.12: 1801686-1801686 on Build GRCh38
rs780992777 Chr.12: 1801603-1801603 on Build GRCh38
rs754289211 Chr.12: 1801677-1801677 on Build GRCh38

View Full Product Details