Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4288783-4289050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCND2
Entrez Gene ID
894
Gene Name
cyclin D2
Gene Aliases
KIAK0002, MPPH3
UniGene
Hs.376071
Gene Chromosome Location
Chr.12: 4273736-4305356 on Build GRCh38
Transcripts
NM_001759.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7321326 c.610A>T p.T204S Chr.12: 4288880-4288880 on Build GRCh38
5697452 c.701A>T p.K234M Chr.12: 4288971-4288971 on Build GRCh38
5608024 c.578A>G p.K193R Chr.12: 4288848-4288848 on Build GRCh38
6930783 c.717C>A p.D239E Chr.12: 4288987-4288987 on Build GRCh38
7326962 c.611C>G p.T204S Chr.12: 4288881-4288881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562182290 Chr.12: 4288870-4288870 on Build GRCh38
rs755471154 Chr.12: 4289006-4289006 on Build GRCh38
rs766225061 Chr.12: 4288936-4288936 on Build GRCh38
rs932643313 Chr.12: 4288895-4288895 on Build GRCh38
rs752282475 Chr.12: 4288981-4288981 on Build GRCh38
rs759408487 Chr.12: 4288961-4288961 on Build GRCh38
rs758807595 Chr.12: 4288853-4288853 on Build GRCh38
rs139239250 Chr.12: 4288972-4288972 on Build GRCh38
rs535798018 Chr.12: 4288819-4288819 on Build GRCh38
rs770027846 Chr.12: 4288887-4288887 on Build GRCh38
rs781201256 Chr.12: 4289020-4289020 on Build GRCh38
rs1012989280 Chr.12: 4288950-4288950 on Build GRCh38
rs755776688 Chr.12: 4288987-4288987 on Build GRCh38
rs748329879 Chr.12: 4288879-4288879 on Build GRCh38
rs199882809 Chr.12: 4288904-4288904 on Build GRCh38
rs61755286 Chr.12: 4288880-4288880 on Build GRCh38
rs976936457 Chr.12: 4288855-4288855 on Build GRCh38
rs934573100 Chr.12: 4289014-4289014 on Build GRCh38
rs758862176 Chr.12: 4288994-4288994 on Build GRCh38
rs915300093 Chr.12: 4288969-4288969 on Build GRCh38
rs369033113 Chr.12: 4288804-4288804 on Build GRCh38
rs780410687 Chr.12: 4288997-4288997 on Build GRCh38
rs567626308 Chr.12: 4288838-4288838 on Build GRCh38
rs376238740 Chr.12: 4289005-4289005 on Build GRCh38
rs747054950 Chr.12: 4288867-4288867 on Build GRCh38
rs769975071 Chr.12: 4289024-4289024 on Build GRCh38
rs199552192 Chr.12: 4288876-4288876 on Build GRCh38
rs780649921 Chr.12: 4288854-4288854 on Build GRCh38
rs373099767 Chr.12: 4288988-4288988 on Build GRCh38
rs375356578 Chr.12: 4288957-4288957 on Build GRCh38
rs748277013 Chr.12: 4289021-4289021 on Build GRCh38
rs202056442 Chr.12: 4288898-4288898 on Build GRCh38
rs777949060 Chr.12: 4288992-4289014 on Build GRCh38
rs779377897 Chr.12: 4288834-4288834 on Build GRCh38
rs750783951 Chr.12: 4288989-4288989 on Build GRCh38
rs762849458 Chr.12: 4288935-4288935 on Build GRCh38

View Full Product Details