Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27222105-27222364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAD
Entrez Gene ID
790
Gene Name
carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase
Gene Aliases
CDG1Z
UniGene
Hs.377010
Gene Chromosome Location
Chr.2: 27217257-27243792 on Build GRCh38
Transcripts
NM_001306079.1 NM_004341.4 XM_005264555.3 XM_006712101.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5661571 c.474G>T p.L158L Chr.2: 27222315-27222315 on Build GRCh38
5987270 c.425G>C p.G142A Chr.2: 27222266-27222266 on Build GRCh38
4093500 c.434C>A p.P145H Chr.2: 27222275-27222275 on Build GRCh38
3962729 c.442C>T p.L148L Chr.2: 27222283-27222283 on Build GRCh38
3962730 c.444G>T p.L148L Chr.2: 27222285-27222285 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758733421 Chr.2: 27222178-27222179 on Build GRCh38
rs953388667 Chr.2: 27222137-27222137 on Build GRCh38
rs781212395 Chr.2: 27222299-27222299 on Build GRCh38
rs773521281 Chr.2: 27222220-27222220 on Build GRCh38
rs747221303 Chr.2: 27222208-27222208 on Build GRCh38
rs147103288 Chr.2: 27222308-27222308 on Build GRCh38
rs768762689 Chr.2: 27222209-27222209 on Build GRCh38
rs1048689342 Chr.2: 27222233-27222233 on Build GRCh38
rs767215684 Chr.2: 27222331-27222331 on Build GRCh38
rs374839707 Chr.2: 27222227-27222227 on Build GRCh38
rs539232010 Chr.2: 27222216-27222216 on Build GRCh38
rs371844501 Chr.2: 27222182-27222182 on Build GRCh38
rs150948363 Chr.2: 27222241-27222241 on Build GRCh38
rs778646517 Chr.2: 27222285-27222285 on Build GRCh38
rs145509871 Chr.2: 27222235-27222235 on Build GRCh38
rs753772157 Chr.2: 27222270-27222270 on Build GRCh38
rs372965533 Chr.2: 27222298-27222298 on Build GRCh38
rs780733991 Chr.2: 27222148-27222148 on Build GRCh38
rs771465104 Chr.2: 27222316-27222316 on Build GRCh38
rs774890129 Chr.2: 27222327-27222327 on Build GRCh38
rs369739874 Chr.2: 27222343-27222343 on Build GRCh38
rs749123379 Chr.2: 27222183-27222183 on Build GRCh38
rs745620112 Chr.2: 27222297-27222297 on Build GRCh38
rs759235850 Chr.2: 27222232-27222232 on Build GRCh38
rs756979309 Chr.2: 27222199-27222199 on Build GRCh38
rs147694325 Chr.2: 27222302-27222302 on Build GRCh38
rs370344157 Chr.2: 27222288-27222288 on Build GRCh38
rs762394968 Chr.2: 27222218-27222218 on Build GRCh38
rs948293256 Chr.2: 27222255-27222255 on Build GRCh38
rs769754191 Chr.2: 27222307-27222307 on Build GRCh38
rs759935188 Chr.2: 27222329-27222329 on Build GRCh38
rs138979522 Chr.2: 27222276-27222276 on Build GRCh38
rs749788077 Chr.2: 27222311-27222311 on Build GRCh38
rs906691445 Chr.2: 27222279-27222279 on Build GRCh38
rs922057070 Chr.2: 27222249-27222249 on Build GRCh38
rs760300403 Chr.2: 27222333-27222333 on Build GRCh38
rs145371458 Chr.2: 27222223-27222223 on Build GRCh38
rs764098153 Chr.2: 27222260-27222260 on Build GRCh38
rs754590127 Chr.2: 27222146-27222146 on Build GRCh38
rs892516093 Chr.2: 27222319-27222319 on Build GRCh38
rs766834759 Chr.2: 27222226-27222226 on Build GRCh38
rs752277453 Chr.2: 27222237-27222237 on Build GRCh38
rs963908880 Chr.2: 27222229-27222229 on Build GRCh38

View Full Product Details