Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112186700-112186973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf32
Entrez Gene ID
132720
Gene Name
chromosome 4 open reading frame 32
Gene Aliases
-
UniGene
Hs.23439
Gene Chromosome Location
Chr.4: 112145397-112189081 on Build GRCh38
Transcripts
NM_152400.2 XM_017007732.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4187138 c.270G>A p.M90I Chr.4: 112186809-112186809 on Build GRCh38
3824962 c.206A>C p.K69T Chr.4: 112186745-112186745 on Build GRCh38
7162012 c.272A>G p.Y91C Chr.4: 112186811-112186811 on Build GRCh38
6866411 c.232C>T p.L78L Chr.4: 112186771-112186771 on Build GRCh38
3599170 c.312C>T p.F104F Chr.4: 112186851-112186851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977682754 Chr.4: 112186839-112186839 on Build GRCh38
rs200706775 Chr.4: 112186727-112186727 on Build GRCh38
rs923640375 Chr.4: 112186866-112186866 on Build GRCh38
rs777862360 Chr.4: 112186931-112186931 on Build GRCh38
rs772344788 Chr.4: 112186803-112186803 on Build GRCh38
rs751196818 Chr.4: 112186881-112186881 on Build GRCh38
rs199842368 Chr.4: 112186911-112186911 on Build GRCh38
rs958745483 Chr.4: 112186773-112186773 on Build GRCh38
rs761025644 Chr.4: 112186817-112186817 on Build GRCh38
rs765877248 Chr.4: 112186869-112186869 on Build GRCh38
rs1050714162 Chr.4: 112186912-112186912 on Build GRCh38
rs144235488 Chr.4: 112186909-112186909 on Build GRCh38
rs538521310 Chr.4: 112186814-112186814 on Build GRCh38
rs754286283 Chr.4: 112186830-112186830 on Build GRCh38
rs1026262414 Chr.4: 112186761-112186761 on Build GRCh38
rs777845898 Chr.4: 112186740-112186740 on Build GRCh38
rs571639524 Chr.4: 112186934-112186934 on Build GRCh38
rs1034406130 Chr.4: 112186763-112186763 on Build GRCh38
rs11548409 Chr.4: 112186733-112186733 on Build GRCh38
rs914714979 Chr.4: 112186802-112186802 on Build GRCh38
rs866694073 Chr.4: 112186776-112186776 on Build GRCh38
rs889540759 Chr.4: 112186915-112186915 on Build GRCh38
rs376066122 Chr.4: 112186780-112186780 on Build GRCh38
rs866555738 Chr.4: 112186823-112186823 on Build GRCh38
rs764644257 Chr.4: 112186818-112186818 on Build GRCh38
rs139612441 Chr.4: 112186908-112186908 on Build GRCh38
rs200998231 Chr.4: 112186851-112186851 on Build GRCh38
rs755957136 Chr.4: 112186929-112186929 on Build GRCh38
rs774436785 Chr.4: 112186778-112186778 on Build GRCh38
rs146826929 Chr.4: 112186757-112186757 on Build GRCh38
rs770981967 Chr.4: 112186771-112186771 on Build GRCh38
rs754711443 Chr.4: 112186884-112186884 on Build GRCh38
rs747936613 Chr.4: 112186728-112186728 on Build GRCh38
rs774334194 Chr.4: 112186787-112186787 on Build GRCh38
rs749433746 Chr.4: 112186760-112186760 on Build GRCh38

View Full Product Details