Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119582079-119582312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADPRH
Entrez Gene ID
141
Gene Name
ADP-ribosylarginine hydrolase
Gene Aliases
ARH1
UniGene
Hs.99884
Gene Chromosome Location
Chr.3: 119579433-119589954 on Build GRCh38
Transcripts
NM_001125.3 NM_001291949.1 NM_001291950.1 XM_011512438.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
728055 c.31A>C p.S11R Chr.3: 119582200-119582200 on Build GRCh38
3727346 c.76T>C p.F26L Chr.3: 119582245-119582245 on Build GRCh38
3585753 c.71G>A p.W24* Chr.3: 119582240-119582240 on Build GRCh38
6595346 c.18T>C p.A6A Chr.3: 119582187-119582187 on Build GRCh38
728057 c.30G>T p.L10L Chr.3: 119582199-119582199 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755537082 Chr.3: 119582238-119582238 on Build GRCh38
rs764111355 Chr.3: 119582186-119582187 on Build GRCh38
rs368027725 Chr.3: 119582271-119582271 on Build GRCh38
rs369666515 Chr.3: 119582129-119582129 on Build GRCh38
rs780528254 Chr.3: 119582231-119582231 on Build GRCh38
rs925917049 Chr.3: 119582260-119582260 on Build GRCh38
rs371536198 Chr.3: 119582141-119582141 on Build GRCh38
rs753907297 Chr.3: 119582188-119582189 on Build GRCh38
rs754354521 Chr.3: 119582235-119582235 on Build GRCh38
rs370199855 Chr.3: 119582185-119582185 on Build GRCh38
rs752569734 Chr.3: 119582218-119582218 on Build GRCh38
rs758479726 Chr.3: 119582272-119582272 on Build GRCh38
rs371596030 Chr.3: 119582143-119582143 on Build GRCh38
rs764982975 Chr.3: 119582214-119582214 on Build GRCh38
rs368811348 Chr.3: 119582276-119582277 on Build GRCh38
rs745753637 Chr.3: 119582273-119582273 on Build GRCh38
rs763608980 Chr.3: 119582229-119582229 on Build GRCh38
rs769794255 Chr.3: 119582288-119582288 on Build GRCh38
rs757329794 Chr.3: 119582125-119582125 on Build GRCh38
rs775121593 Chr.3: 119582193-119582193 on Build GRCh38
rs750439969 Chr.3: 119582221-119582221 on Build GRCh38
rs750525007 Chr.3: 119582127-119582127 on Build GRCh38
rs748554433 Chr.3: 119582250-119582250 on Build GRCh38
rs762491673 Chr.3: 119582207-119582207 on Build GRCh38
rs764928020 Chr.3: 119582159-119582159 on Build GRCh38
rs772786039 Chr.3: 119582191-119582191 on Build GRCh38
rs750303103 Chr.3: 119582163-119582163 on Build GRCh38
rs762746053 Chr.3: 119582293-119582293 on Build GRCh38
rs768962047 Chr.3: 119582149-119582149 on Build GRCh38
rs147901245 Chr.3: 119582225-119582225 on Build GRCh38
rs779094664 Chr.3: 119582248-119582248 on Build GRCh38
rs759350893 Chr.3: 119582209-119582209 on Build GRCh38
rs781209289 Chr.3: 119582126-119582126 on Build GRCh38
rs201822828 Chr.3: 119582266-119582266 on Build GRCh38
rs146571791 Chr.3: 119582169-119582169 on Build GRCh38
rs1019270358 Chr.3: 119582112-119582112 on Build GRCh38
rs762243117 Chr.3: 119582269-119582283 on Build GRCh38
rs20568 Chr.3: 119582292-119582292 on Build GRCh38
rs377477506 Chr.3: 119582253-119582253 on Build GRCh38
rs761975944 Chr.3: 119582219-119582219 on Build GRCh38
rs778594436 Chr.3: 119582154-119582154 on Build GRCh38

View Full Product Details