Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38765327-38765586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR5
Entrez Gene ID
79913
Gene Name
ARP5 actin-related protein 5 homolog
Gene Aliases
Arp5, INO80M
UniGene
Hs.371585
Gene Chromosome Location
Chr.20: 38748454-38772446 on Build GRCh38
Transcripts
NM_024855.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5947750 c.1202C>T p.P401L Chr.20: 38765427-38765427 on Build GRCh38
135879 c.1205C>T p.S402F Chr.20: 38765430-38765430 on Build GRCh38
227168 c.1240C>T p.P414S Chr.20: 38765465-38765465 on Build GRCh38
4462322 c.1241C>T p.P414L Chr.20: 38765466-38765466 on Build GRCh38
6593126 c.1278G>A p.P426P Chr.20: 38765503-38765503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201347262 Chr.20: 38765457-38765457 on Build GRCh38
rs772472184 Chr.20: 38765493-38765493 on Build GRCh38
rs773398124 Chr.20: 38765416-38765416 on Build GRCh38
rs765871079 Chr.20: 38765542-38765542 on Build GRCh38
rs768436470 Chr.20: 38765392-38765392 on Build GRCh38
rs764766327 Chr.20: 38765445-38765445 on Build GRCh38
rs754313094 Chr.20: 38765524-38765524 on Build GRCh38
rs991989205 Chr.20: 38765506-38765506 on Build GRCh38
rs200626089 Chr.20: 38765393-38765393 on Build GRCh38
rs759756974 Chr.20: 38765485-38765485 on Build GRCh38
rs770050639 Chr.20: 38765399-38765399 on Build GRCh38
rs781201417 Chr.20: 38765559-38765559 on Build GRCh38
rs754647483 Chr.20: 38765546-38765546 on Build GRCh38
rs759825005 Chr.20: 38765440-38765440 on Build GRCh38
rs926370286 Chr.20: 38765463-38765463 on Build GRCh38
rs878912896 Chr.20: 38765464-38765464 on Build GRCh38
rs767830598 Chr.20: 38765442-38765442 on Build GRCh38
rs757916197 Chr.20: 38765548-38765548 on Build GRCh38
rs762373580 Chr.20: 38765536-38765536 on Build GRCh38
rs749930437 Chr.20: 38765447-38765447 on Build GRCh38
rs373486054 Chr.20: 38765451-38765451 on Build GRCh38
rs761236439 Chr.20: 38765502-38765502 on Build GRCh38
rs374665514 Chr.20: 38765427-38765427 on Build GRCh38
rs951815903 Chr.20: 38765434-38765434 on Build GRCh38
rs753222436 Chr.20: 38765444-38765444 on Build GRCh38
rs534821560 Chr.20: 38765545-38765545 on Build GRCh38
rs761735427 Chr.20: 38765510-38765510 on Build GRCh38
rs376034458 Chr.20: 38765476-38765476 on Build GRCh38
rs199604949 Chr.20: 38765470-38765470 on Build GRCh38
rs772578329 Chr.20: 38765525-38765525 on Build GRCh38
rs748139042 Chr.20: 38765466-38765466 on Build GRCh38
rs142246739 Chr.20: 38765438-38765438 on Build GRCh38
rs996452108 Chr.20: 38765541-38765541 on Build GRCh38
rs746602299 Chr.20: 38765449-38765449 on Build GRCh38
rs369247942 Chr.20: 38765448-38765448 on Build GRCh38
rs745568810 Chr.20: 38765367-38765367 on Build GRCh38
rs200033323 Chr.20: 38765565-38765565 on Build GRCh38
rs766497395 Chr.20: 38765428-38765428 on Build GRCh38
rs748052809 Chr.20: 38765398-38765398 on Build GRCh38
rs749341643 Chr.20: 38765475-38765475 on Build GRCh38
rs778343960 Chr.20: 38765357-38765357 on Build GRCh38
rs774470082 Chr.20: 38765484-38765484 on Build GRCh38
rs778061414 Chr.20: 38765474-38765474 on Build GRCh38
rs748457186 Chr.20: 38765528-38765528 on Build GRCh38
rs765585565 Chr.20: 38765379-38765379 on Build GRCh38
rs10485651 Chr.20: 38765389-38765389 on Build GRCh38
rs900729927 Chr.20: 38765503-38765503 on Build GRCh38
rs138160614 Chr.20: 38765391-38765391 on Build GRCh38
rs775800510 Chr.20: 38765497-38765497 on Build GRCh38
rs866114656 Chr.20: 38765501-38765501 on Build GRCh38
rs959152053 Chr.20: 38765486-38765486 on Build GRCh38

View Full Product Details