Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116340651-116340876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC172
Entrez Gene ID
374355
Gene Name
coiled-coil domain containing 172
Gene Aliases
C10orf96
UniGene
Hs.233407
Gene Chromosome Location
Chr.10: 116324097-116380029 on Build GRCh38
Transcripts
NM_198515.2 XM_006717820.3 XM_011539771.2 XM_017016206.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5521548 c.203T>G p.L68R Chr.10: 116340771-116340771 on Build GRCh38
5969672 c.211G>T p.A71S Chr.10: 116340779-116340779 on Build GRCh38
7299471 c.189C>T p.S63S Chr.10: 116340757-116340757 on Build GRCh38
1728509 c.180T>A p.F60L Chr.10: 116340748-116340748 on Build GRCh38
1474381 c.207G>C p.L69F Chr.10: 116340775-116340775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937701722 Chr.10: 116340758-116340758 on Build GRCh38
rs377376030 Chr.10: 116340847-116340847 on Build GRCh38
rs773084922 Chr.10: 116340716-116340716 on Build GRCh38
rs373054155 Chr.10: 116340786-116340786 on Build GRCh38
rs775943821 Chr.10: 116340792-116340792 on Build GRCh38
rs749432649 Chr.10: 116340796-116340796 on Build GRCh38
rs137998964 Chr.10: 116340746-116340746 on Build GRCh38
rs981898413 Chr.10: 116340709-116340709 on Build GRCh38
rs760535953 Chr.10: 116340825-116340825 on Build GRCh38
rs200926954 Chr.10: 116340816-116340816 on Build GRCh38
rs752211066 Chr.10: 116340740-116340740 on Build GRCh38
rs752662578 Chr.10: 116340835-116340835 on Build GRCh38
rs766286879 Chr.10: 116340827-116340827 on Build GRCh38
rs757708235 Chr.10: 116340770-116340770 on Build GRCh38
rs768604566 Chr.10: 116340809-116340809 on Build GRCh38
rs781469666 Chr.10: 116340775-116340775 on Build GRCh38
rs373300112 Chr.10: 116340731-116340731 on Build GRCh38
rs746631943 Chr.10: 116340764-116340764 on Build GRCh38
rs746390087 Chr.10: 116340783-116340783 on Build GRCh38
rs774412345 Chr.10: 116340688-116340688 on Build GRCh38
rs766082125 Chr.10: 116340719-116340719 on Build GRCh38
rs753826983 Chr.10: 116340721-116340721 on Build GRCh38
rs189853450 Chr.10: 116340807-116340807 on Build GRCh38
rs117513116 Chr.10: 116340756-116340756 on Build GRCh38
rs145954478 Chr.10: 116340826-116340826 on Build GRCh38
rs760576448 Chr.10: 116340700-116340700 on Build GRCh38
rs748942710 Chr.10: 116340757-116340759 on Build GRCh38
rs1018903190 Chr.10: 116340734-116340734 on Build GRCh38
rs764859297 Chr.10: 116340736-116340736 on Build GRCh38
rs754900913 Chr.10: 116340723-116340723 on Build GRCh38
rs770238940 Chr.10: 116340791-116340791 on Build GRCh38
rs765308958 Chr.10: 116340834-116340834 on Build GRCh38
rs757988499 Chr.10: 116340836-116340836 on Build GRCh38
rs759574921 Chr.10: 116340832-116340832 on Build GRCh38
rs1030789797 Chr.10: 116340755-116340755 on Build GRCh38
rs776516928 Chr.10: 116340828-116340828 on Build GRCh38
rs1005431715 Chr.10: 116340846-116340846 on Build GRCh38
rs779626879 Chr.10: 116340743-116340743 on Build GRCh38

View Full Product Details