Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34268892-34269165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHCY
Entrez Gene ID
191
Gene Name
adenosylhomocysteinase
Gene Aliases
SAHH, adoHcyase
UniGene
Hs.388004
Gene Chromosome Location
Chr.20: 34235012-34311976 on Build GRCh38
Transcripts
XM_011528657.2 NM_001672.2 XM_005260412.3 XM_011528820.2 XM_011528821.1

Gene Symbol
ASIP
Entrez Gene ID
434
Gene Name
agouti signaling protein
Gene Aliases
AGSW, AGTI, AGTIL, ASP, SHEP9
UniGene
Hs.659995
Gene Chromosome Location
Chr.20: 34186493-34269344 on Build GRCh38
Transcripts
XM_011528657.2 NM_001672.2 XM_005260412.3 XM_011528820.2 XM_011528821.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1411285 c.290G>A p.R97H Chr.20: 34269058-34269058 on Build GRCh38
3545524 c.262C>T p.P88S Chr.20: 34269030-34269030 on Build GRCh38
4807896 c.264C>A p.P88P Chr.20: 34269032-34269032 on Build GRCh38
7356609 c.294C>A p.N98K Chr.20: 34269062-34269062 on Build GRCh38
4937386 c.256C>A p.R86R Chr.20: 34269024-34269024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749841638 Chr.20: 34268945-34268945 on Build GRCh38
rs567393493 Chr.20: 34268950-34268950 on Build GRCh38
rs758913102 Chr.20: 34269008-34269008 on Build GRCh38
rs955878401 Chr.20: 34269100-34269100 on Build GRCh38
rs775299058 Chr.20: 34269081-34269081 on Build GRCh38
rs1032902529 Chr.20: 34269095-34269095 on Build GRCh38
rs1000123331 Chr.20: 34269080-34269080 on Build GRCh38
rs780888249 Chr.20: 34269030-34269030 on Build GRCh38
rs772599508 Chr.20: 34269101-34269125 on Build GRCh38
rs776744511 Chr.20: 34269058-34269058 on Build GRCh38
rs369598328 Chr.20: 34268998-34268998 on Build GRCh38
rs988622815 Chr.20: 34269114-34269114 on Build GRCh38
rs761605725 Chr.20: 34269136-34269136 on Build GRCh38
rs201107532 Chr.20: 34268954-34268954 on Build GRCh38
rs937888696 Chr.20: 34269117-34269117 on Build GRCh38
rs769701477 Chr.20: 34269076-34269076 on Build GRCh38
rs1036765174 Chr.20: 34268941-34268941 on Build GRCh38
rs767121271 Chr.20: 34269028-34269028 on Build GRCh38
rs768466168 Chr.20: 34269125-34269125 on Build GRCh38
rs763823641 Chr.20: 34269121-34269121 on Build GRCh38
rs548682853 Chr.20: 34268926-34268926 on Build GRCh38
rs576337181 Chr.20: 34269055-34269055 on Build GRCh38
rs761208609 Chr.20: 34268989-34268991 on Build GRCh38
rs940164108 Chr.20: 34268933-34268933 on Build GRCh38
rs759460072 Chr.20: 34269065-34269065 on Build GRCh38
rs76347145 Chr.20: 34268972-34268972 on Build GRCh38
rs554065691 Chr.20: 34269028-34269028 on Build GRCh38
rs748304026 Chr.20: 34268973-34268973 on Build GRCh38
rs373157539 Chr.20: 34268958-34268958 on Build GRCh38
rs375221899 Chr.20: 34268977-34268977 on Build GRCh38
rs760690218 Chr.20: 34268982-34268982 on Build GRCh38
rs577827889 Chr.20: 34268974-34268974 on Build GRCh38
rs762757094 Chr.20: 34269098-34269098 on Build GRCh38
rs763741682 Chr.20: 34269022-34269022 on Build GRCh38
rs1021484408 Chr.20: 34269118-34269118 on Build GRCh38
rs965482419 Chr.20: 34269122-34269122 on Build GRCh38
rs36093428 Chr.20: 34269032-34269032 on Build GRCh38
rs772338707 Chr.20: 34269033-34269033 on Build GRCh38
rs767121119 Chr.20: 34269143-34269143 on Build GRCh38
rs758128920 Chr.20: 34269021-34269021 on Build GRCh38
rs752600295 Chr.20: 34269013-34269013 on Build GRCh38
rs574186399 Chr.20: 34268993-34268993 on Build GRCh38
rs770970694 Chr.20: 34269056-34269056 on Build GRCh38
rs766471312 Chr.20: 34268987-34268987 on Build GRCh38
rs554657361 Chr.20: 34269037-34269037 on Build GRCh38
rs538816237 Chr.20: 34268991-34268991 on Build GRCh38
rs964902549 Chr.20: 34268935-34268935 on Build GRCh38

View Full Product Details