Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64923810-64924045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHURC1
Entrez Gene ID
91612
Gene Name
churchill domain containing 1
Gene Aliases
C14orf52, My015, chch
UniGene
Hs.731537
Gene Chromosome Location
Chr.14: 64914361-64935368 on Build GRCh38
Transcripts
NM_001204063.1 NM_001204064.1 NM_145165.3 NM_001202558.1 NM_001202559.1

Gene Symbol
CHURC1-FNTB
Entrez Gene ID
100529261
Gene Name
CHURC1-FNTB readthrough
Gene Aliases
-
UniGene
Hs.325531
Gene Chromosome Location
Chr.14: 64914361-65062655 on Build GRCh38
Transcripts
NM_001204063.1 NM_001204064.1 NM_145165.3 NM_001202558.1 NM_001202559.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7200865 c.121-10G>T p.? Chr.14: 64923981-64923981 on Build GRCh38
1207381 c.67T>G p.F23V Chr.14: 64924018-64924018 on Build GRCh38
4672412 c.156G>A p.L52L Chr.14: 64924026-64924026 on Build GRCh38
7200866 c.40-10G>T p.? Chr.14: 64923981-64923981 on Build GRCh38
2248764 c.69C>G p.F23L Chr.14: 64924020-64924020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039826798 Chr.14: 64923955-64923955 on Build GRCh38
rs147829082 Chr.14: 64924011-64924011 on Build GRCh38
rs899583275 Chr.14: 64923990-64923990 on Build GRCh38
rs929530061 Chr.14: 64923911-64923911 on Build GRCh38
rs761495524 Chr.14: 64923972-64923972 on Build GRCh38
rs1007739062 Chr.14: 64923934-64923934 on Build GRCh38
rs950295382 Chr.14: 64923933-64923933 on Build GRCh38
rs919289837 Chr.14: 64923864-64923864 on Build GRCh38
rs751686250 Chr.14: 64923984-64923985 on Build GRCh38
rs781747031 Chr.14: 64923995-64923995 on Build GRCh38
rs1033088131 Chr.14: 64923998-64923998 on Build GRCh38
rs753096380 Chr.14: 64924003-64924003 on Build GRCh38
rs750019122 Chr.14: 64923984-64923984 on Build GRCh38
rs185853369 Chr.14: 64923915-64923915 on Build GRCh38
rs887920572 Chr.14: 64923924-64923924 on Build GRCh38
rs1055948397 Chr.14: 64923874-64923874 on Build GRCh38
rs557991248 Chr.14: 64924018-64924018 on Build GRCh38
rs920324594 Chr.14: 64923858-64923858 on Build GRCh38
rs778045732 Chr.14: 64924016-64924016 on Build GRCh38
rs753541251 Chr.14: 64923964-64923964 on Build GRCh38
rs371958910 Chr.14: 64923980-64923980 on Build GRCh38
rs376584321 Chr.14: 64923992-64923992 on Build GRCh38
rs930811219 Chr.14: 64923868-64923868 on Build GRCh38
rs545691848 Chr.14: 64923859-64923859 on Build GRCh38

View Full Product Details