Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36244696-36244955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAR2
Entrez Gene ID
25980
Gene Name
AAR2 splicing factor homolog
Gene Aliases
C20orf4, CGI-23
UniGene
Hs.11314
Gene Chromosome Location
Chr.20: 36236136-36256941 on Build GRCh38
Transcripts
NM_001271874.1 NM_015511.4 XM_006723770.3 XM_011528762.2 XM_011528763.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5527703 c.929C>T p.P310L Chr.20: 36244868-36244868 on Build GRCh38
5552056 c.962A>G p.D321G Chr.20: 36244901-36244901 on Build GRCh38
4954503 c.902T>G p.I301S Chr.20: 36244841-36244841 on Build GRCh38
301830 c.930C>A p.P310P Chr.20: 36244869-36244869 on Build GRCh38
6589281 c.857C>A p.A286E Chr.20: 36244796-36244796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746573248 Chr.20: 36244909-36244909 on Build GRCh38
rs539592841 Chr.20: 36244896-36244896 on Build GRCh38
rs377157882 Chr.20: 36244889-36244889 on Build GRCh38
rs1006847964 Chr.20: 36244818-36244818 on Build GRCh38
rs983831798 Chr.20: 36244919-36244919 on Build GRCh38
rs372921853 Chr.20: 36244743-36244743 on Build GRCh38
rs140703484 Chr.20: 36244794-36244794 on Build GRCh38
rs113156393 Chr.20: 36244933-36244933 on Build GRCh38
rs989159070 Chr.20: 36244916-36244916 on Build GRCh38
rs904578033 Chr.20: 36244770-36244770 on Build GRCh38
rs138478673 Chr.20: 36244824-36244824 on Build GRCh38
rs778970346 Chr.20: 36244737-36244737 on Build GRCh38
rs376767059 Chr.20: 36244863-36244863 on Build GRCh38
rs1001115790 Chr.20: 36244783-36244783 on Build GRCh38
rs761398969 Chr.20: 36244923-36244923 on Build GRCh38
rs117200013 Chr.20: 36244833-36244833 on Build GRCh38
rs202224152 Chr.20: 36244823-36244823 on Build GRCh38
rs750451424 Chr.20: 36244739-36244739 on Build GRCh38
rs374099380 Chr.20: 36244883-36244883 on Build GRCh38
rs758937745 Chr.20: 36244802-36244802 on Build GRCh38
rs1042212591 Chr.20: 36244757-36244757 on Build GRCh38
rs775133590 Chr.20: 36244908-36244908 on Build GRCh38
rs150912491 Chr.20: 36244744-36244744 on Build GRCh38
rs550754862 Chr.20: 36244848-36244848 on Build GRCh38
rs369290761 Chr.20: 36244869-36244869 on Build GRCh38
rs750338713 Chr.20: 36244856-36244856 on Build GRCh38
rs776383295 Chr.20: 36244917-36244917 on Build GRCh38
rs781449292 Chr.20: 36244870-36244870 on Build GRCh38
rs148948059 Chr.20: 36244924-36244924 on Build GRCh38
rs145635219 Chr.20: 36244766-36244766 on Build GRCh38
rs758286265 Chr.20: 36244857-36244857 on Build GRCh38
rs768875388 Chr.20: 36244773-36244773 on Build GRCh38
rs768398840 Chr.20: 36244911-36244911 on Build GRCh38
rs144176613 Chr.20: 36244819-36244820 on Build GRCh38
rs147679615 Chr.20: 36244787-36244787 on Build GRCh38
rs748440774 Chr.20: 36244881-36244881 on Build GRCh38
rs781465836 Chr.20: 36244786-36244786 on Build GRCh38
rs901436722 Chr.20: 36244746-36244746 on Build GRCh38
rs201599475 Chr.20: 36244882-36244882 on Build GRCh38
rs763908537 Chr.20: 36244825-36244825 on Build GRCh38
rs917276647 Chr.20: 36244738-36244738 on Build GRCh38
rs773817924 Chr.20: 36244801-36244801 on Build GRCh38

View Full Product Details