Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31462517-31462756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC5
Entrez Gene ID
79798
Gene Name
armadillo repeat containing 5
Gene Aliases
AIMAH2
UniGene
Hs.732945
Gene Chromosome Location
Chr.16: 31458273-31467167 on Build GRCh38
Transcripts
NM_001105247.1 NM_001288767.1 NM_001301820.1 NM_024742.2 XM_006721091.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7263877 c.1070G>A p.R357H Chr.16: 31462617-31462617 on Build GRCh38
6078981 c.1085G>C p.R362P Chr.16: 31462632-31462632 on Build GRCh38
4460433 c.1165C>T p.R389C Chr.16: 31462712-31462712 on Build GRCh38
1588445 c.1038G>A p.Q346Q Chr.16: 31462585-31462585 on Build GRCh38
3707023 c.1099G>T p.D367Y Chr.16: 31462646-31462646 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755134664 Chr.16: 31462568-31462568 on Build GRCh38
rs781382967 Chr.16: 31462570-31462570 on Build GRCh38
rs374639681 Chr.16: 31462616-31462616 on Build GRCh38
rs767757331 Chr.16: 31462700-31462700 on Build GRCh38
rs751777905 Chr.16: 31462562-31462562 on Build GRCh38
rs777575042 Chr.16: 31462725-31462725 on Build GRCh38
rs750797663 Chr.16: 31462550-31462550 on Build GRCh38
rs774899884 Chr.16: 31462649-31462649 on Build GRCh38
rs587777663 Chr.16: 31462641-31462641 on Build GRCh38
rs777214700 Chr.16: 31462541-31462541 on Build GRCh38
rs541189681 Chr.16: 31462578-31462578 on Build GRCh38
rs371525408 Chr.16: 31462692-31462692 on Build GRCh38
rs574169297 Chr.16: 31462548-31462548 on Build GRCh38
rs778548538 Chr.16: 31462621-31462621 on Build GRCh38
rs1039442244 Chr.16: 31462554-31462554 on Build GRCh38
rs1022188408 Chr.16: 31462674-31462674 on Build GRCh38
rs746823662 Chr.16: 31462734-31462735 on Build GRCh38
rs79238971 Chr.16: 31462586-31462586 on Build GRCh38
rs1037784138 Chr.16: 31462542-31462542 on Build GRCh38
rs373906994 Chr.16: 31462691-31462691 on Build GRCh38
rs890718116 Chr.16: 31462681-31462681 on Build GRCh38
rs201395563 Chr.16: 31462694-31462694 on Build GRCh38
rs745472071 Chr.16: 31462638-31462638 on Build GRCh38
rs753715053 Chr.16: 31462734-31462734 on Build GRCh38
rs771598546 Chr.16: 31462640-31462640 on Build GRCh38
rs752777691 Chr.16: 31462702-31462702 on Build GRCh38
rs201436376 Chr.16: 31462699-31462699 on Build GRCh38
rs763409959 Chr.16: 31462551-31462551 on Build GRCh38
rs368308507 Chr.16: 31462617-31462617 on Build GRCh38
rs371292365 Chr.16: 31462696-31462696 on Build GRCh38
rs370832156 Chr.16: 31462682-31462682 on Build GRCh38
rs551854881 Chr.16: 31462683-31462683 on Build GRCh38
rs777753363 Chr.16: 31462595-31462595 on Build GRCh38
rs539440145 Chr.16: 31462547-31462547 on Build GRCh38
rs997822943 Chr.16: 31462573-31462573 on Build GRCh38
rs756214079 Chr.16: 31462715-31462715 on Build GRCh38

View Full Product Details