Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 22055074-22055347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMAS
Entrez Gene ID
55907
Gene Name
cytidine monophosphate N-acetylneuraminic acid synthetase
Gene Aliases
CSS
UniGene
Hs.311346
Gene Chromosome Location
Chr.12: 22046174-22065674 on Build GRCh38
Transcripts
NM_018686.5 NR_135117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1628499 c.281A>G p.H94R Chr.12: 22055169-22055169 on Build GRCh38
3954543 c.325C>A p.H109N Chr.12: 22055213-22055213 on Build GRCh38
268906 c.353A>T p.D118V Chr.12: 22055241-22055241 on Build GRCh38
5817603 c.348A>G p.S116S Chr.12: 22055236-22055236 on Build GRCh38
7176972 c.382G>C p.E128Q Chr.12: 22055270-22055270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372910806 Chr.12: 22055249-22055249 on Build GRCh38
rs202065613 Chr.12: 22055136-22055136 on Build GRCh38
rs771403320 Chr.12: 22055280-22055280 on Build GRCh38
rs755254310 Chr.12: 22055232-22055232 on Build GRCh38
rs767530307 Chr.12: 22055178-22055178 on Build GRCh38
rs753560501 Chr.12: 22055217-22055217 on Build GRCh38
rs779106335 Chr.12: 22055244-22055244 on Build GRCh38
rs141990459 Chr.12: 22055183-22055183 on Build GRCh38
rs369640237 Chr.12: 22055248-22055248 on Build GRCh38
rs771222826 Chr.12: 22055109-22055109 on Build GRCh38
rs750302169 Chr.12: 22055197-22055197 on Build GRCh38
rs751170788 Chr.12: 22055140-22055140 on Build GRCh38
rs201015690 Chr.12: 22055242-22055242 on Build GRCh38
rs748270722 Chr.12: 22055245-22055245 on Build GRCh38
rs763779607 Chr.12: 22055131-22055131 on Build GRCh38
rs762700948 Chr.12: 22055126-22055126 on Build GRCh38
rs150694299 Chr.12: 22055199-22055199 on Build GRCh38
rs761224753 Chr.12: 22055161-22055161 on Build GRCh38
rs766189836 Chr.12: 22055216-22055216 on Build GRCh38
rs941123829 Chr.12: 22055267-22055267 on Build GRCh38
rs371108787 Chr.12: 22055268-22055268 on Build GRCh38
rs985540096 Chr.12: 22055203-22055203 on Build GRCh38
rs200718250 Chr.12: 22055310-22055310 on Build GRCh38
rs749797170 Chr.12: 22055278-22055278 on Build GRCh38
rs777097662 Chr.12: 22055119-22055119 on Build GRCh38

View Full Product Details