Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76081314-76081585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC7
Entrez Gene ID
23265
Gene Name
exocyst complex component 7
Gene Aliases
2-5-3p, EX070, EXO70, EXOC1, Exo70p, YJL085W
UniGene
Hs.514496
Gene Chromosome Location
Chr.17: 76080993-76104221 on Build GRCh38
Transcripts
NM_001013839.3 NM_001145297.3 NM_001145298.3 NM_001145299.3 NM_001282313.1 NM_015219.4 XM_006721786.3 XM_006721787.3 XM_006721788.3 XM_006721789.3 XM_011524571.2 XM_017024397.1 NM_180990.3

Gene Symbol
ZACN
Entrez Gene ID
353174
Gene Name
zinc activated ion channel
Gene Aliases
L2, LGICZ, LGICZ1, ZAC, ZAC1
UniGene
Hs.714919
Gene Chromosome Location
Chr.17: 76079182-76082804 on Build GRCh38
Transcripts
NM_001013839.3 NM_001145297.3 NM_001145298.3 NM_001145299.3 NM_001282313.1 NM_015219.4 XM_006721786.3 XM_006721787.3 XM_006721788.3 XM_006721789.3 XM_011524571.2 XM_017024397.1 NM_180990.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3388152 c.689C>T p.A230V Chr.17: 76081564-76081564 on Build GRCh38
4069802 c.687G>T p.T229T Chr.17: 76081562-76081562 on Build GRCh38
6824645 c.706G>A p.A236T Chr.17: 76081581-76081581 on Build GRCh38
5055796 c.705C>T p.I235I Chr.17: 76081580-76081580 on Build GRCh38
984602 c.688G>A p.A230T Chr.17: 76081563-76081563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757815055 Chr.17: 76081532-76081532 on Build GRCh38
rs758791520 Chr.17: 76081549-76081549 on Build GRCh38
rs976104701 Chr.17: 76081505-76081505 on Build GRCh38
rs767826167 Chr.17: 76081419-76081419 on Build GRCh38
rs771704797 Chr.17: 76081511-76081511 on Build GRCh38
rs763976488 Chr.17: 76081413-76081413 on Build GRCh38
rs1027914331 Chr.17: 76081440-76081440 on Build GRCh38
rs972233197 Chr.17: 76081482-76081482 on Build GRCh38
rs542726890 Chr.17: 76081459-76081459 on Build GRCh38
rs747590255 Chr.17: 76081560-76081560 on Build GRCh38
rs747707246 Chr.17: 76081506-76081507 on Build GRCh38
rs531328918 Chr.17: 76081487-76081487 on Build GRCh38
rs755188097 Chr.17: 76081363-76081363 on Build GRCh38
rs200623073 Chr.17: 76081401-76081401 on Build GRCh38
rs377168006 Chr.17: 76081339-76081339 on Build GRCh38
rs1000607854 Chr.17: 76081563-76081563 on Build GRCh38
rs778314165 Chr.17: 76081398-76081398 on Build GRCh38
rs749977473 Chr.17: 76081367-76081367 on Build GRCh38
rs910457903 Chr.17: 76081491-76081491 on Build GRCh38
rs369528499 Chr.17: 76081438-76081438 on Build GRCh38
rs751849469 Chr.17: 76081357-76081357 on Build GRCh38
rs373727958 Chr.17: 76081441-76081441 on Build GRCh38
rs371601457 Chr.17: 76081424-76081424 on Build GRCh38
rs146836718 Chr.17: 76081556-76081556 on Build GRCh38
rs202074550 Chr.17: 76081564-76081564 on Build GRCh38
rs746214535 Chr.17: 76081453-76081453 on Build GRCh38
rs201632789 Chr.17: 76081399-76081399 on Build GRCh38
rs1056381794 Chr.17: 76081411-76081411 on Build GRCh38
rs780959820 Chr.17: 76081438-76081439 on Build GRCh38
rs373637080 Chr.17: 76081514-76081514 on Build GRCh38
rs912131844 Chr.17: 76081461-76081461 on Build GRCh38
rs1055335286 Chr.17: 76081522-76081522 on Build GRCh38
rs143316623 Chr.17: 76081340-76081340 on Build GRCh38
rs201124965 Chr.17: 76081536-76081536 on Build GRCh38
rs780227901 Chr.17: 76081551-76081551 on Build GRCh38
rs754741502 Chr.17: 76081416-76081418 on Build GRCh38
rs894118946 Chr.17: 76081415-76081415 on Build GRCh38
rs930680424 Chr.17: 76081486-76081486 on Build GRCh38
rs201163620 Chr.17: 76081446-76081446 on Build GRCh38
rs8836 Chr.17: 76081416-76081416 on Build GRCh38
rs199930768 Chr.17: 76081412-76081412 on Build GRCh38
rs143922990 Chr.17: 76081512-76081512 on Build GRCh38
rs145200784 Chr.17: 76081402-76081402 on Build GRCh38
rs750823224 Chr.17: 76081541-76081541 on Build GRCh38
rs964729027 Chr.17: 76081492-76081492 on Build GRCh38
rs779504228 Chr.17: 76081562-76081562 on Build GRCh38
rs577823187 Chr.17: 76081425-76081425 on Build GRCh38
rs376126613 Chr.17: 76081465-76081465 on Build GRCh38
rs779646744 Chr.17: 76081507-76081507 on Build GRCh38
rs1049094481 Chr.17: 76081491-76081499 on Build GRCh38
rs140656208 Chr.17: 76081561-76081561 on Build GRCh38
rs562611925 Chr.17: 76081463-76081463 on Build GRCh38
rs372981369 Chr.17: 76081445-76081445 on Build GRCh38
rs373431850 Chr.17: 76081555-76081555 on Build GRCh38
rs746595295 Chr.17: 76081508-76081508 on Build GRCh38
rs1006390876 Chr.17: 76081410-76081410 on Build GRCh38
rs141382007 Chr.17: 76081403-76081403 on Build GRCh38
rs148665598 Chr.17: 76081562-76081562 on Build GRCh38
rs762932459 Chr.17: 76081565-76081565 on Build GRCh38
rs781489327 Chr.17: 76081370-76081370 on Build GRCh38
rs201573073 Chr.17: 76081426-76081426 on Build GRCh38
rs759896218 Chr.17: 76081418-76081418 on Build GRCh38
rs775035609 Chr.17: 76081404-76081404 on Build GRCh38
rs564905386 Chr.17: 76081517-76081517 on Build GRCh38
rs919347518 Chr.17: 76081484-76081484 on Build GRCh38
rs771757847 Chr.17: 76081500-76081500 on Build GRCh38
rs780817546 Chr.17: 76081558-76081558 on Build GRCh38
rs755529553 Chr.17: 76081507-76081507 on Build GRCh38
rs776655580 Chr.17: 76081414-76081414 on Build GRCh38
rs764238220 Chr.17: 76081516-76081516 on Build GRCh38
rs374650693 Chr.17: 76081396-76081396 on Build GRCh38
rs147360559 Chr.17: 76081380-76081380 on Build GRCh38
rs189863330 Chr.17: 76081469-76081469 on Build GRCh38

View Full Product Details