Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123937398-123937671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC92
Entrez Gene ID
80212
Gene Name
coiled-coil domain containing 92
Gene Aliases
-
UniGene
Hs.114111
Gene Chromosome Location
Chr.12: 123936409-123972985 on Build GRCh38
Transcripts
NM_001304957.1 NM_001304958.1 NM_001304959.1 NM_001304960.1 NM_001304961.1 NM_025140.2 XM_005253624.2 XM_017019984.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2226829 c.466G>A p.A156T Chr.12: 123937588-123937588 on Build GRCh38
2226827 c.540G>A p.P180P Chr.12: 123937514-123937514 on Build GRCh38
4670124 c.477C>T p.T159T Chr.12: 123937577-123937577 on Build GRCh38
4670123 c.579C>A p.P193P Chr.12: 123937475-123937475 on Build GRCh38
6520796 c.519C>T p.S173S Chr.12: 123937535-123937535 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150284078 Chr.12: 123937452-123937452 on Build GRCh38
rs758089612 Chr.12: 123937522-123937522 on Build GRCh38
rs908706507 Chr.12: 123937466-123937466 on Build GRCh38
rs770966187 Chr.12: 123937444-123937444 on Build GRCh38
rs763083665 Chr.12: 123937457-123937457 on Build GRCh38
rs770592291 Chr.12: 123937543-123937543 on Build GRCh38
rs1128802 Chr.12: 123937475-123937475 on Build GRCh38
rs144657516 Chr.12: 123937562-123937562 on Build GRCh38
rs777524677 Chr.12: 123937615-123937615 on Build GRCh38
rs761814821 Chr.12: 123937584-123937584 on Build GRCh38
rs780565796 Chr.12: 123937628-123937628 on Build GRCh38
rs747767968 Chr.12: 123937440-123937440 on Build GRCh38
rs758420754 Chr.12: 123937540-123937540 on Build GRCh38
rs771590551 Chr.12: 123937471-123937472 on Build GRCh38
rs141016743 Chr.12: 123937589-123937589 on Build GRCh38
rs369556523 Chr.12: 123937476-123937476 on Build GRCh38
rs781701790 Chr.12: 123937425-123937425 on Build GRCh38
rs775416812 Chr.12: 123937456-123937456 on Build GRCh38
rs138698948 Chr.12: 123937616-123937616 on Build GRCh38
rs774587971 Chr.12: 123937565-123937565 on Build GRCh38
rs778561559 Chr.12: 123937438-123937438 on Build GRCh38
rs761079952 Chr.12: 123937601-123937601 on Build GRCh38
rs149289004 Chr.12: 123937635-123937635 on Build GRCh38
rs781130374 Chr.12: 123937552-123937552 on Build GRCh38
rs140100051 Chr.12: 123937427-123937427 on Build GRCh38
rs140945803 Chr.12: 123937561-123937561 on Build GRCh38
rs756443057 Chr.12: 123937485-123937485 on Build GRCh38
rs746464773 Chr.12: 123937535-123937535 on Build GRCh38
rs767973007 Chr.12: 123937478-123937478 on Build GRCh38
rs759312013 Chr.12: 123937451-123937451 on Build GRCh38
rs775277901 Chr.12: 123937550-123937552 on Build GRCh38
rs200976444 Chr.12: 123937446-123937446 on Build GRCh38
rs267603367 Chr.12: 123937472-123937472 on Build GRCh38
rs756466263 Chr.12: 123937434-123937434 on Build GRCh38
rs137881473 Chr.12: 123937477-123937477 on Build GRCh38
rs757733147 Chr.12: 123937424-123937424 on Build GRCh38
rs745427235 Chr.12: 123937637-123937637 on Build GRCh38
rs375777447 Chr.12: 123937501-123937501 on Build GRCh38
rs200893122 Chr.12: 123937588-123937588 on Build GRCh38
rs373710834 Chr.12: 123937479-123937479 on Build GRCh38
rs764341645 Chr.12: 123937473-123937473 on Build GRCh38
rs528091347 Chr.12: 123937609-123937609 on Build GRCh38
rs898908164 Chr.12: 123937521-123937521 on Build GRCh38
rs757033113 Chr.12: 123937623-123937623 on Build GRCh38
rs768819776 Chr.12: 123937564-123937564 on Build GRCh38
rs745784029 Chr.12: 123937559-123937559 on Build GRCh38
rs372052672 Chr.12: 123937421-123937421 on Build GRCh38
rs746554659 Chr.12: 123937443-123937443 on Build GRCh38
rs766947844 Chr.12: 123937602-123937602 on Build GRCh38
rs777614867 Chr.12: 123937534-123937534 on Build GRCh38
rs772842988 Chr.12: 123937441-123937441 on Build GRCh38
rs745429394 Chr.12: 123937456-123937457 on Build GRCh38
rs370137076 Chr.12: 123937502-123937502 on Build GRCh38

View Full Product Details