Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34385556-34385821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf24
Entrez Gene ID
84688
Gene Name
chromosome 9 open reading frame 24
Gene Aliases
CBE1, NYD-SP22, SMRP1, bA573M23.4
UniGene
Hs.50334
Gene Chromosome Location
Chr.9: 34379019-34400833 on Build GRCh38
Transcripts
NM_032596.3 XM_005251616.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7135992 c.33G>A p.P11P Chr.9: 34385780-34385780 on Build GRCh38
6322999 c.176C>A p.A59D Chr.9: 34385741-34385741 on Build GRCh38
7358783 c.158G>A p.R53Q Chr.9: 34385759-34385759 on Build GRCh38
4925745 c.157C>A p.R53R Chr.9: 34385760-34385760 on Build GRCh38
6616895 c.267T>G p.L89L Chr.9: 34385650-34385650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761338563 Chr.9: 34385683-34385683 on Build GRCh38
rs199593654 Chr.9: 34385653-34385653 on Build GRCh38
rs754349571 Chr.9: 34385698-34385698 on Build GRCh38
rs961833460 Chr.9: 34385650-34385650 on Build GRCh38
rs765744200 Chr.9: 34385795-34385795 on Build GRCh38
rs564045199 Chr.9: 34385619-34385619 on Build GRCh38
rs377596770 Chr.9: 34385621-34385621 on Build GRCh38
rs868752559 Chr.9: 34385640-34385640 on Build GRCh38
rs758588671 Chr.9: 34385726-34385726 on Build GRCh38
rs766977544 Chr.9: 34385692-34385692 on Build GRCh38
rs758200252 Chr.9: 34385593-34385594 on Build GRCh38
rs780503622 Chr.9: 34385596-34385596 on Build GRCh38
rs139282413 Chr.9: 34385702-34385702 on Build GRCh38
rs1037798426 Chr.9: 34385717-34385735 on Build GRCh38
rs751065059 Chr.9: 34385587-34385587 on Build GRCh38
rs748334291 Chr.9: 34385611-34385611 on Build GRCh38
rs1042126326 Chr.9: 34385594-34385594 on Build GRCh38
rs753103140 Chr.9: 34385720-34385720 on Build GRCh38
rs775212662 Chr.9: 34385754-34385754 on Build GRCh38
rs772697066 Chr.9: 34385780-34385780 on Build GRCh38
rs781360978 Chr.9: 34385596-34385596 on Build GRCh38
rs41303241 Chr.9: 34385793-34385793 on Build GRCh38
rs779205366 Chr.9: 34385710-34385710 on Build GRCh38
rs987012442 Chr.9: 34385630-34385630 on Build GRCh38
rs371496210 Chr.9: 34385599-34385599 on Build GRCh38
rs148681444 Chr.9: 34385625-34385625 on Build GRCh38
rs112332239 Chr.9: 34385759-34385759 on Build GRCh38
rs991120392 Chr.9: 34385701-34385701 on Build GRCh38
rs747178254 Chr.9: 34385734-34385734 on Build GRCh38
rs762569717 Chr.9: 34385667-34385667 on Build GRCh38
rs920316836 Chr.9: 34385665-34385665 on Build GRCh38
rs761391464 Chr.9: 34385767-34385767 on Build GRCh38
rs1015883805 Chr.9: 34385637-34385637 on Build GRCh38
rs778034425 Chr.9: 34385729-34385729 on Build GRCh38
rs752693407 Chr.9: 34385588-34385589 on Build GRCh38
rs377488201 Chr.9: 34385675-34385675 on Build GRCh38
rs757443260 Chr.9: 34385745-34385745 on Build GRCh38
rs756676945 Chr.9: 34385589-34385589 on Build GRCh38
rs745882367 Chr.9: 34385633-34385633 on Build GRCh38
rs769714789 Chr.9: 34385654-34385654 on Build GRCh38
rs756404307 Chr.9: 34385780-34385780 on Build GRCh38
rs371128055 Chr.9: 34385747-34385747 on Build GRCh38
rs576056264 Chr.9: 34385677-34385677 on Build GRCh38
rs774026748 Chr.9: 34385766-34385766 on Build GRCh38
rs755369141 Chr.9: 34385601-34385601 on Build GRCh38
rs372723683 Chr.9: 34385662-34385662 on Build GRCh38
rs745811804 Chr.9: 34385749-34385749 on Build GRCh38
rs772327045 Chr.9: 34385613-34385613 on Build GRCh38
rs781352259 Chr.9: 34385746-34385746 on Build GRCh38
rs768238547 Chr.9: 34385760-34385760 on Build GRCh38
rs769753783 Chr.9: 34385752-34385752 on Build GRCh38
rs767030544 Chr.9: 34385771-34385771 on Build GRCh38

View Full Product Details