Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50568840-50569101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1G
Entrez Gene ID
8913
Gene Name
calcium voltage-gated channel subunit alpha1 G
Gene Aliases
Ca(V)T.1, Cav3.1, NBR13, SCA42
UniGene
Hs.591169
Gene Chromosome Location
Chr.17: 50560929-50629625 on Build GRCh38
Transcripts
NM_001256324.1 NM_001256325.1 NM_001256326.1 NM_001256327.1 NM_001256328.1 NM_001256329.1 NM_001256330.1 NM_001256331.1 NM_001256332.1 NM_001256333.1 NM_001256334.1 NM_001256359.1 NM_001256360.1 NM_001256361.1 NM_018896.4 NM_198376.2 NM_198377.2 NM_198378.2 NM_198379.2 NM_198380.2 NM_198382.2 NM_198383.2 NM_198384.2 NM_198385.2 NM_198386.2 NM_198387.2 NM_198388.2 NM_198396.2 NR_046054.1 NR_046055.1 NR_046056.1 NR_046057.1 NR_046058.1 XM_006722160.3 XM_006722161.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6618045 c.304C>T p.R102W Chr.17: 50568931-50568931 on Build GRCh38
287344 c.321C>T p.I107I Chr.17: 50568948-50568948 on Build GRCh38
5369575 c.277C>T p.L93F Chr.17: 50568904-50568904 on Build GRCh38
3519174 c.270C>T p.V90V Chr.17: 50568897-50568897 on Build GRCh38
3192055 c.285C>T p.C95C Chr.17: 50568912-50568912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900319892 Chr.17: 50569060-50569062 on Build GRCh38
rs1042419466 Chr.17: 50568986-50568989 on Build GRCh38
rs139870591 Chr.17: 50568985-50568985 on Build GRCh38
rs751572113 Chr.17: 50568990-50568991 on Build GRCh38
rs762356545 Chr.17: 50569026-50569027 on Build GRCh38
rs770271368 Chr.17: 50569023-50569023 on Build GRCh38
rs371342918 Chr.17: 50569037-50569037 on Build GRCh38
rs3833150 Chr.17: 50568985-50568986 on Build GRCh38
rs763813233 Chr.17: 50568932-50568932 on Build GRCh38
rs529908416 Chr.17: 50568948-50568948 on Build GRCh38
rs756048612 Chr.17: 50569006-50569006 on Build GRCh38
rs775178189 Chr.17: 50569021-50569022 on Build GRCh38
rs755276697 Chr.17: 50568969-50568969 on Build GRCh38
rs1039159897 Chr.17: 50569039-50569044 on Build GRCh38
rs401388 Chr.17: 50569016-50569016 on Build GRCh38
rs755064335 Chr.17: 50568995-50568996 on Build GRCh38
rs79838522 Chr.17: 50569015-50569015 on Build GRCh38
rs767153145 Chr.17: 50569011-50569015 on Build GRCh38
rs771649000 Chr.17: 50568881-50568881 on Build GRCh38
rs771736617 Chr.17: 50569017-50569018 on Build GRCh38
rs918200860 Chr.17: 50568970-50568970 on Build GRCh38
rs939434619 Chr.17: 50569082-50569082 on Build GRCh38
rs372490164 Chr.17: 50569011-50569011 on Build GRCh38
rs997794272 Chr.17: 50569063-50569063 on Build GRCh38
rs777605910 Chr.17: 50569009-50569015 on Build GRCh38
rs371434876 Chr.17: 50569015-50569015 on Build GRCh38
rs780071833 Chr.17: 50568960-50568960 on Build GRCh38
rs748117263 Chr.17: 50568999-50569000 on Build GRCh38
rs760078615 Chr.17: 50569029-50569029 on Build GRCh38
rs750305086 Chr.17: 50568949-50568949 on Build GRCh38
rs749099745 Chr.17: 50569012-50569013 on Build GRCh38
rs781240948 Chr.17: 50568971-50568971 on Build GRCh38
rs749745167 Chr.17: 50569024-50569024 on Build GRCh38
rs945318433 Chr.17: 50568986-50568987 on Build GRCh38
rs537989352 Chr.17: 50569044-50569044 on Build GRCh38
rs201146969 Chr.17: 50568913-50568913 on Build GRCh38
rs774155893 Chr.17: 50569013-50569014 on Build GRCh38
rs771738729 Chr.17: 50569025-50569025 on Build GRCh38
rs576295333 Chr.17: 50568957-50568957 on Build GRCh38
rs377732195 Chr.17: 50569013-50569015 on Build GRCh38
rs781059473 Chr.17: 50568998-50568999 on Build GRCh38
rs751757027 Chr.17: 50568962-50568962 on Build GRCh38
rs752427792 Chr.17: 50569011-50569013 on Build GRCh38
rs58450142 Chr.17: 50569015-50569016 on Build GRCh38
rs910163279 Chr.17: 50569028-50569028 on Build GRCh38
rs78259124 Chr.17: 50569014-50569014 on Build GRCh38
rs775015926 Chr.17: 50569027-50569027 on Build GRCh38
rs569312786 Chr.17: 50569036-50569036 on Build GRCh38
rs770616342 Chr.17: 50569013-50569013 on Build GRCh38
rs770465443 Chr.17: 50568867-50568867 on Build GRCh38
rs963107685 Chr.17: 50568931-50568931 on Build GRCh38
rs371274681 Chr.17: 50568939-50568939 on Build GRCh38
rs1025329505 Chr.17: 50568989-50568989 on Build GRCh38
rs1006719185 Chr.17: 50568940-50568940 on Build GRCh38
rs370361162 Chr.17: 50568912-50568912 on Build GRCh38
rs765720302 Chr.17: 50569027-50569028 on Build GRCh38
rs925328853 Chr.17: 50569013-50569013 on Build GRCh38
rs773853745 Chr.17: 50568870-50568870 on Build GRCh38

View Full Product Details