Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101731759-101732028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP97
Entrez Gene ID
79598
Gene Name
centrosomal protein 97
Gene Aliases
2810403B08Rik, LRRIQ2
UniGene
Hs.444135
Gene Chromosome Location
Chr.3: 101724098-101770562 on Build GRCh38
Transcripts
NM_001303401.1 NM_024548.3 XM_006713743.3 XM_011513127.1 XM_017007178.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1616668 c.547C>T p.R183* Chr.3: 101731939-101731939 on Build GRCh38
1616669 c.547C>T p.R183* Chr.3: 101731939-101731939 on Build GRCh38
7172293 c.547C>A p.R183R Chr.3: 101731939-101731939 on Build GRCh38
1130391 c.510A>T p.R170S Chr.3: 101731902-101731902 on Build GRCh38
1130390 c.510A>T p.R170S Chr.3: 101731902-101731902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768584483 Chr.3: 101731823-101731823 on Build GRCh38
rs763363498 Chr.3: 101731989-101731989 on Build GRCh38
rs774720492 Chr.3: 101731998-101731998 on Build GRCh38
rs753847232 Chr.3: 101731915-101731915 on Build GRCh38
rs953304840 Chr.3: 101731820-101731820 on Build GRCh38
rs763126349 Chr.3: 101731889-101731889 on Build GRCh38
rs776072662 Chr.3: 101731966-101731966 on Build GRCh38
rs746594612 Chr.3: 101731951-101731951 on Build GRCh38
rs199611157 Chr.3: 101731837-101731837 on Build GRCh38
rs770307504 Chr.3: 101731980-101731980 on Build GRCh38
rs764356930 Chr.3: 101731892-101731892 on Build GRCh38
rs201862164 Chr.3: 101731943-101731943 on Build GRCh38
rs772644232 Chr.3: 101731844-101731844 on Build GRCh38
rs9864426 Chr.3: 101732008-101732008 on Build GRCh38
rs139940133 Chr.3: 101731926-101731926 on Build GRCh38
rs767646219 Chr.3: 101731912-101731912 on Build GRCh38
rs780127687 Chr.3: 101731797-101731797 on Build GRCh38
rs984029829 Chr.3: 101731807-101731807 on Build GRCh38
rs992885938 Chr.3: 101731894-101731894 on Build GRCh38
rs759983065 Chr.3: 101731879-101731879 on Build GRCh38
rs770443689 Chr.3: 101731961-101731961 on Build GRCh38
rs746410340 Chr.3: 101731974-101731974 on Build GRCh38
rs952614010 Chr.3: 101731802-101731802 on Build GRCh38
rs753068491 Chr.3: 101731888-101731888 on Build GRCh38
rs762075996 Chr.3: 101731999-101731999 on Build GRCh38
rs796995655 Chr.3: 101731786-101731786 on Build GRCh38
rs767734270 Chr.3: 101732003-101732003 on Build GRCh38
rs747949237 Chr.3: 101731934-101731934 on Build GRCh38
rs182599710 Chr.3: 101731919-101731919 on Build GRCh38
rs948864139 Chr.3: 101731917-101731917 on Build GRCh38
rs914451601 Chr.3: 101731924-101731924 on Build GRCh38
rs757410297 Chr.3: 101731910-101731910 on Build GRCh38
rs765614535 Chr.3: 101731884-101731884 on Build GRCh38
rs908352603 Chr.3: 101731824-101731824 on Build GRCh38
rs751650337 Chr.3: 101731939-101731939 on Build GRCh38
rs751650810 Chr.3: 101731905-101731905 on Build GRCh38
rs756170944 Chr.3: 101731792-101731792 on Build GRCh38
rs778721030 Chr.3: 101731836-101731836 on Build GRCh38
rs777475294 Chr.3: 101731940-101731940 on Build GRCh38
rs760163007 Chr.3: 101731863-101731863 on Build GRCh38
rs187342025 Chr.3: 101731995-101731995 on Build GRCh38
rs771491755 Chr.3: 101731871-101731871 on Build GRCh38
rs201836749 Chr.3: 101731873-101731873 on Build GRCh38
rs771723851 Chr.3: 101731841-101731841 on Build GRCh38
rs749163775 Chr.3: 101731809-101731809 on Build GRCh38
rs961520201 Chr.3: 101731853-101731853 on Build GRCh38
rs1027987811 Chr.3: 101731800-101731800 on Build GRCh38

View Full Product Details