Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 441981-442212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA11P
Entrez Gene ID
79963
Gene Name
ATP binding cassette subfamily A member 11, pseudogene
Gene Aliases
ABCA11, EST1133530
UniGene
Hs.677493
Gene Chromosome Location
Chr.4: 425435-474209 on Build GRCh38
Transcripts
NR_002451.2 NM_133474.3

Gene Symbol
ZNF721
Entrez Gene ID
170960
Gene Name
zinc finger protein 721
Gene Aliases
-
UniGene
Hs.428360
Gene Chromosome Location
Chr.4: 439984-499171 on Build GRCh38
Transcripts
NR_002451.2 NM_133474.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6832694 c.1661A>C p.K554T Chr.4: 442116-442116 on Build GRCh38
5017092 c.1573T>C p.C525R Chr.4: 442204-442204 on Build GRCh38
278402 c.2318A>C p.K773T Chr.4: 442149-442149 on Build GRCh38
3604031 c.2358T>G p.C786W Chr.4: 442109-442109 on Build GRCh38
3783805 c.1625A>T p.E542V Chr.4: 442152-442152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782416079 Chr.4: 442077-442077 on Build GRCh38
rs782592476 Chr.4: 442085-442085 on Build GRCh38
rs782715544 Chr.4: 442140-442140 on Build GRCh38
rs13120917 Chr.4: 442055-442055 on Build GRCh38
rs186714659 Chr.4: 442016-442016 on Build GRCh38
rs782289856 Chr.4: 442017-442017 on Build GRCh38
rs374783735 Chr.4: 442134-442134 on Build GRCh38
rs782701975 Chr.4: 442024-442024 on Build GRCh38
rs368506813 Chr.4: 442047-442047 on Build GRCh38
rs782151801 Chr.4: 442120-442120 on Build GRCh38
rs190754079 Chr.4: 442063-442063 on Build GRCh38
rs782355386 Chr.4: 442178-442178 on Build GRCh38
rs61793703 Chr.4: 442106-442106 on Build GRCh38
rs782684732 Chr.4: 442171-442171 on Build GRCh38
rs998857640 Chr.4: 442108-442108 on Build GRCh38
rs371250652 Chr.4: 442094-442094 on Build GRCh38
rs781876306 Chr.4: 442053-442053 on Build GRCh38
rs782165509 Chr.4: 442044-442044 on Build GRCh38
rs372133028 Chr.4: 442156-442156 on Build GRCh38
rs180899199 Chr.4: 442071-442071 on Build GRCh38
rs1040448771 Chr.4: 442103-442103 on Build GRCh38
rs564127195 Chr.4: 442118-442118 on Build GRCh38
rs782648764 Chr.4: 442095-442095 on Build GRCh38
rs781866787 Chr.4: 442162-442162 on Build GRCh38
rs782130229 Chr.4: 442110-442110 on Build GRCh38
rs782568321 Chr.4: 442117-442118 on Build GRCh38
rs542073111 Chr.4: 442056-442056 on Build GRCh38
rs201461396 Chr.4: 442114-442114 on Build GRCh38
rs1048827808 Chr.4: 442037-442037 on Build GRCh38
rs782716841 Chr.4: 442157-442157 on Build GRCh38
rs13114490 Chr.4: 442052-442052 on Build GRCh38
rs781789961 Chr.4: 442152-442152 on Build GRCh38
rs782236652 Chr.4: 442076-442076 on Build GRCh38
rs781885128 Chr.4: 442035-442035 on Build GRCh38
rs267600163 Chr.4: 442144-442144 on Build GRCh38
rs782191131 Chr.4: 442088-442088 on Build GRCh38
rs782662525 Chr.4: 442147-442147 on Build GRCh38
rs376721590 Chr.4: 442065-442065 on Build GRCh38
rs531627742 Chr.4: 442170-442170 on Build GRCh38
rs554405280 Chr.4: 442061-442061 on Build GRCh38

View Full Product Details