Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 124242163-124242436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP5
Entrez Gene ID
129684
Gene Name
contactin associated protein like 5
Gene Aliases
caspr5
UniGene
Hs.660653
Gene Chromosome Location
Chr.2: 124025287-124921201 on Build GRCh38
Transcripts
NM_130773.3 XM_006712258.1 XM_017003316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1613360 c.275C>T p.A92V Chr.2: 124242287-124242287 on Build GRCh38
1613361 c.287G>T p.R96I Chr.2: 124242299-124242299 on Build GRCh38
5034521 c.380G>T p.W127L Chr.2: 124242392-124242392 on Build GRCh38
319559 c.248G>C p.G83A Chr.2: 124242260-124242260 on Build GRCh38
5994275 c.251A>T p.N84I Chr.2: 124242263-124242263 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776009823 Chr.2: 124242196-124242196 on Build GRCh38
rs756004265 Chr.2: 124242218-124242220 on Build GRCh38
rs780303323 Chr.2: 124242406-124242406 on Build GRCh38
rs867651102 Chr.2: 124242363-124242363 on Build GRCh38
rs775431778 Chr.2: 124242323-124242323 on Build GRCh38
rs200562881 Chr.2: 124242357-124242357 on Build GRCh38
rs953863371 Chr.2: 124242270-124242270 on Build GRCh38
rs369199254 Chr.2: 124242221-124242221 on Build GRCh38
rs749040103 Chr.2: 124242264-124242264 on Build GRCh38
rs776949563 Chr.2: 124242208-124242208 on Build GRCh38
rs371368855 Chr.2: 124242372-124242372 on Build GRCh38
rs756084592 Chr.2: 124242240-124242240 on Build GRCh38
rs778561420 Chr.2: 124242382-124242382 on Build GRCh38
rs757022526 Chr.2: 124242379-124242379 on Build GRCh38
rs752229409 Chr.2: 124242369-124242369 on Build GRCh38
rs566067392 Chr.2: 124242339-124242339 on Build GRCh38
rs750303852 Chr.2: 124242387-124242387 on Build GRCh38
rs1016002492 Chr.2: 124242326-124242326 on Build GRCh38
rs769130518 Chr.2: 124242330-124242330 on Build GRCh38
rs373476119 Chr.2: 124242228-124242228 on Build GRCh38
rs751174282 Chr.2: 124242220-124242220 on Build GRCh38
rs886110078 Chr.2: 124242253-124242253 on Build GRCh38
rs773366678 Chr.2: 124242356-124242356 on Build GRCh38
rs960964264 Chr.2: 124242403-124242403 on Build GRCh38
rs747133475 Chr.2: 124242324-124242324 on Build GRCh38
rs1029405540 Chr.2: 124242216-124242216 on Build GRCh38
rs374804172 Chr.2: 124242378-124242378 on Build GRCh38
rs752321298 Chr.2: 124242230-124242230 on Build GRCh38
rs766008579 Chr.2: 124242215-124242215 on Build GRCh38
rs753828713 Chr.2: 124242374-124242374 on Build GRCh38
rs374807595 Chr.2: 124242190-124242190 on Build GRCh38
rs772052519 Chr.2: 124242193-124242193 on Build GRCh38
rs369801109 Chr.2: 124242303-124242303 on Build GRCh38
rs200770909 Chr.2: 124242212-124242212 on Build GRCh38
rs377746974 Chr.2: 124242364-124242364 on Build GRCh38
rs377465900 Chr.2: 124242392-124242392 on Build GRCh38
rs777902823 Chr.2: 124242248-124242248 on Build GRCh38
rs146843368 Chr.2: 124242207-124242207 on Build GRCh38
rs760280589 Chr.2: 124242370-124242370 on Build GRCh38
rs777147651 Chr.2: 124242346-124242346 on Build GRCh38
rs376635773 Chr.2: 124242276-124242276 on Build GRCh38
rs1034276241 Chr.2: 124242367-124242367 on Build GRCh38
rs373323066 Chr.2: 124242304-124242304 on Build GRCh38
rs779127795 Chr.2: 124242291-124242291 on Build GRCh38
rs762127739 Chr.2: 124242355-124242355 on Build GRCh38

View Full Product Details