Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 24692330-24692603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP12A
Entrez Gene ID
479
Gene Name
ATPase H+/K+ transporting non-gastric alpha2 subunit
Gene Aliases
#945, ATP1AL1, HK
UniGene
Hs.147111
Gene Chromosome Location
Chr.13: 24680411-24711785 on Build GRCh38
Transcripts
NM_001185085.1 NM_001676.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6372128 c.1112G>T p.R371L Chr.13: 24692454-24692454 on Build GRCh38
7412272 c.1200G>T p.R400S Chr.13: 24692560-24692560 on Build GRCh38
7210830 c.1098G>A p.S366S Chr.13: 24692440-24692440 on Build GRCh38
6001027 c.1164C>T p.L388L Chr.13: 24692506-24692506 on Build GRCh38
6406773 c.1133T>C p.L378P Chr.13: 24692475-24692475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777140060 Chr.13: 24692541-24692542 on Build GRCh38
rs191179077 Chr.13: 24692409-24692409 on Build GRCh38
rs755893211 Chr.13: 24692459-24692459 on Build GRCh38
rs555243224 Chr.13: 24692538-24692538 on Build GRCh38
rs200648050 Chr.13: 24692534-24692534 on Build GRCh38
rs1033107295 Chr.13: 24692507-24692507 on Build GRCh38
rs760695882 Chr.13: 24692563-24692563 on Build GRCh38
rs761069302 Chr.13: 24692435-24692435 on Build GRCh38
rs568295692 Chr.13: 24692400-24692400 on Build GRCh38
rs766221397 Chr.13: 24692564-24692564 on Build GRCh38
rs778320466 Chr.13: 24692506-24692506 on Build GRCh38
rs940712729 Chr.13: 24692365-24692365 on Build GRCh38
rs142661524 Chr.13: 24692490-24692490 on Build GRCh38
rs565720035 Chr.13: 24692363-24692363 on Build GRCh38
rs748547110 Chr.13: 24692411-24692411 on Build GRCh38
rs534559668 Chr.13: 24692374-24692374 on Build GRCh38
rs750215883 Chr.13: 24692394-24692394 on Build GRCh38
rs769987029 Chr.13: 24692544-24692544 on Build GRCh38
rs536291211 Chr.13: 24692393-24692393 on Build GRCh38
rs755907407 Chr.13: 24692399-24692399 on Build GRCh38
rs202155951 Chr.13: 24692439-24692439 on Build GRCh38
rs1050545648 Chr.13: 24692392-24692392 on Build GRCh38
rs575878580 Chr.13: 24692539-24692539 on Build GRCh38
rs982274398 Chr.13: 24692546-24692546 on Build GRCh38
rs543553750 Chr.13: 24692446-24692446 on Build GRCh38
rs748974707 Chr.13: 24692407-24692407 on Build GRCh38
rs759783098 Chr.13: 24692440-24692440 on Build GRCh38
rs899130264 Chr.13: 24692386-24692386 on Build GRCh38
rs772620660 Chr.13: 24692433-24692433 on Build GRCh38
rs146073882 Chr.13: 24692495-24692495 on Build GRCh38
rs775638009 Chr.13: 24692545-24692545 on Build GRCh38
rs556805114 Chr.13: 24692419-24692419 on Build GRCh38
rs778766710 Chr.13: 24692489-24692489 on Build GRCh38
rs747492140 Chr.13: 24692510-24692510 on Build GRCh38
rs557563746 Chr.13: 24692529-24692532 on Build GRCh38
rs112162533 Chr.13: 24692541-24692541 on Build GRCh38
rs753674751 Chr.13: 24692572-24692572 on Build GRCh38
rs764107424 Chr.13: 24692557-24692557 on Build GRCh38
rs765321632 Chr.13: 24692378-24692378 on Build GRCh38
rs752570110 Chr.13: 24692529-24692530 on Build GRCh38
rs751336859 Chr.13: 24692446-24692446 on Build GRCh38
rs375256989 Chr.13: 24692475-24692475 on Build GRCh38
rs201938406 Chr.13: 24692390-24692390 on Build GRCh38
rs930735107 Chr.13: 24692388-24692388 on Build GRCh38
rs757885776 Chr.13: 24692491-24692491 on Build GRCh38
rs746028094 Chr.13: 24692533-24692533 on Build GRCh38
rs768243716 Chr.13: 24692408-24692408 on Build GRCh38
rs771242833 Chr.13: 24692529-24692529 on Build GRCh38
rs769933522 Chr.13: 24692454-24692454 on Build GRCh38
rs562286871 Chr.13: 24692538-24692538 on Build GRCh38
rs754888521 Chr.13: 24692578-24692578 on Build GRCh38
rs867224558 Chr.13: 24692573-24692573 on Build GRCh38
rs372274390 Chr.13: 24692547-24692547 on Build GRCh38
rs774093370 Chr.13: 24692562-24692562 on Build GRCh38
rs754809955 Chr.13: 24692469-24692470 on Build GRCh38
rs1036775613 Chr.13: 24692368-24692368 on Build GRCh38
rs367998322 Chr.13: 24692530-24692530 on Build GRCh38
rs750380289 Chr.13: 24692455-24692455 on Build GRCh38
rs576837427 Chr.13: 24692434-24692434 on Build GRCh38
rs553240041 Chr.13: 24692461-24692461 on Build GRCh38
rs752833147 Chr.13: 24692453-24692453 on Build GRCh38
rs771332778 Chr.13: 24692436-24692436 on Build GRCh38
rs867536379 Chr.13: 24692531-24692531 on Build GRCh38
rs754694161 Chr.13: 24692476-24692476 on Build GRCh38

View Full Product Details