Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17942679-17942908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC124
Entrez Gene ID
115098
Gene Name
coiled-coil domain containing 124
Gene Aliases
-
UniGene
Hs.100043
Gene Chromosome Location
Chr.19: 17933015-17943985 on Build GRCh38
Transcripts
NM_001136203.1 NM_138442.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6295973 c.263G>T p.R88L Chr.19: 17942759-17942759 on Build GRCh38
6043250 c.289C>T p.R97W Chr.19: 17942785-17942785 on Build GRCh38
307393 c.271A>T p.T91S Chr.19: 17942767-17942767 on Build GRCh38
1391594 c.187C>T p.Q63* Chr.19: 17942683-17942683 on Build GRCh38
5719023 c.342G>A p.P114P Chr.19: 17942838-17942838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs931486784 Chr.19: 17942851-17942851 on Build GRCh38
rs779314366 Chr.19: 17942752-17942752 on Build GRCh38
rs371128459 Chr.19: 17942745-17942745 on Build GRCh38
rs986461196 Chr.19: 17942769-17942769 on Build GRCh38
rs200503204 Chr.19: 17942822-17942822 on Build GRCh38
rs919656617 Chr.19: 17942832-17942832 on Build GRCh38
rs953953979 Chr.19: 17942763-17942763 on Build GRCh38
rs1035685707 Chr.19: 17942706-17942706 on Build GRCh38
rs192049836 Chr.19: 17942857-17942857 on Build GRCh38
rs749329035 Chr.19: 17942746-17942746 on Build GRCh38
rs1021679254 Chr.19: 17942733-17942733 on Build GRCh38
rs570543487 Chr.19: 17942885-17942885 on Build GRCh38
rs143035747 Chr.19: 17942864-17942864 on Build GRCh38
rs1044565164 Chr.19: 17942852-17942852 on Build GRCh38
rs560139923 Chr.19: 17942813-17942813 on Build GRCh38
rs143989433 Chr.19: 17942794-17942794 on Build GRCh38
rs973598090 Chr.19: 17942849-17942849 on Build GRCh38
rs774887396 Chr.19: 17942818-17942818 on Build GRCh38
rs201985798 Chr.19: 17942715-17942715 on Build GRCh38
rs376271505 Chr.19: 17942762-17942762 on Build GRCh38
rs4808106 Chr.19: 17942748-17942748 on Build GRCh38
rs541765064 Chr.19: 17942826-17942826 on Build GRCh38
rs770906965 Chr.19: 17942789-17942789 on Build GRCh38
rs770700367 Chr.19: 17942717-17942718 on Build GRCh38
rs575804409 Chr.19: 17942739-17942739 on Build GRCh38
rs2302173 Chr.19: 17942713-17942713 on Build GRCh38
rs187506397 Chr.19: 17942836-17942836 on Build GRCh38
rs750953589 Chr.19: 17942721-17942721 on Build GRCh38
rs556085759 Chr.19: 17942725-17942725 on Build GRCh38
rs756588390 Chr.19: 17942727-17942727 on Build GRCh38
rs540312451 Chr.19: 17942796-17942796 on Build GRCh38
rs1057346419 Chr.19: 17942882-17942882 on Build GRCh38

View Full Product Details