Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67438559-67438830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO1B
Entrez Gene ID
57175
Gene Name
coronin 1B
Gene Aliases
CORONIN-2
UniGene
Hs.6191
Gene Chromosome Location
Chr.11: 67438047-67443821 on Build GRCh38
Transcripts
NM_001018070.2 NM_020441.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
379844 c.1188C>G p.Y396* Chr.11: 67438827-67438827 on Build GRCh38
2041967 c.1204C>T p.R402W Chr.11: 67438811-67438811 on Build GRCh38
4524284 c.1263G>A p.P421P Chr.11: 67438752-67438752 on Build GRCh38
6442360 c.1271C>A p.S424Y Chr.11: 67438744-67438744 on Build GRCh38
6436442 c.1333G>A p.A445T Chr.11: 67438682-67438682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886873269 Chr.11: 67438739-67438739 on Build GRCh38
rs749943867 Chr.11: 67438661-67438661 on Build GRCh38
rs371788569 Chr.11: 67438674-67438674 on Build GRCh38
rs752068899 Chr.11: 67438642-67438642 on Build GRCh38
rs560903102 Chr.11: 67438752-67438752 on Build GRCh38
rs932691284 Chr.11: 67438595-67438595 on Build GRCh38
rs939532839 Chr.11: 67438789-67438789 on Build GRCh38
rs142509476 Chr.11: 67438753-67438753 on Build GRCh38
rs760208883 Chr.11: 67438758-67438758 on Build GRCh38
rs1015926857 Chr.11: 67438737-67438737 on Build GRCh38
rs141781443 Chr.11: 67438727-67438728 on Build GRCh38
rs367989759 Chr.11: 67438785-67438785 on Build GRCh38
rs749475319 Chr.11: 67438790-67438790 on Build GRCh38
rs746960019 Chr.11: 67438782-67438782 on Build GRCh38
rs906698215 Chr.11: 67438786-67438786 on Build GRCh38
rs766998871 Chr.11: 67438660-67438660 on Build GRCh38
rs761822297 Chr.11: 67438790-67438813 on Build GRCh38
rs1028986097 Chr.11: 67438594-67438594 on Build GRCh38
rs762282775 Chr.11: 67438749-67438749 on Build GRCh38
rs769021054 Chr.11: 67438734-67438734 on Build GRCh38
rs893887926 Chr.11: 67438614-67438614 on Build GRCh38
rs979948699 Chr.11: 67438643-67438643 on Build GRCh38
rs867313056 Chr.11: 67438714-67438714 on Build GRCh38
rs772581541 Chr.11: 67438796-67438796 on Build GRCh38
rs200675963 Chr.11: 67438792-67438792 on Build GRCh38
rs745942973 Chr.11: 67438715-67438715 on Build GRCh38
rs764390354 Chr.11: 67438641-67438641 on Build GRCh38
rs34026289 Chr.11: 67438776-67438776 on Build GRCh38
rs921244054 Chr.11: 67438593-67438593 on Build GRCh38
rs761297209 Chr.11: 67438810-67438810 on Build GRCh38
rs548209940 Chr.11: 67438806-67438806 on Build GRCh38
rs148492786 Chr.11: 67438692-67438692 on Build GRCh38
rs906729128 Chr.11: 67438664-67438664 on Build GRCh38
rs536517452 Chr.11: 67438793-67438793 on Build GRCh38
rs770062333 Chr.11: 67438703-67438705 on Build GRCh38
rs765939658 Chr.11: 67438811-67438811 on Build GRCh38
rs778606005 Chr.11: 67438691-67438691 on Build GRCh38
rs199730254 Chr.11: 67438768-67438768 on Build GRCh38
rs1009717076 Chr.11: 67438629-67438629 on Build GRCh38
rs574741792 Chr.11: 67438727-67438727 on Build GRCh38
rs931057641 Chr.11: 67438699-67438699 on Build GRCh38
rs777672699 Chr.11: 67438781-67438781 on Build GRCh38
rs923785030 Chr.11: 67438644-67438644 on Build GRCh38
rs754737752 Chr.11: 67438684-67438684 on Build GRCh38
rs762880383 Chr.11: 67438779-67438779 on Build GRCh38
rs781604322 Chr.11: 67438629-67438633 on Build GRCh38
rs754610168 Chr.11: 67438769-67438769 on Build GRCh38
rs1053843310 Chr.11: 67438605-67438605 on Build GRCh38
rs562837525 Chr.11: 67438705-67438705 on Build GRCh38
rs780084285 Chr.11: 67438706-67438708 on Build GRCh38
rs968158939 Chr.11: 67438624-67438624 on Build GRCh38
rs542125021 Chr.11: 67438747-67438747 on Build GRCh38
rs943968409 Chr.11: 67438578-67438578 on Build GRCh38
rs577352348 Chr.11: 67438678-67438678 on Build GRCh38
rs762234711 Chr.11: 67438651-67438651 on Build GRCh38
rs749405134 Chr.11: 67438764-67438764 on Build GRCh38
rs11601245 Chr.11: 67438763-67438763 on Build GRCh38
rs779827114 Chr.11: 67438673-67438673 on Build GRCh38
rs935131202 Chr.11: 67438653-67438653 on Build GRCh38
rs755721620 Chr.11: 67438663-67438663 on Build GRCh38
rs763283760 Chr.11: 67438625-67438625 on Build GRCh38
rs749513920 Chr.11: 67438733-67438733 on Build GRCh38
rs1040968659 Chr.11: 67438588-67438588 on Build GRCh38
rs768817457 Chr.11: 67438766-67438789 on Build GRCh38
rs756117196 Chr.11: 67438784-67438784 on Build GRCh38
rs550750255 Chr.11: 67438773-67438773 on Build GRCh38
rs377019509 Chr.11: 67438764-67438764 on Build GRCh38
rs746139828 Chr.11: 67438635-67438643 on Build GRCh38
rs771882753 Chr.11: 67438698-67438698 on Build GRCh38
rs775629291 Chr.11: 67438728-67438728 on Build GRCh38
rs777727726 Chr.11: 67438713-67438713 on Build GRCh38

View Full Product Details