Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 103091116-103091389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDH2
Entrez Gene ID
56898
Gene Name
3-hydroxybutyrate dehydrogenase, type 2
Gene Aliases
DHRS6, EFA6R, PRO20933, SDR15C1, UCPA-OR, UNQ6308
UniGene
Hs.124696
Gene Chromosome Location
Chr.4: 103077618-103099867 on Build GRCh38
Transcripts
NM_020139.3 XM_005263140.3 XM_006714274.3 XM_011532127.2 XM_011532128.2 XM_017008461.1 XM_017008462.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6399728 c.294G>T p.W98C Chr.4: 103091240-103091240 on Build GRCh38
1426167 c.300C>T p.F100F Chr.4: 103091234-103091234 on Build GRCh38
6038753 c.319C>T p.R107C Chr.4: 103091215-103091215 on Build GRCh38
6406024 c.290A>G p.D97G Chr.4: 103091244-103091244 on Build GRCh38
3598755 c.271C>T p.L91L Chr.4: 103091263-103091263 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1033211942 Chr.4: 103091288-103091288 on Build GRCh38
rs941411861 Chr.4: 103091143-103091143 on Build GRCh38
rs140261412 Chr.4: 103091208-103091208 on Build GRCh38
rs761682620 Chr.4: 103091152-103091153 on Build GRCh38
rs750218021 Chr.4: 103091293-103091293 on Build GRCh38
rs878877824 Chr.4: 103091248-103091248 on Build GRCh38
rs765187678 Chr.4: 103091169-103091169 on Build GRCh38
rs776431891 Chr.4: 103091153-103091155 on Build GRCh38
rs764213224 Chr.4: 103091252-103091252 on Build GRCh38
rs563526577 Chr.4: 103091229-103091229 on Build GRCh38
rs755830134 Chr.4: 103091308-103091308 on Build GRCh38
rs865999026 Chr.4: 103091204-103091204 on Build GRCh38
rs989099424 Chr.4: 103091336-103091336 on Build GRCh38
rs143985505 Chr.4: 103091214-103091214 on Build GRCh38
rs140021532 Chr.4: 103091181-103091181 on Build GRCh38
rs549075878 Chr.4: 103091302-103091302 on Build GRCh38
rs751474542 Chr.4: 103091255-103091255 on Build GRCh38
rs768580395 Chr.4: 103091140-103091142 on Build GRCh38
rs779685455 Chr.4: 103091316-103091316 on Build GRCh38
rs956394431 Chr.4: 103091317-103091317 on Build GRCh38
rs753429424 Chr.4: 103091328-103091328 on Build GRCh38
rs151335490 Chr.4: 103091231-103091231 on Build GRCh38
rs748791893 Chr.4: 103091209-103091209 on Build GRCh38
rs780993568 Chr.4: 103091138-103091138 on Build GRCh38
rs745538959 Chr.4: 103091151-103091151 on Build GRCh38
rs775783782 Chr.4: 103091246-103091246 on Build GRCh38
rs763133896 Chr.4: 103091251-103091251 on Build GRCh38
rs767257056 Chr.4: 103091281-103091281 on Build GRCh38
rs777020376 Chr.4: 103091227-103091227 on Build GRCh38
rs148470137 Chr.4: 103091220-103091220 on Build GRCh38
rs192392266 Chr.4: 103091215-103091215 on Build GRCh38
rs761943079 Chr.4: 103091141-103091141 on Build GRCh38
rs187331164 Chr.4: 103091168-103091168 on Build GRCh38
rs964513187 Chr.4: 103091245-103091245 on Build GRCh38
rs1006949133 Chr.4: 103091274-103091274 on Build GRCh38
rs761772154 Chr.4: 103091276-103091276 on Build GRCh38
rs755740381 Chr.4: 103091179-103091179 on Build GRCh38
rs754502753 Chr.4: 103091334-103091334 on Build GRCh38
rs1037558937 Chr.4: 103091150-103091150 on Build GRCh38
rs368646051 Chr.4: 103091213-103091213 on Build GRCh38

View Full Product Details