Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103560219-103560490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG5
Entrez Gene ID
9529
Gene Name
BCL2 associated athanogene 5
Gene Aliases
BAG-5
UniGene
Hs.5443
Gene Chromosome Location
Chr.14: 103556544-103562814 on Build GRCh38
Transcripts
NM_001015048.2 NM_001015049.2 NM_004873.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
953645 c.858G>C p.M286I Chr.14: 103560307-103560307 on Build GRCh38
953646 c.754T>G p.L252V Chr.14: 103560411-103560411 on Build GRCh38
4633874 c.1029G>A p.L343L Chr.14: 103560259-103560259 on Build GRCh38
256385 c.862G>T p.E288* Chr.14: 103560303-103560303 on Build GRCh38
7413951 c.704G>T p.R235L Chr.14: 103560461-103560461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753330598 Chr.14: 103560271-103560271 on Build GRCh38
rs1007408769 Chr.14: 103560372-103560372 on Build GRCh38
rs761089992 Chr.14: 103560353-103560353 on Build GRCh38
rs766951129 Chr.14: 103560374-103560374 on Build GRCh38
rs759091597 Chr.14: 103560403-103560403 on Build GRCh38
rs368650975 Chr.14: 103560343-103560343 on Build GRCh38
rs758276272 Chr.14: 103560419-103560419 on Build GRCh38
rs758093982 Chr.14: 103560301-103560301 on Build GRCh38
rs768693021 Chr.14: 103560465-103560465 on Build GRCh38
rs780114319 Chr.14: 103560318-103560318 on Build GRCh38
rs921759055 Chr.14: 103560370-103560370 on Build GRCh38
rs768889647 Chr.14: 103560320-103560320 on Build GRCh38
rs1014753951 Chr.14: 103560245-103560245 on Build GRCh38
rs770847049 Chr.14: 103560311-103560311 on Build GRCh38
rs372348583 Chr.14: 103560441-103560441 on Build GRCh38
rs972727028 Chr.14: 103560416-103560416 on Build GRCh38
rs372338173 Chr.14: 103560400-103560400 on Build GRCh38
rs777446271 Chr.14: 103560430-103560430 on Build GRCh38
rs1019682000 Chr.14: 103560382-103560382 on Build GRCh38
rs749328414 Chr.14: 103560319-103560319 on Build GRCh38
rs778658043 Chr.14: 103560286-103560286 on Build GRCh38
rs141215432 Chr.14: 103560461-103560461 on Build GRCh38
rs777828990 Chr.14: 103560305-103560305 on Build GRCh38
rs151161046 Chr.14: 103560408-103560408 on Build GRCh38
rs753044829 Chr.14: 103560330-103560330 on Build GRCh38
rs753418462 Chr.14: 103560376-103560376 on Build GRCh38
rs751304561 Chr.14: 103560432-103560432 on Build GRCh38
rs201699944 Chr.14: 103560466-103560466 on Build GRCh38
rs746911735 Chr.14: 103560308-103560308 on Build GRCh38
rs752283078 Chr.14: 103560415-103560415 on Build GRCh38
rs762278649 Chr.14: 103560335-103560335 on Build GRCh38
rs772310219 Chr.14: 103560341-103560341 on Build GRCh38
rs752583831 Chr.14: 103560299-103560299 on Build GRCh38
rs963271892 Chr.14: 103560256-103560256 on Build GRCh38
rs374522616 Chr.14: 103560330-103560330 on Build GRCh38
rs749420089 Chr.14: 103560462-103560462 on Build GRCh38
rs756420106 Chr.14: 103560426-103560428 on Build GRCh38

View Full Product Details