Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 18057427-18057674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAH1
Entrez Gene ID
427
Gene Name
N-acylsphingosine amidohydrolase 1
Gene Aliases
AC, ACDase, ASAH, PHP, PHP32, SMAPME
UniGene
Hs.527412
Gene Chromosome Location
Chr.8: 18056299-18084998 on Build GRCh38
Transcripts
NM_001127505.2 NM_004315.5 NM_177924.4 XM_005273504.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4654357 c.1199A>G p.Y400C Chr.8: 18057571-18057571 on Build GRCh38
3898566 c.1095A>G p.T365T Chr.8: 18057609-18057609 on Build GRCh38
3675085 c.1205G>T p.R402L Chr.8: 18057565-18057565 on Build GRCh38
2873541 c.1102A>G p.I368V Chr.8: 18057602-18057602 on Build GRCh38
5475160 c.1144G>A p.E382K Chr.8: 18057578-18057578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916778796 Chr.8: 18057515-18057515 on Build GRCh38
rs775704760 Chr.8: 18057640-18057640 on Build GRCh38
rs374718391 Chr.8: 18057497-18057497 on Build GRCh38
rs748511962 Chr.8: 18057484-18057484 on Build GRCh38
rs769587137 Chr.8: 18057565-18057565 on Build GRCh38
rs778324510 Chr.8: 18057490-18057490 on Build GRCh38
rs767698652 Chr.8: 18057514-18057514 on Build GRCh38
rs759825713 Chr.8: 18057513-18057513 on Build GRCh38
rs943868608 Chr.8: 18057595-18057595 on Build GRCh38
rs372186704 Chr.8: 18057532-18057532 on Build GRCh38
rs369660352 Chr.8: 18057518-18057518 on Build GRCh38
rs200503522 Chr.8: 18057528-18057528 on Build GRCh38
rs761017863 Chr.8: 18057641-18057641 on Build GRCh38
rs757860989 Chr.8: 18057542-18057542 on Build GRCh38
rs754140197 Chr.8: 18057603-18057603 on Build GRCh38
rs773149410 Chr.8: 18057507-18057507 on Build GRCh38
rs373012279 Chr.8: 18057553-18057553 on Build GRCh38
rs756453402 Chr.8: 18057521-18057521 on Build GRCh38
rs905502103 Chr.8: 18057636-18057636 on Build GRCh38
rs148105771 Chr.8: 18057619-18057619 on Build GRCh38
rs372661447 Chr.8: 18057618-18057618 on Build GRCh38
rs773025886 Chr.8: 18057566-18057566 on Build GRCh38
rs746513660 Chr.8: 18057547-18057547 on Build GRCh38
rs376552739 Chr.8: 18057635-18057635 on Build GRCh38
rs368345612 Chr.8: 18057568-18057568 on Build GRCh38
rs759594722 Chr.8: 18057582-18057582 on Build GRCh38
rs556143081 Chr.8: 18057476-18057476 on Build GRCh38
rs370106165 Chr.8: 18057612-18057612 on Build GRCh38
rs779529827 Chr.8: 18057610-18057610 on Build GRCh38
rs182371905 Chr.8: 18057510-18057510 on Build GRCh38
rs764463279 Chr.8: 18057602-18057602 on Build GRCh38
rs376831762 Chr.8: 18057544-18057544 on Build GRCh38
rs770955290 Chr.8: 18057624-18057624 on Build GRCh38
rs751629328 Chr.8: 18057512-18057512 on Build GRCh38
rs201151177 Chr.8: 18057608-18057608 on Build GRCh38
rs756656718 Chr.8: 18057489-18057489 on Build GRCh38
rs888031541 Chr.8: 18057462-18057462 on Build GRCh38
rs375983141 Chr.8: 18057579-18057579 on Build GRCh38
rs772074338 Chr.8: 18057639-18057639 on Build GRCh38
rs768462100 Chr.8: 18057550-18057550 on Build GRCh38
rs770772909 Chr.8: 18057569-18057569 on Build GRCh38
rs548760913 Chr.8: 18057585-18057585 on Build GRCh38
rs753111968 Chr.8: 18057517-18057517 on Build GRCh38
rs190110128 Chr.8: 18057500-18057500 on Build GRCh38
rs879107867 Chr.8: 18057556-18057556 on Build GRCh38
rs148976489 Chr.8: 18057578-18057578 on Build GRCh38
rs568755414 Chr.8: 18057504-18057504 on Build GRCh38
rs371008353 Chr.8: 18057509-18057509 on Build GRCh38
rs138912339 Chr.8: 18057563-18057563 on Build GRCh38
rs746603674 Chr.8: 18057499-18057499 on Build GRCh38
rs1046087544 Chr.8: 18057527-18057527 on Build GRCh38
rs947753778 Chr.8: 18057520-18057520 on Build GRCh38
rs935049109 Chr.8: 18057545-18057545 on Build GRCh38
rs781019071 Chr.8: 18057615-18057615 on Build GRCh38
rs775010336 Chr.8: 18057498-18057498 on Build GRCh38
rs200054096 Chr.8: 18057598-18057598 on Build GRCh38
rs774226930 Chr.8: 18057631-18057631 on Build GRCh38
rs544879078 Chr.8: 18057506-18057506 on Build GRCh38
rs17636067 Chr.8: 18057617-18057617 on Build GRCh38

View Full Product Details