Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35864210-35864483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEATR9
Entrez Gene ID
256957
Gene Name
HEAT repeat containing 9
Gene Aliases
C17orf66
UniGene
Hs.514820
Gene Chromosome Location
Chr.17: 35854946-35868891 on Build GRCh38
Transcripts
NM_001321395.1 NM_152781.3 NR_135630.1 XM_005278233.2 XM_005278235.1 XM_006721802.3 XM_011524586.2 XM_017024424.1

SNP Detail

SNP ID Chromosome Location
rs2291299 Chr.17: 35864402-35864402 on Build GRCh38
rs771812903 Chr.17: 35864436-35864436 on Build GRCh38
rs761221303 Chr.17: 35864457-35864458 on Build GRCh38
rs371185832 Chr.17: 35864256-35864256 on Build GRCh38
rs1049314175 Chr.17: 35864260-35864260 on Build GRCh38
rs751200279 Chr.17: 35864447-35864447 on Build GRCh38
rs930829983 Chr.17: 35864250-35864250 on Build GRCh38
rs768820908 Chr.17: 35864287-35864287 on Build GRCh38
rs534150583 Chr.17: 35864401-35864401 on Build GRCh38
rs370184768 Chr.17: 35864333-35864333 on Build GRCh38
rs777466904 Chr.17: 35864252-35864252 on Build GRCh38
rs377744012 Chr.17: 35864372-35864372 on Build GRCh38
rs191363860 Chr.17: 35864320-35864320 on Build GRCh38
rs758134448 Chr.17: 35864243-35864243 on Build GRCh38
rs1053280776 Chr.17: 35864231-35864231 on Build GRCh38
rs760430325 Chr.17: 35864294-35864294 on Build GRCh38
rs144324311 Chr.17: 35864295-35864295 on Build GRCh38
rs374346786 Chr.17: 35864354-35864354 on Build GRCh38
rs1020690591 Chr.17: 35864338-35864338 on Build GRCh38
rs576032774 Chr.17: 35864384-35864384 on Build GRCh38
rs548112802 Chr.17: 35864276-35864276 on Build GRCh38
rs889400372 Chr.17: 35864265-35864265 on Build GRCh38
rs991462361 Chr.17: 35864375-35864375 on Build GRCh38
rs1033595573 Chr.17: 35864365-35864365 on Build GRCh38
rs371873648 Chr.17: 35864347-35864347 on Build GRCh38
rs775553290 Chr.17: 35864278-35864278 on Build GRCh38
rs771294757 Chr.17: 35864270-35864270 on Build GRCh38
rs571714369 Chr.17: 35864418-35864418 on Build GRCh38
rs772751042 Chr.17: 35864237-35864238 on Build GRCh38
rs372258896 Chr.17: 35864421-35864421 on Build GRCh38
rs765204737 Chr.17: 35864341-35864341 on Build GRCh38
rs375623449 Chr.17: 35864351-35864351 on Build GRCh38
rs774631439 Chr.17: 35864293-35864293 on Build GRCh38
rs919374538 Chr.17: 35864425-35864425 on Build GRCh38
rs139294415 Chr.17: 35864420-35864420 on Build GRCh38
rs776160203 Chr.17: 35864448-35864449 on Build GRCh38
rs141245415 Chr.17: 35864286-35864286 on Build GRCh38
rs368070140 Chr.17: 35864340-35864340 on Build GRCh38
rs893335233 Chr.17: 35864245-35864245 on Build GRCh38
rs766177309 Chr.17: 35864306-35864306 on Build GRCh38
rs570995588 Chr.17: 35864277-35864277 on Build GRCh38

View Full Product Details