Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 104034164-104034389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL17A1
Entrez Gene ID
1308
Gene Name
collagen type XVII alpha 1
Gene Aliases
BA16H23.2, BP180, BPA-2, BPAG2, ERED, LAD-1
UniGene
Hs.117938
Gene Chromosome Location
Chr.10: 104031288-104085880 on Build GRCh38
Transcripts
NM_000494.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6433412 c.3907C>T p.R1303W Chr.10: 104034194-104034194 on Build GRCh38
6632577 c.3816G>A p.P1272P Chr.10: 104034285-104034285 on Build GRCh38
4638665 c.3798C>T p.P1266P Chr.10: 104034303-104034303 on Build GRCh38
4631933 c.3873T>C p.G1291G Chr.10: 104034228-104034228 on Build GRCh38
2060196 c.3800delC p.P1267fs*45 Chr.10: 104034301-104034301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763644228 Chr.10: 104034235-104034236 on Build GRCh38
rs779043586 Chr.10: 104034285-104034285 on Build GRCh38
rs749875359 Chr.10: 104034329-104034329 on Build GRCh38
rs936464293 Chr.10: 104034353-104034353 on Build GRCh38
rs756973019 Chr.10: 104034300-104034301 on Build GRCh38
rs121912771 Chr.10: 104034193-104034193 on Build GRCh38
rs756783234 Chr.10: 104034228-104034228 on Build GRCh38
rs886046684 Chr.10: 104034276-104034276 on Build GRCh38
rs771509812 Chr.10: 104034311-104034311 on Build GRCh38
rs760163289 Chr.10: 104034340-104034340 on Build GRCh38
rs201407886 Chr.10: 104034214-104034214 on Build GRCh38
rs770168630 Chr.10: 104034323-104034323 on Build GRCh38
rs766961106 Chr.10: 104034272-104034272 on Build GRCh38
rs754779591 Chr.10: 104034360-104034360 on Build GRCh38
rs759888854 Chr.10: 104034322-104034322 on Build GRCh38
rs767016278 Chr.10: 104034326-104034326 on Build GRCh38
rs751058779 Chr.10: 104034273-104034274 on Build GRCh38
rs1052194032 Chr.10: 104034358-104034358 on Build GRCh38
rs749116147 Chr.10: 104034185-104034185 on Build GRCh38
rs752418541 Chr.10: 104034205-104034205 on Build GRCh38
rs374031668 Chr.10: 104034202-104034202 on Build GRCh38
rs772301107 Chr.10: 104034287-104034287 on Build GRCh38
rs749528648 Chr.10: 104034236-104034236 on Build GRCh38
rs772247816 Chr.10: 104034248-104034248 on Build GRCh38
rs781143436 Chr.10: 104034229-104034229 on Build GRCh38
rs779836450 Chr.10: 104034278-104034278 on Build GRCh38
rs745779563 Chr.10: 104034232-104034232 on Build GRCh38
rs181559014 Chr.10: 104034324-104034324 on Build GRCh38
rs986793152 Chr.10: 104034314-104034314 on Build GRCh38
rs750388712 Chr.10: 104034274-104034274 on Build GRCh38
rs751224626 Chr.10: 104034261-104034261 on Build GRCh38
rs766292786 Chr.10: 104034344-104034344 on Build GRCh38
rs112324301 Chr.10: 104034335-104034335 on Build GRCh38
rs762683429 Chr.10: 104034253-104034253 on Build GRCh38
rs956376879 Chr.10: 104034210-104034210 on Build GRCh38
rs138210414 Chr.10: 104034241-104034241 on Build GRCh38
rs753777678 Chr.10: 104034349-104034349 on Build GRCh38
rs527309056 Chr.10: 104034341-104034341 on Build GRCh38
rs1039207664 Chr.10: 104034363-104034363 on Build GRCh38
rs150432542 Chr.10: 104034233-104034233 on Build GRCh38
rs372395647 Chr.10: 104034249-104034249 on Build GRCh38
rs763668125 Chr.10: 104034220-104034220 on Build GRCh38
rs368630310 Chr.10: 104034286-104034286 on Build GRCh38
rs576816351 Chr.10: 104034262-104034262 on Build GRCh38
rs776796314 Chr.10: 104034252-104034252 on Build GRCh38
rs754797158 Chr.10: 104034284-104034284 on Build GRCh38
rs747525421 Chr.10: 104034310-104034310 on Build GRCh38
rs367870580 Chr.10: 104034244-104034244 on Build GRCh38
rs761162295 Chr.10: 104034194-104034194 on Build GRCh38
rs777962104 Chr.10: 104034307-104034307 on Build GRCh38
rs541222706 Chr.10: 104034235-104034235 on Build GRCh38
rs903694312 Chr.10: 104034344-104034345 on Build GRCh38
rs563128052 Chr.10: 104034237-104034237 on Build GRCh38
rs750979545 Chr.10: 104034222-104034222 on Build GRCh38
rs377068542 Chr.10: 104034250-104034250 on Build GRCh38
rs372244733 Chr.10: 104034313-104034313 on Build GRCh38
rs749093771 Chr.10: 104034280-104034280 on Build GRCh38

View Full Product Details