Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46016091-46016320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC61
Entrez Gene ID
729440
Gene Name
coiled-coil domain containing 61
Gene Aliases
-
UniGene
Hs.515479
Gene Chromosome Location
Chr.19: 45995457-46018616 on Build GRCh38
Transcripts
NM_001267723.1 XM_005259191.3 XM_005259192.4 XM_011527257.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
998366 c.1072+1G>A p.? Chr.19: 46016224-46016224 on Build GRCh38
1612430 c.1034G>T p.G345V Chr.19: 46016185-46016185 on Build GRCh38
6435626 c.1043G>A p.R348H Chr.19: 46016194-46016194 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs377131956 Chr.19: 46016264-46016264 on Build GRCh38
rs749845324 Chr.19: 46016269-46016269 on Build GRCh38
rs779851134 Chr.19: 46016209-46016209 on Build GRCh38
rs765146197 Chr.19: 46016200-46016200 on Build GRCh38
rs888792431 Chr.19: 46016217-46016217 on Build GRCh38
rs756212235 Chr.19: 46016183-46016183 on Build GRCh38
rs538643382 Chr.19: 46016146-46016146 on Build GRCh38
rs1050009299 Chr.19: 46016215-46016215 on Build GRCh38
rs935494298 Chr.19: 46016163-46016163 on Build GRCh38
rs113745251 Chr.19: 46016148-46016148 on Build GRCh38
rs201360874 Chr.19: 46016294-46016294 on Build GRCh38
rs369340867 Chr.19: 46016274-46016274 on Build GRCh38
rs574291544 Chr.19: 46016296-46016296 on Build GRCh38
rs755686232 Chr.19: 46016303-46016303 on Build GRCh38
rs894026323 Chr.19: 46016178-46016178 on Build GRCh38
rs758226056 Chr.19: 46016206-46016206 on Build GRCh38
rs556156591 Chr.19: 46016107-46016124 on Build GRCh38
rs570212080 Chr.19: 46016131-46016131 on Build GRCh38
rs892701397 Chr.19: 46016289-46016289 on Build GRCh38
rs78197451 Chr.19: 46016165-46016165 on Build GRCh38
rs754597762 Chr.19: 46016220-46016220 on Build GRCh38
rs766244815 Chr.19: 46016288-46016288 on Build GRCh38
rs760716968 Chr.19: 46016278-46016278 on Build GRCh38
rs1008553242 Chr.19: 46016222-46016222 on Build GRCh38
rs780759823 Chr.19: 46016297-46016297 on Build GRCh38
rs775766359 Chr.19: 46016277-46016277 on Build GRCh38
rs747672280 Chr.19: 46016234-46016234 on Build GRCh38
rs753870369 Chr.19: 46016283-46016283 on Build GRCh38
rs762895994 Chr.19: 46016284-46016284 on Build GRCh38
rs769329471 Chr.19: 46016243-46016243 on Build GRCh38
rs764201706 Chr.19: 46016280-46016280 on Build GRCh38
rs773571825 Chr.19: 46016149-46016149 on Build GRCh38
rs372618168 Chr.19: 46016211-46016211 on Build GRCh38
rs369281254 Chr.19: 46016203-46016203 on Build GRCh38
rs780895662 Chr.19: 46016221-46016221 on Build GRCh38
rs534397820 Chr.19: 46016273-46016273 on Build GRCh38
rs1018697375 Chr.19: 46016237-46016237 on Build GRCh38
rs933611768 Chr.19: 46016238-46016238 on Build GRCh38
rs916603518 Chr.19: 46016139-46016139 on Build GRCh38

View Full Product Details